Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219540009-219540506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF
Entrez Gene ID
79586
Gene Name
chondroitin polymerizing factor
Gene Aliases
CHSY2, CSS2
UniGene
Hs.516711
Gene Chromosome Location
Chr.2: 219538947-219543765 on Build GRCh38
Transcripts
NM_001195731.1 NM_024536.5 XM_011511838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6304984 c.1247G>T p.G416V Chr.2: 219540464-219540464 on Build GRCh38
6628552 c.1678G>A p.A560T Chr.2: 219540033-219540033 on Build GRCh38
1565058 c.1338G>A p.Q446Q Chr.2: 219540373-219540373 on Build GRCh38
6628551 c.1436G>T p.R479L Chr.2: 219540275-219540275 on Build GRCh38
4592919 c.1277T>G p.L426R Chr.2: 219540434-219540434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769313279 Chr.2: 219540339-219540339 on Build GRCh38
rs768978070 Chr.2: 219540167-219540167 on Build GRCh38
rs1038876666 Chr.2: 219540318-219540318 on Build GRCh38
rs765155405 Chr.2: 219540456-219540456 on Build GRCh38
rs557875110 Chr.2: 219540468-219540468 on Build GRCh38
rs377089380 Chr.2: 219540059-219540059 on Build GRCh38
rs138908715 Chr.2: 219540211-219540211 on Build GRCh38
rs150325362 Chr.2: 219540186-219540186 on Build GRCh38
rs754703207 Chr.2: 219540091-219540091 on Build GRCh38
rs771626436 Chr.2: 219540260-219540260 on Build GRCh38
rs768694057 Chr.2: 219540235-219540235 on Build GRCh38
rs140166294 Chr.2: 219540405-219540405 on Build GRCh38
rs772950229 Chr.2: 219540419-219540419 on Build GRCh38
rs773005895 Chr.2: 219540481-219540481 on Build GRCh38
rs150850620 Chr.2: 219540387-219540387 on Build GRCh38
rs745894220 Chr.2: 219540152-219540152 on Build GRCh38
rs775950895 Chr.2: 219540263-219540263 on Build GRCh38
rs763280868 Chr.2: 219540057-219540057 on Build GRCh38
rs750764885 Chr.2: 219540467-219540467 on Build GRCh38
rs778272074 Chr.2: 219540319-219540319 on Build GRCh38
rs149456271 Chr.2: 219540205-219540205 on Build GRCh38
rs949661363 Chr.2: 219540282-219540282 on Build GRCh38
rs200369127 Chr.2: 219540252-219540252 on Build GRCh38
rs764366642 Chr.2: 219540447-219540447 on Build GRCh38
rs765355562 Chr.2: 219540377-219540377 on Build GRCh38
rs778435737 Chr.2: 219540473-219540473 on Build GRCh38
rs1002212562 Chr.2: 219540330-219540330 on Build GRCh38
rs771279965 Chr.2: 219540320-219540320 on Build GRCh38
rs777590978 Chr.2: 219540220-219540220 on Build GRCh38
rs199870967 Chr.2: 219540289-219540289 on Build GRCh38
rs758638648 Chr.2: 219540384-219540384 on Build GRCh38
rs755717664 Chr.2: 219540076-219540076 on Build GRCh38
rs777229183 Chr.2: 219540321-219540321 on Build GRCh38
rs145792368 Chr.2: 219540408-219540408 on Build GRCh38
rs368679810 Chr.2: 219540246-219540246 on Build GRCh38
rs1024562539 Chr.2: 219540151-219540151 on Build GRCh38
rs746407058 Chr.2: 219540335-219540335 on Build GRCh38
rs760395442 Chr.2: 219540485-219540485 on Build GRCh38
rs1010437879 Chr.2: 219540358-219540358 on Build GRCh38
rs201075985 Chr.2: 219540035-219540035 on Build GRCh38
rs146854124 Chr.2: 219540163-219540163 on Build GRCh38
rs752391344 Chr.2: 219540213-219540213 on Build GRCh38
rs907591853 Chr.2: 219540465-219540465 on Build GRCh38
rs572962492 Chr.2: 219540386-219540386 on Build GRCh38
rs748037251 Chr.2: 219540099-219540099 on Build GRCh38
rs986801190 Chr.2: 219540232-219540232 on Build GRCh38
rs774521625 Chr.2: 219540177-219540177 on Build GRCh38
rs749888212 Chr.2: 219540075-219540075 on Build GRCh38
rs778571617 Chr.2: 219540092-219540092 on Build GRCh38
rs758207651 Chr.2: 219540219-219540219 on Build GRCh38
rs773753712 Chr.2: 219540247-219540247 on Build GRCh38
rs773878341 Chr.2: 219540191-219540191 on Build GRCh38
rs538123825 Chr.2: 219540288-219540288 on Build GRCh38
rs765957822 Chr.2: 219540199-219540199 on Build GRCh38
rs371775506 Chr.2: 219540353-219540353 on Build GRCh38
rs774921566 Chr.2: 219540398-219540398 on Build GRCh38
rs745949162 Chr.2: 219540044-219540044 on Build GRCh38
rs779228805 Chr.2: 219540241-219540241 on Build GRCh38
rs144193296 Chr.2: 219540404-219540404 on Build GRCh38
rs200411864 Chr.2: 219540155-219540155 on Build GRCh38
rs200725116 Chr.2: 219540279-219540279 on Build GRCh38
rs764333095 Chr.2: 219540058-219540058 on Build GRCh38
rs761315379 Chr.2: 219540192-219540192 on Build GRCh38
rs766436141 Chr.2: 219540441-219540441 on Build GRCh38
rs906087063 Chr.2: 219540327-219540327 on Build GRCh38
rs540827592 Chr.2: 219540395-219540395 on Build GRCh38
rs201025089 Chr.2: 219540359-219540359 on Build GRCh38
rs372841467 Chr.2: 219540043-219540043 on Build GRCh38
rs145738935 Chr.2: 219540455-219540455 on Build GRCh38
rs1045433928 Chr.2: 219540298-219540298 on Build GRCh38
rs750352855 Chr.2: 219540366-219540366 on Build GRCh38
rs142947245 Chr.2: 219540439-219540439 on Build GRCh38
rs770955890 Chr.2: 219540487-219540487 on Build GRCh38
rs749399783 Chr.2: 219540234-219540234 on Build GRCh38
rs147396475 Chr.2: 219540275-219540275 on Build GRCh38
rs757250970 Chr.2: 219540231-219540231 on Build GRCh38
rs763868875 Chr.2: 219540274-219540274 on Build GRCh38
rs758277175 Chr.2: 219540105-219540105 on Build GRCh38
rs748351385 Chr.2: 219540293-219540293 on Build GRCh38
rs201029716 Chr.2: 219540264-219540264 on Build GRCh38
rs112268222 Chr.2: 219540278-219540278 on Build GRCh38
rs764743400 Chr.2: 219540265-219540265 on Build GRCh38
rs775044196 Chr.2: 219540340-219540340 on Build GRCh38
rs758761506 Chr.2: 219540311-219540311 on Build GRCh38
rs768149745 Chr.2: 219540072-219540072 on Build GRCh38
rs754751372 Chr.2: 219540034-219540034 on Build GRCh38
rs776122883 Chr.2: 219540276-219540276 on Build GRCh38
rs746493090 Chr.2: 219540144-219540144 on Build GRCh38
rs748475880 Chr.2: 219540245-219540245 on Build GRCh38
rs11556306 Chr.2: 219540351-219540351 on Build GRCh38
rs960827873 Chr.2: 219540182-219540182 on Build GRCh38
rs1020395562 Chr.2: 219540193-219540193 on Build GRCh38
rs766081327 Chr.2: 219540078-219540078 on Build GRCh38
rs370130402 Chr.2: 219540088-219540088 on Build GRCh38
rs764960369 Chr.2: 219540212-219540212 on Build GRCh38
rs762631718 Chr.2: 219540272-219540272 on Build GRCh38
rs533280206 Chr.2: 219540042-219540042 on Build GRCh38
rs757616174 Chr.2: 219540390-219540390 on Build GRCh38
rs769960716 Chr.2: 219540050-219540050 on Build GRCh38
rs774014211 Chr.2: 219540407-219540407 on Build GRCh38
rs775706899 Chr.2: 219540056-219540056 on Build GRCh38
rs761626632 Chr.2: 219540357-219540357 on Build GRCh38
rs200756992 Chr.2: 219540369-219540369 on Build GRCh38
rs781445929 Chr.2: 219540391-219540391 on Build GRCh38
rs746379463 Chr.2: 219540392-219540392 on Build GRCh38
rs115884312 Chr.2: 219540472-219540472 on Build GRCh38
rs866240988 Chr.2: 219540242-219540242 on Build GRCh38
rs751465265 Chr.2: 219540228-219540228 on Build GRCh38
rs752367537 Chr.2: 219540271-219540271 on Build GRCh38
rs749465783 Chr.2: 219540164-219540164 on Build GRCh38
rs575630904 Chr.2: 219540442-219540442 on Build GRCh38
rs774065443 Chr.2: 219540355-219540355 on Build GRCh38
rs138114486 Chr.2: 219540479-219540479 on Build GRCh38
rs752885494 Chr.2: 219540378-219540378 on Build GRCh38
rs747834322 Chr.2: 219540478-219540478 on Build GRCh38
rs867030173 Chr.2: 219540129-219540129 on Build GRCh38
rs777715508 Chr.2: 219540116-219540116 on Build GRCh38
rs772667451 Chr.2: 219540189-219540189 on Build GRCh38
rs779983221 Chr.2: 219540233-219540233 on Build GRCh38
rs145096722 Chr.2: 219540156-219540156 on Build GRCh38
rs767313477 Chr.2: 219540412-219540412 on Build GRCh38
rs758585887 Chr.2: 219540446-219540446 on Build GRCh38

View Full Product Details