Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219541913-219542423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF
Entrez Gene ID
79586
Gene Name
chondroitin polymerizing factor
Gene Aliases
CHSY2, CSS2
UniGene
Hs.516711
Gene Chromosome Location
Chr.2: 219538947-219543765 on Build GRCh38
Transcripts
NM_001195731.1 NM_024536.5 XM_011511838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407078 c.439C>T p.R147C Chr.2: 219542065-219542065 on Build GRCh38
4091734 c.358C>T p.L120L Chr.2: 219542146-219542146 on Build GRCh38
319445 c.424G>A p.G142R Chr.2: 219542080-219542080 on Build GRCh38
6342633 c.528C>A p.H176Q Chr.2: 219541976-219541976 on Build GRCh38
3798667 c.571G>A p.D191N Chr.2: 219541933-219541933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897601690 Chr.2: 219542283-219542283 on Build GRCh38
rs757737130 Chr.2: 219541976-219541976 on Build GRCh38
rs894386675 Chr.2: 219542048-219542048 on Build GRCh38
rs752889377 Chr.2: 219542040-219542040 on Build GRCh38
rs778460864 Chr.2: 219542131-219542131 on Build GRCh38
rs938036745 Chr.2: 219541978-219541978 on Build GRCh38
rs772005890 Chr.2: 219542031-219542031 on Build GRCh38
rs570337446 Chr.2: 219542371-219542371 on Build GRCh38
rs781584670 Chr.2: 219541981-219541981 on Build GRCh38
rs982177811 Chr.2: 219542261-219542261 on Build GRCh38
rs760742426 Chr.2: 219542033-219542033 on Build GRCh38
rs1024774602 Chr.2: 219542339-219542339 on Build GRCh38
rs750792607 Chr.2: 219542232-219542232 on Build GRCh38
rs377629763 Chr.2: 219541936-219541936 on Build GRCh38
rs571316585 Chr.2: 219542032-219542032 on Build GRCh38
rs369689645 Chr.2: 219541990-219541990 on Build GRCh38
rs35657922 Chr.2: 219542189-219542189 on Build GRCh38
rs984276245 Chr.2: 219542236-219542236 on Build GRCh38
rs774416614 Chr.2: 219542229-219542229 on Build GRCh38
rs751916266 Chr.2: 219542067-219542067 on Build GRCh38
rs765392601 Chr.2: 219542036-219542036 on Build GRCh38
rs1036014128 Chr.2: 219542269-219542269 on Build GRCh38
rs781660239 Chr.2: 219542078-219542078 on Build GRCh38
rs767865008 Chr.2: 219542231-219542231 on Build GRCh38
rs1013262870 Chr.2: 219542329-219542329 on Build GRCh38
rs764279137 Chr.2: 219542227-219542227 on Build GRCh38
rs763197544 Chr.2: 219542053-219542053 on Build GRCh38
rs376366880 Chr.2: 219542074-219542074 on Build GRCh38
rs1055545116 Chr.2: 219542313-219542313 on Build GRCh38
rs752942735 Chr.2: 219541942-219541942 on Build GRCh38
rs371065957 Chr.2: 219541939-219541939 on Build GRCh38
rs759718223 Chr.2: 219541937-219541937 on Build GRCh38
rs902919631 Chr.2: 219542093-219542093 on Build GRCh38
rs760643891 Chr.2: 219542377-219542377 on Build GRCh38
rs373330576 Chr.2: 219542129-219542129 on Build GRCh38
rs7581251 Chr.2: 219542222-219542222 on Build GRCh38
rs758749064 Chr.2: 219541966-219541966 on Build GRCh38
rs947312471 Chr.2: 219542338-219542338 on Build GRCh38
rs556113650 Chr.2: 219542234-219542234 on Build GRCh38
rs928066185 Chr.2: 219542293-219542293 on Build GRCh38
rs998680009 Chr.2: 219542401-219542401 on Build GRCh38
rs761535669 Chr.2: 219542024-219542024 on Build GRCh38
rs768199469 Chr.2: 219542011-219542011 on Build GRCh38
rs774138074 Chr.2: 219542016-219542016 on Build GRCh38
rs3731912 Chr.2: 219542000-219542000 on Build GRCh38
rs746416855 Chr.2: 219541982-219541982 on Build GRCh38
rs748812043 Chr.2: 219542096-219542096 on Build GRCh38
rs937988865 Chr.2: 219542304-219542304 on Build GRCh38
rs778191760 Chr.2: 219541970-219541970 on Build GRCh38
rs752637660 Chr.2: 219542234-219542234 on Build GRCh38
rs746936842 Chr.2: 219542199-219542199 on Build GRCh38
rs188641011 Chr.2: 219542390-219542390 on Build GRCh38
rs773197011 Chr.2: 219542194-219542194 on Build GRCh38
rs756556391 Chr.2: 219542237-219542237 on Build GRCh38
rs567212015 Chr.2: 219542217-219542217 on Build GRCh38
rs576501247 Chr.2: 219542241-219542241 on Build GRCh38
rs1011376537 Chr.2: 219542069-219542069 on Build GRCh38
rs750919192 Chr.2: 219542084-219542084 on Build GRCh38
rs369325931 Chr.2: 219542088-219542088 on Build GRCh38
rs762156120 Chr.2: 219542230-219542230 on Build GRCh38
rs771741638 Chr.2: 219542167-219542167 on Build GRCh38
rs538724051 Chr.2: 219542337-219542337 on Build GRCh38
rs764232533 Chr.2: 219542065-219542065 on Build GRCh38
rs775650833 Chr.2: 219542221-219542221 on Build GRCh38
rs373176396 Chr.2: 219541991-219541991 on Build GRCh38
rs747873112 Chr.2: 219542160-219542160 on Build GRCh38
rs867711639 Chr.2: 219542175-219542175 on Build GRCh38
rs888701327 Chr.2: 219542203-219542203 on Build GRCh38
rs748936818 Chr.2: 219542003-219542003 on Build GRCh38
rs771655201 Chr.2: 219542270-219542270 on Build GRCh38
rs769273874 Chr.2: 219541983-219541983 on Build GRCh38
rs768536819 Chr.2: 219542378-219542378 on Build GRCh38
rs751870150 Chr.2: 219541975-219541975 on Build GRCh38
rs779474308 Chr.2: 219542089-219542089 on Build GRCh38
rs996956883 Chr.2: 219542211-219542211 on Build GRCh38

View Full Product Details