Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222571718-222572225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSB
Entrez Gene ID
10056
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Gene Aliases
FARSLB, FRSB, HSPC173, PheHB, PheRS
UniGene
Hs.471452
Gene Chromosome Location
Chr.2: 222571443-222656355 on Build GRCh38
Transcripts
NM_005687.4 NR_130154.1 XM_006712169.2 XM_011510466.2 XM_017003110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7206059 c.1626T>C p.A542A Chr.2: 222572015-222572015 on Build GRCh38
3798676 c.1693C>T p.H565Y Chr.2: 222571948-222571948 on Build GRCh38
4470313 c.1653C>T p.I551I Chr.2: 222571988-222571988 on Build GRCh38
4437035 c.1731C>T p.P577P Chr.2: 222571910-222571910 on Build GRCh38
720549 c.1701C>T p.D567D Chr.2: 222571940-222571940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006535933 Chr.2: 222572088-222572088 on Build GRCh38
rs535728309 Chr.2: 222571954-222571954 on Build GRCh38
rs775673628 Chr.2: 222571860-222571860 on Build GRCh38
rs1014983674 Chr.2: 222571779-222571779 on Build GRCh38
rs969358399 Chr.2: 222571911-222571911 on Build GRCh38
rs200455799 Chr.2: 222571938-222571938 on Build GRCh38
rs950041375 Chr.2: 222572097-222572097 on Build GRCh38
rs894452569 Chr.2: 222572089-222572089 on Build GRCh38
rs768843030 Chr.2: 222571973-222571973 on Build GRCh38
rs763107652 Chr.2: 222571970-222571970 on Build GRCh38
rs530056752 Chr.2: 222571978-222571978 on Build GRCh38
rs746775581 Chr.2: 222572062-222572062 on Build GRCh38
rs777665977 Chr.2: 222571950-222571950 on Build GRCh38
rs369687965 Chr.2: 222571831-222571831 on Build GRCh38
rs1033255916 Chr.2: 222571846-222571846 on Build GRCh38
rs182018748 Chr.2: 222572156-222572156 on Build GRCh38
rs770904791 Chr.2: 222572065-222572065 on Build GRCh38
rs748777302 Chr.2: 222571900-222571900 on Build GRCh38
rs766444416 Chr.2: 222571829-222571829 on Build GRCh38
rs758835288 Chr.2: 222571852-222571852 on Build GRCh38
rs189464589 Chr.2: 222572153-222572153 on Build GRCh38
rs776755911 Chr.2: 222571969-222571969 on Build GRCh38
rs758031471 Chr.2: 222572012-222572012 on Build GRCh38
rs184700021 Chr.2: 222572137-222572137 on Build GRCh38
rs559887916 Chr.2: 222572122-222572122 on Build GRCh38
rs7185 Chr.2: 222571888-222571888 on Build GRCh38
rs893321092 Chr.2: 222572142-222572142 on Build GRCh38
rs3768976 Chr.2: 222572141-222572141 on Build GRCh38
rs753794587 Chr.2: 222572108-222572108 on Build GRCh38
rs778426408 Chr.2: 222571924-222571924 on Build GRCh38
rs192201095 Chr.2: 222572101-222572101 on Build GRCh38
rs752060290 Chr.2: 222572058-222572058 on Build GRCh38
rs1802198 Chr.2: 222572001-222572001 on Build GRCh38
rs767855853 Chr.2: 222571882-222571882 on Build GRCh38
rs553112461 Chr.2: 222571797-222571797 on Build GRCh38
rs761078821 Chr.2: 222571990-222571990 on Build GRCh38
rs561882024 Chr.2: 222571908-222571908 on Build GRCh38
rs903298628 Chr.2: 222572104-222572104 on Build GRCh38
rs887715408 Chr.2: 222572077-222572077 on Build GRCh38
rs766858931 Chr.2: 222571998-222571998 on Build GRCh38
rs267599217 Chr.2: 222571957-222571957 on Build GRCh38
rs750775563 Chr.2: 222571824-222571825 on Build GRCh38
rs17851564 Chr.2: 222571988-222571988 on Build GRCh38
rs868589459 Chr.2: 222571945-222571945 on Build GRCh38
rs757554435 Chr.2: 222571880-222571880 on Build GRCh38
rs963642313 Chr.2: 222572197-222572197 on Build GRCh38
rs770964019 Chr.2: 222571967-222571967 on Build GRCh38
rs116655892 Chr.2: 222571867-222571867 on Build GRCh38
rs377716916 Chr.2: 222572006-222572006 on Build GRCh38
rs778432967 Chr.2: 222572005-222572005 on Build GRCh38
rs545995156 Chr.2: 222571746-222571746 on Build GRCh38
rs976436389 Chr.2: 222571943-222571943 on Build GRCh38
rs774298156 Chr.2: 222571977-222571977 on Build GRCh38
rs77827981 Chr.2: 222572000-222572000 on Build GRCh38
rs747770664 Chr.2: 222571939-222571939 on Build GRCh38
rs763162761 Chr.2: 222571862-222571862 on Build GRCh38
rs781267081 Chr.2: 222572070-222572070 on Build GRCh38
rs944178081 Chr.2: 222571975-222571975 on Build GRCh38
rs745747750 Chr.2: 222572071-222572071 on Build GRCh38
rs141988189 Chr.2: 222571884-222571884 on Build GRCh38
rs141929079 Chr.2: 222571940-222571940 on Build GRCh38
rs750743562 Chr.2: 222571984-222571984 on Build GRCh38
rs746834771 Chr.2: 222571964-222571964 on Build GRCh38
rs961850067 Chr.2: 222571803-222571803 on Build GRCh38
rs201693414 Chr.2: 222572031-222572031 on Build GRCh38
rs532447595 Chr.2: 222572076-222572076 on Build GRCh38
rs780568817 Chr.2: 222571897-222571897 on Build GRCh38
rs751862914 Chr.2: 222571876-222571876 on Build GRCh38
rs780549915 Chr.2: 222572039-222572039 on Build GRCh38
rs894207194 Chr.2: 222572179-222572179 on Build GRCh38

View Full Product Details