Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222922943-222923485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
UniGene
Hs.655772
Gene Chromosome Location
Chr.2: 222860984-222944624 on Build GRCh38
Transcripts
NM_004457.3 NM_203372.1 XM_017003584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904136 c.1081-1G>A p.? Chr.2: 222923077-222923077 on Build GRCh38
4991707 c.1103G>T p.S368I Chr.2: 222923100-222923100 on Build GRCh38
5508935 c.1152+2delT p.? Chr.2: 222923151-222923151 on Build GRCh38
242786 c.1082C>T p.S361F Chr.2: 222923079-222923079 on Build GRCh38
278599 c.1087A>C p.K363Q Chr.2: 222923084-222923084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375798869 Chr.2: 222923074-222923074 on Build GRCh38
rs997549859 Chr.2: 222923058-222923058 on Build GRCh38
rs1006829142 Chr.2: 222923115-222923115 on Build GRCh38
rs762419923 Chr.2: 222923047-222923048 on Build GRCh38
rs775251003 Chr.2: 222923114-222923114 on Build GRCh38
rs772524713 Chr.2: 222923032-222923032 on Build GRCh38
rs973838774 Chr.2: 222923187-222923187 on Build GRCh38
rs754516691 Chr.2: 222922966-222922966 on Build GRCh38
rs767629776 Chr.2: 222923150-222923150 on Build GRCh38
rs900970563 Chr.2: 222923021-222923021 on Build GRCh38
rs764004849 Chr.2: 222923130-222923130 on Build GRCh38
rs112352030 Chr.2: 222923056-222923056 on Build GRCh38
rs768200414 Chr.2: 222923018-222923018 on Build GRCh38
rs566961930 Chr.2: 222922982-222922982 on Build GRCh38
rs747303315 Chr.2: 222923197-222923197 on Build GRCh38
rs374025445 Chr.2: 222923356-222923356 on Build GRCh38
rs942365587 Chr.2: 222923402-222923402 on Build GRCh38
rs535801552 Chr.2: 222923023-222923023 on Build GRCh38
rs925488320 Chr.2: 222923346-222923350 on Build GRCh38
rs757265756 Chr.2: 222923149-222923149 on Build GRCh38
rs146287451 Chr.2: 222923291-222923291 on Build GRCh38
rs554864113 Chr.2: 222923321-222923321 on Build GRCh38
rs771365458 Chr.2: 222923064-222923064 on Build GRCh38
rs776996984 Chr.2: 222923040-222923053 on Build GRCh38
rs917502246 Chr.2: 222923323-222923323 on Build GRCh38
rs901101006 Chr.2: 222923441-222923441 on Build GRCh38
rs993005118 Chr.2: 222923296-222923296 on Build GRCh38
rs762475796 Chr.2: 222923117-222923117 on Build GRCh38
rs187358631 Chr.2: 222923280-222923280 on Build GRCh38
rs751342377 Chr.2: 222923148-222923148 on Build GRCh38
rs750314497 Chr.2: 222923155-222923155 on Build GRCh38
rs555864781 Chr.2: 222923061-222923061 on Build GRCh38
rs764633563 Chr.2: 222923100-222923100 on Build GRCh38
rs755133422 Chr.2: 222923157-222923157 on Build GRCh38
rs10932978 Chr.2: 222922973-222922973 on Build GRCh38
rs994022258 Chr.2: 222923050-222923050 on Build GRCh38
rs1025703687 Chr.2: 222923052-222923052 on Build GRCh38
rs572203101 Chr.2: 222923290-222923290 on Build GRCh38
rs910855874 Chr.2: 222923394-222923394 on Build GRCh38
rs368892612 Chr.2: 222923075-222923075 on Build GRCh38
rs933372024 Chr.2: 222923429-222923429 on Build GRCh38
rs538712684 Chr.2: 222923254-222923254 on Build GRCh38
rs914015702 Chr.2: 222923400-222923400 on Build GRCh38
rs991817118 Chr.2: 222923277-222923277 on Build GRCh38
rs569617901 Chr.2: 222923225-222923225 on Build GRCh38
rs962584503 Chr.2: 222923174-222923174 on Build GRCh38
rs989670861 Chr.2: 222923364-222923364 on Build GRCh38
rs977547088 Chr.2: 222923342-222923342 on Build GRCh38
rs373978348 Chr.2: 222923163-222923163 on Build GRCh38
rs752992653 Chr.2: 222923161-222923161 on Build GRCh38
rs756441514 Chr.2: 222923331-222923331 on Build GRCh38
rs778825017 Chr.2: 222923160-222923160 on Build GRCh38
rs951127912 Chr.2: 222923272-222923272 on Build GRCh38
rs1052522089 Chr.2: 222922996-222922996 on Build GRCh38
rs936891070 Chr.2: 222923347-222923347 on Build GRCh38
rs773522343 Chr.2: 222923053-222923053 on Build GRCh38
rs535031870 Chr.2: 222923358-222923358 on Build GRCh38
rs759147663 Chr.2: 222923095-222923095 on Build GRCh38
rs563013801 Chr.2: 222923447-222923447 on Build GRCh38
rs574085534 Chr.2: 222923332-222923332 on Build GRCh38
rs926147970 Chr.2: 222923344-222923344 on Build GRCh38
rs771277131 Chr.2: 222923198-222923198 on Build GRCh38
rs879124059 Chr.2: 222923411-222923411 on Build GRCh38

View Full Product Details