Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224770453-224770852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
UniGene
Hs.46578
Gene Chromosome Location
Chr.2: 224765090-225042689 on Build GRCh38
Transcripts
NM_001290263.1 NM_014689.2 XM_005246673.4 XM_006712617.3 XM_017004441.1 XM_017004442.1 XM_017004443.1 XM_017004444.1 XM_017004445.1 XM_017004446.1 XM_017004447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4867092 c.6223C>A p.L2075I Chr.2: 224770609-224770609 on Build GRCh38
4931437 c.6272T>G p.L2091W Chr.2: 224770560-224770560 on Build GRCh38
720507 c.1826G>A p.R609K Chr.2: 224770545-224770545 on Build GRCh38
1017133 c.6235C>A p.L2079I Chr.2: 224770609-224770609 on Build GRCh38
4931438 c.1811T>G p.L604W Chr.2: 224770560-224770560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187626634 Chr.2: 224770704-224770704 on Build GRCh38
rs190425827 Chr.2: 224770499-224770499 on Build GRCh38
rs746097994 Chr.2: 224770625-224770625 on Build GRCh38
rs377007980 Chr.2: 224770714-224770714 on Build GRCh38
rs1044644737 Chr.2: 224770621-224770621 on Build GRCh38
rs776515478 Chr.2: 224770666-224770668 on Build GRCh38
rs547840387 Chr.2: 224770744-224770744 on Build GRCh38
rs766175724 Chr.2: 224770582-224770582 on Build GRCh38
rs757333504 Chr.2: 224770495-224770495 on Build GRCh38
rs969556389 Chr.2: 224770555-224770555 on Build GRCh38
rs749654060 Chr.2: 224770652-224770652 on Build GRCh38
rs758017836 Chr.2: 224770598-224770598 on Build GRCh38
rs1005981501 Chr.2: 224770754-224770754 on Build GRCh38
rs994114911 Chr.2: 224770506-224770506 on Build GRCh38
rs1025632871 Chr.2: 224770514-224770514 on Build GRCh38
rs201549682 Chr.2: 224770558-224770558 on Build GRCh38
rs554496052 Chr.2: 224770533-224770533 on Build GRCh38
rs772883004 Chr.2: 224770565-224770565 on Build GRCh38
rs770285454 Chr.2: 224770509-224770509 on Build GRCh38
rs368336784 Chr.2: 224770617-224770617 on Build GRCh38
rs779311670 Chr.2: 224770691-224770691 on Build GRCh38
rs907372379 Chr.2: 224770628-224770628 on Build GRCh38
rs375370127 Chr.2: 224770661-224770661 on Build GRCh38
rs765132018 Chr.2: 224770605-224770605 on Build GRCh38
rs923123131 Chr.2: 224770482-224770482 on Build GRCh38
rs182634471 Chr.2: 224770568-224770568 on Build GRCh38
rs769109007 Chr.2: 224770539-224770539 on Build GRCh38
rs1024149644 Chr.2: 224770553-224770553 on Build GRCh38
rs192063573 Chr.2: 224770752-224770752 on Build GRCh38
rs147106645 Chr.2: 224770741-224770741 on Build GRCh38
rs1021235244 Chr.2: 224770747-224770747 on Build GRCh38
rs764606340 Chr.2: 224770815-224770815 on Build GRCh38
rs753694327 Chr.2: 224770586-224770586 on Build GRCh38
rs754801101 Chr.2: 224770596-224770596 on Build GRCh38
rs1023698644 Chr.2: 224770780-224770780 on Build GRCh38
rs769041405 Chr.2: 224770678-224770678 on Build GRCh38
rs1042783841 Chr.2: 224770685-224770685 on Build GRCh38
rs781352125 Chr.2: 224770619-224770619 on Build GRCh38
rs780792694 Chr.2: 224770745-224770745 on Build GRCh38
rs774891050 Chr.2: 224770540-224770540 on Build GRCh38
rs377567213 Chr.2: 224770504-224770504 on Build GRCh38
rs543390552 Chr.2: 224770703-224770703 on Build GRCh38
rs1040883744 Chr.2: 224770721-224770722 on Build GRCh38
rs901087753 Chr.2: 224770735-224770735 on Build GRCh38
rs1048075444 Chr.2: 224770651-224770651 on Build GRCh38
rs996051107 Chr.2: 224770781-224770781 on Build GRCh38
rs761278104 Chr.2: 224770543-224770543 on Build GRCh38
rs1003419550 Chr.2: 224770686-224770686 on Build GRCh38
rs370874234 Chr.2: 224770641-224770641 on Build GRCh38
rs1011222318 Chr.2: 224770726-224770726 on Build GRCh38
rs752661665 Chr.2: 224770606-224770606 on Build GRCh38
rs200296578 Chr.2: 224770501-224770501 on Build GRCh38
rs756461033 Chr.2: 224770629-224770629 on Build GRCh38

View Full Product Details