Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232481399-232481896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECEL1
Entrez Gene ID
9427
Gene Name
endothelin converting enzyme like 1
Gene Aliases
DA5D, DINE, ECEX, XCE
UniGene
Hs.26880
Gene Chromosome Location
Chr.2: 232479827-232487859 on Build GRCh38
Transcripts
NM_001290787.1 NM_004826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651322 c.1969T>C p.F657L Chr.2: 232481526-232481526 on Build GRCh38
310801 c.1959C>G p.L653L Chr.2: 232481536-232481536 on Build GRCh38
3909837 c.1929C>T p.F643F Chr.2: 232481566-232481566 on Build GRCh38
7239745 c.1894C>T p.L632L Chr.2: 232481601-232481601 on Build GRCh38
6192630 c.1988G>A p.R663Q Chr.2: 232481507-232481507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140594660 Chr.2: 232481616-232481616 on Build GRCh38
rs1016421273 Chr.2: 232481446-232481446 on Build GRCh38
rs766572719 Chr.2: 232481797-232481797 on Build GRCh38
rs148671826 Chr.2: 232481587-232481587 on Build GRCh38
rs755456884 Chr.2: 232481636-232481636 on Build GRCh38
rs749748838 Chr.2: 232481635-232481635 on Build GRCh38
rs780485015 Chr.2: 232481571-232481571 on Build GRCh38
rs145010012 Chr.2: 232481615-232481615 on Build GRCh38
rs749969915 Chr.2: 232481427-232481427 on Build GRCh38
rs771635783 Chr.2: 232481642-232481642 on Build GRCh38
rs558933471 Chr.2: 232481459-232481459 on Build GRCh38
rs138272112 Chr.2: 232481627-232481627 on Build GRCh38
rs981783065 Chr.2: 232481442-232481442 on Build GRCh38
rs997757623 Chr.2: 232481570-232481570 on Build GRCh38
rs754749774 Chr.2: 232481544-232481544 on Build GRCh38
rs753679242 Chr.2: 232481458-232481458 on Build GRCh38
rs149922272 Chr.2: 232481558-232481558 on Build GRCh38
rs780570845 Chr.2: 232481631-232481631 on Build GRCh38
rs142492002 Chr.2: 232481788-232481788 on Build GRCh38
rs981157141 Chr.2: 232481781-232481781 on Build GRCh38
rs760929496 Chr.2: 232481796-232481796 on Build GRCh38
rs766889105 Chr.2: 232481522-232481522 on Build GRCh38
rs924450163 Chr.2: 232481556-232481556 on Build GRCh38
rs942039552 Chr.2: 232481841-232481841 on Build GRCh38
rs767771279 Chr.2: 232481626-232481626 on Build GRCh38
rs770512434 Chr.2: 232481656-232481656 on Build GRCh38
rs201176879 Chr.2: 232481676-232481676 on Build GRCh38
rs775435337 Chr.2: 232481791-232481791 on Build GRCh38
rs769168956 Chr.2: 232481870-232481870 on Build GRCh38
rs781173464 Chr.2: 232481561-232481561 on Build GRCh38
rs575131423 Chr.2: 232481637-232481637 on Build GRCh38
rs376446735 Chr.2: 232481837-232481837 on Build GRCh38
rs751965556 Chr.2: 232481830-232481830 on Build GRCh38
rs753697610 Chr.2: 232481541-232481541 on Build GRCh38
rs745378093 Chr.2: 232481836-232481836 on Build GRCh38
rs767641178 Chr.2: 232481673-232481673 on Build GRCh38
rs748529947 Chr.2: 232481641-232481641 on Build GRCh38
rs768312431 Chr.2: 232481508-232481508 on Build GRCh38
rs11285283 Chr.2: 232481672-232481672 on Build GRCh38
rs770673747 Chr.2: 232481768-232481768 on Build GRCh38
rs540904303 Chr.2: 232481666-232481666 on Build GRCh38
rs758805366 Chr.2: 232481826-232481826 on Build GRCh38
rs753195318 Chr.2: 232481823-232481823 on Build GRCh38
rs68130818 Chr.2: 232481745-232481745 on Build GRCh38
rs764405950 Chr.2: 232481545-232481545 on Build GRCh38
rs764591336 Chr.2: 232481829-232481829 on Build GRCh38
rs770259285 Chr.2: 232481486-232481486 on Build GRCh38
rs760203132 Chr.2: 232481611-232481611 on Build GRCh38
rs746705900 Chr.2: 232481765-232481765 on Build GRCh38
rs139791336 Chr.2: 232481532-232481532 on Build GRCh38
rs543950131 Chr.2: 232481832-232481832 on Build GRCh38
rs777586315 Chr.2: 232481764-232481764 on Build GRCh38
rs374941711 Chr.2: 232481660-232481660 on Build GRCh38
rs777417335 Chr.2: 232481643-232481643 on Build GRCh38
rs759583251 Chr.2: 232481657-232481657 on Build GRCh38
rs764987058 Chr.2: 232481618-232481618 on Build GRCh38
rs749296865 Chr.2: 232481840-232481840 on Build GRCh38
rs76290356 Chr.2: 232481859-232481859 on Build GRCh38
rs757490576 Chr.2: 232481624-232481624 on Build GRCh38
rs200251847 Chr.2: 232481861-232481861 on Build GRCh38
rs761418483 Chr.2: 232481520-232481520 on Build GRCh38
rs771146485 Chr.2: 232481868-232481868 on Build GRCh38
rs376306807 Chr.2: 232481757-232481757 on Build GRCh38
rs374276757 Chr.2: 232481750-232481750 on Build GRCh38
rs775902800 Chr.2: 232481494-232481494 on Build GRCh38
rs759553480 Chr.2: 232481876-232481876 on Build GRCh38
rs780937186 Chr.2: 232481833-232481833 on Build GRCh38
rs1011343381 Chr.2: 232481730-232481730 on Build GRCh38
rs967430473 Chr.2: 232481749-232481749 on Build GRCh38
rs756675006 Chr.2: 232481562-232481562 on Build GRCh38
rs958652798 Chr.2: 232481809-232481809 on Build GRCh38
rs989774338 Chr.2: 232481430-232481430 on Build GRCh38
rs953693275 Chr.2: 232481425-232481425 on Build GRCh38
rs746415040 Chr.2: 232481478-232481478 on Build GRCh38
rs766496935 Chr.2: 232481736-232481736 on Build GRCh38
rs1025250133 Chr.2: 232481716-232481716 on Build GRCh38
rs1013322162 Chr.2: 232481682-232481682 on Build GRCh38
rs745360606 Chr.2: 232481789-232481789 on Build GRCh38
rs768971199 Chr.2: 232481579-232481579 on Build GRCh38
rs201366755 Chr.2: 232481527-232481527 on Build GRCh38
rs756826367 Chr.2: 232481683-232481683 on Build GRCh38
rs957143258 Chr.2: 232481429-232481429 on Build GRCh38
rs760766235 Chr.2: 232481678-232481678 on Build GRCh38
rs367703178 Chr.2: 232481576-232481576 on Build GRCh38
rs756121085 Chr.2: 232481628-232481628 on Build GRCh38
rs530513811 Chr.2: 232481806-232481806 on Build GRCh38
rs146580059 Chr.2: 232481588-232481588 on Build GRCh38
rs370173849 Chr.2: 232481619-232481619 on Build GRCh38
rs772654309 Chr.2: 232481605-232481605 on Build GRCh38
rs772747375 Chr.2: 232481528-232481528 on Build GRCh38
rs967290967 Chr.2: 232481436-232481436 on Build GRCh38
rs368507400 Chr.2: 232481838-232481838 on Build GRCh38
rs773809634 Chr.2: 232481511-232481511 on Build GRCh38
rs372170199 Chr.2: 232481792-232481792 on Build GRCh38
rs569210107 Chr.2: 232481540-232481540 on Build GRCh38
rs747113853 Chr.2: 232481460-232481460 on Build GRCh38
rs1017208484 Chr.2: 232481685-232481685 on Build GRCh38
rs968811682 Chr.2: 232481743-232481743 on Build GRCh38
rs765693591 Chr.2: 232481810-232481810 on Build GRCh38
rs369916383 Chr.2: 232481465-232481465 on Build GRCh38
rs781731856 Chr.2: 232481473-232481473 on Build GRCh38
rs1017264586 Chr.2: 232481521-232481521 on Build GRCh38
rs558915148 Chr.2: 232481507-232481507 on Build GRCh38
rs762755047 Chr.2: 232481669-232481669 on Build GRCh38
rs538623666 Chr.2: 232481453-232481453 on Build GRCh38
rs746546278 Chr.2: 232481649-232481649 on Build GRCh38
rs57474503 Chr.2: 232481610-232481610 on Build GRCh38
rs186105776 Chr.2: 232481426-232481426 on Build GRCh38
rs867596526 Chr.2: 232481677-232481677 on Build GRCh38
rs774112459 Chr.2: 232481793-232481793 on Build GRCh38
rs768552137 Chr.2: 232481849-232481849 on Build GRCh38
rs769637000 Chr.2: 232481790-232481790 on Build GRCh38
rs566513569 Chr.2: 232481443-232481443 on Build GRCh38
rs912896342 Chr.2: 232481431-232481431 on Build GRCh38

View Full Product Details