Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232526072-232526579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRND
Entrez Gene ID
1144
Gene Name
cholinergic receptor nicotinic delta subunit
Gene Aliases
ACHRD, CMS2A, CMS3A, CMS3B, CMS3C, FCCMS, SCCMS
UniGene
Hs.156289
Gene Chromosome Location
Chr.2: 232526160-232537907 on Build GRCh38
Transcripts
NM_000751.2 NM_001256657.1 NM_001311195.1 NM_001311196.1 XM_011510524.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6090571 c.55A>T p.S19C Chr.2: 232526531-232526531 on Build GRCh38
4792330 c.88C>T p.R30W Chr.2: 232526564-232526564 on Build GRCh38
4911211 c.51T>C p.C17C Chr.2: 232526266-232526266 on Build GRCh38
6628797 c.89G>A p.R30Q Chr.2: 232526565-232526565 on Build GRCh38
720242 c.42G>A p.L14L Chr.2: 232526257-232526257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047699815 Chr.2: 232526377-232526377 on Build GRCh38
rs2278479 Chr.2: 232526097-232526097 on Build GRCh38
rs778460094 Chr.2: 232526169-232526169 on Build GRCh38
rs992162337 Chr.2: 232526510-232526510 on Build GRCh38
rs974637045 Chr.2: 232526264-232526264 on Build GRCh38
rs1003922268 Chr.2: 232526098-232526098 on Build GRCh38
rs773871977 Chr.2: 232526496-232526496 on Build GRCh38
rs748208954 Chr.2: 232526159-232526159 on Build GRCh38
rs780953178 Chr.2: 232526142-232526142 on Build GRCh38
rs755159887 Chr.2: 232526149-232526149 on Build GRCh38
rs372429801 Chr.2: 232526329-232526329 on Build GRCh38
rs771477279 Chr.2: 232526178-232526178 on Build GRCh38
rs143002322 Chr.2: 232526556-232526556 on Build GRCh38
rs748945280 Chr.2: 232526318-232526318 on Build GRCh38
rs995852848 Chr.2: 232526430-232526430 on Build GRCh38
rs200197532 Chr.2: 232526508-232526508 on Build GRCh38
rs1018725870 Chr.2: 232526407-232526407 on Build GRCh38
rs752197775 Chr.2: 232526129-232526129 on Build GRCh38
rs570354782 Chr.2: 232526205-232526205 on Build GRCh38
rs138609765 Chr.2: 232526260-232526260 on Build GRCh38
rs767302324 Chr.2: 232526200-232526200 on Build GRCh38
rs757095507 Chr.2: 232526140-232526140 on Build GRCh38
rs144310284 Chr.2: 232526546-232526546 on Build GRCh38
rs148184674 Chr.2: 232526450-232526450 on Build GRCh38
rs550189860 Chr.2: 232526187-232526187 on Build GRCh38
rs751643298 Chr.2: 232526299-232526299 on Build GRCh38
rs763294373 Chr.2: 232526526-232526526 on Build GRCh38
rs774897840 Chr.2: 232526191-232526191 on Build GRCh38
rs797044480 Chr.2: 232526549-232526549 on Build GRCh38
rs968410979 Chr.2: 232526453-232526453 on Build GRCh38
rs368375643 Chr.2: 232526376-232526376 on Build GRCh38
rs990161041 Chr.2: 232526204-232526204 on Build GRCh38
rs751467622 Chr.2: 232526134-232526134 on Build GRCh38
rs898774609 Chr.2: 232526364-232526364 on Build GRCh38
rs369256162 Chr.2: 232526314-232526314 on Build GRCh38
rs533586423 Chr.2: 232526150-232526150 on Build GRCh38
rs749539244 Chr.2: 232526148-232526148 on Build GRCh38
rs759225916 Chr.2: 232526122-232526122 on Build GRCh38
rs767394061 Chr.2: 232526283-232526283 on Build GRCh38
rs556184561 Chr.2: 232526464-232526464 on Build GRCh38
rs182404872 Chr.2: 232526304-232526304 on Build GRCh38
rs561839709 Chr.2: 232526111-232526111 on Build GRCh38
rs752826834 Chr.2: 232526237-232526237 on Build GRCh38
rs367844338 Chr.2: 232526168-232526168 on Build GRCh38
rs900956921 Chr.2: 232526370-232526370 on Build GRCh38
rs917893284 Chr.2: 232526322-232526322 on Build GRCh38
rs764285104 Chr.2: 232526298-232526298 on Build GRCh38
rs776242631 Chr.2: 232526513-232526513 on Build GRCh38
rs773782023 Chr.2: 232526279-232526279 on Build GRCh38
rs762565020 Chr.2: 232526192-232526192 on Build GRCh38
rs781437381 Chr.2: 232526250-232526250 on Build GRCh38
rs773247272 Chr.2: 232526511-232526511 on Build GRCh38
rs554645326 Chr.2: 232526242-232526242 on Build GRCh38
rs753019300 Chr.2: 232526296-232526296 on Build GRCh38
rs967873665 Chr.2: 232526285-232526285 on Build GRCh38
rs965976024 Chr.2: 232526102-232526102 on Build GRCh38
rs2245601 Chr.2: 232526227-232526227 on Build GRCh38
rs996084545 Chr.2: 232526225-232526225 on Build GRCh38
rs879020024 Chr.2: 232526380-232526380 on Build GRCh38
rs886225762 Chr.2: 232526233-232526233 on Build GRCh38
rs142531974 Chr.2: 232526259-232526259 on Build GRCh38
rs761751167 Chr.2: 232526282-232526282 on Build GRCh38
rs773295789 Chr.2: 232526383-232526383 on Build GRCh38
rs759127614 Chr.2: 232526186-232526186 on Build GRCh38
rs751557545 Chr.2: 232526195-232526195 on Build GRCh38
rs756559731 Chr.2: 232526308-232526308 on Build GRCh38
rs375578715 Chr.2: 232526243-232526243 on Build GRCh38
rs371414024 Chr.2: 232526196-232526196 on Build GRCh38
rs527741052 Chr.2: 232526112-232526112 on Build GRCh38
rs193259207 Chr.2: 232526123-232526123 on Build GRCh38
rs760750818 Chr.2: 232526420-232526420 on Build GRCh38
rs1031222246 Chr.2: 232526474-232526474 on Build GRCh38
rs776233680 Chr.2: 232526521-232526521 on Build GRCh38
rs750216316 Chr.2: 232526288-232526288 on Build GRCh38
rs1028610535 Chr.2: 232526439-232526439 on Build GRCh38
rs576328637 Chr.2: 232526490-232526490 on Build GRCh38
rs773968785 Chr.2: 232526143-232526143 on Build GRCh38
rs768222852 Chr.2: 232526272-232526272 on Build GRCh38
rs781744411 Chr.2: 232526303-232526303 on Build GRCh38
rs967382414 Chr.2: 232526456-232526456 on Build GRCh38
rs372385617 Chr.2: 232526514-232526514 on Build GRCh38
rs766061648 Chr.2: 232526295-232526295 on Build GRCh38
rs757874864 Chr.2: 232526300-232526300 on Build GRCh38
rs777072923 Chr.2: 232526184-232526184 on Build GRCh38
rs774636657 Chr.2: 232526158-232526158 on Build GRCh38
rs974562134 Chr.2: 232526103-232526103 on Build GRCh38
rs1003889655 Chr.2: 232526332-232526335 on Build GRCh38
rs764374927 Chr.2: 232526535-232526535 on Build GRCh38
rs989804249 Chr.2: 232526446-232526446 on Build GRCh38
rs746207666 Chr.2: 232526257-232526257 on Build GRCh38
rs200921489 Chr.2: 232526555-232526555 on Build GRCh38
rs760555842 Chr.2: 232526290-232526290 on Build GRCh38
rs547486962 Chr.2: 232526119-232526119 on Build GRCh38
rs1045253302 Chr.2: 232526360-232526360 on Build GRCh38
rs750174953 Chr.2: 232526217-232526217 on Build GRCh38
rs980778196 Chr.2: 232526552-232526552 on Build GRCh38
rs747616715 Chr.2: 232526499-232526499 on Build GRCh38
rs763981083 Chr.2: 232526133-232526133 on Build GRCh38
rs957944888 Chr.2: 232526441-232526441 on Build GRCh38
rs747494704 Chr.2: 232526173-232526173 on Build GRCh38
rs770901573 Chr.2: 232526139-232526139 on Build GRCh38
rs755177195 Chr.2: 232526226-232526226 on Build GRCh38
rs1004608200 Chr.2: 232526384-232526384 on Build GRCh38

View Full Product Details