Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232542741-232543260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNG
Entrez Gene ID
1146
Gene Name
cholinergic receptor nicotinic gamma subunit
Gene Aliases
ACHRG
UniGene
Hs.248101
Gene Chromosome Location
Chr.2: 232539727-232546328 on Build GRCh38
Transcripts
NM_005199.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963328 c.662C>T p.A221V Chr.2: 232542939-232542939 on Build GRCh38
4597923 c.711C>G p.I237M Chr.2: 232542988-232542988 on Build GRCh38
1578617 c.708C>T p.L236L Chr.2: 232542985-232542985 on Build GRCh38
1017834 c.768C>A p.S256S Chr.2: 232543045-232543045 on Build GRCh38
1406242 c.749delC p.C252fs*58 Chr.2: 232543026-232543026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778517594 Chr.2: 232543041-232543041 on Build GRCh38
rs936925551 Chr.2: 232543090-232543090 on Build GRCh38
rs759808539 Chr.2: 232543131-232543131 on Build GRCh38
rs774616155 Chr.2: 232542999-232542999 on Build GRCh38
rs557970419 Chr.2: 232543010-232543010 on Build GRCh38
rs752042214 Chr.2: 232542862-232542862 on Build GRCh38
rs775776788 Chr.2: 232542921-232542921 on Build GRCh38
rs561484280 Chr.2: 232542818-232542818 on Build GRCh38
rs772048048 Chr.2: 232542987-232542987 on Build GRCh38
rs747094975 Chr.2: 232542885-232542885 on Build GRCh38
rs770024550 Chr.2: 232543157-232543157 on Build GRCh38
rs760442965 Chr.2: 232542914-232542914 on Build GRCh38
rs549490400 Chr.2: 232543115-232543115 on Build GRCh38
rs776450073 Chr.2: 232543085-232543085 on Build GRCh38
rs995913282 Chr.2: 232542886-232542886 on Build GRCh38
rs150556220 Chr.2: 232542940-232542940 on Build GRCh38
rs1024372201 Chr.2: 232542821-232542821 on Build GRCh38
rs748417616 Chr.2: 232543122-232543122 on Build GRCh38
rs145847096 Chr.2: 232543031-232543031 on Build GRCh38
rs138125827 Chr.2: 232543052-232543052 on Build GRCh38
rs755046404 Chr.2: 232543012-232543017 on Build GRCh38
rs1012323697 Chr.2: 232542774-232542774 on Build GRCh38
rs764938298 Chr.2: 232543132-232543132 on Build GRCh38
rs748912147 Chr.2: 232542944-232542944 on Build GRCh38
rs1011205791 Chr.2: 232543048-232543048 on Build GRCh38
rs752319888 Chr.2: 232542858-232542859 on Build GRCh38
rs961945316 Chr.2: 232543197-232543197 on Build GRCh38
rs377079049 Chr.2: 232543096-232543096 on Build GRCh38
rs2697782 Chr.2: 232542806-232542806 on Build GRCh38
rs754276441 Chr.2: 232543120-232543120 on Build GRCh38
rs777027483 Chr.2: 232543051-232543051 on Build GRCh38
rs17838626 Chr.2: 232542991-232542991 on Build GRCh38
rs1049133814 Chr.2: 232542997-232542997 on Build GRCh38
rs566138449 Chr.2: 232543139-232543139 on Build GRCh38
rs757262184 Chr.2: 232543232-232543232 on Build GRCh38
rs766716676 Chr.2: 232543024-232543024 on Build GRCh38
rs201149972 Chr.2: 232543117-232543117 on Build GRCh38
rs184315065 Chr.2: 232542875-232542875 on Build GRCh38
rs751486796 Chr.2: 232543230-232543230 on Build GRCh38
rs747624528 Chr.2: 232542980-232542980 on Build GRCh38
rs374951822 Chr.2: 232543029-232543029 on Build GRCh38
rs773121173 Chr.2: 232542988-232542988 on Build GRCh38
rs778445156 Chr.2: 232542973-232542973 on Build GRCh38
rs772217042 Chr.2: 232543106-232543106 on Build GRCh38
rs955751639 Chr.2: 232542817-232542817 on Build GRCh38
rs752137444 Chr.2: 232542936-232542936 on Build GRCh38
rs56755137 Chr.2: 232543206-232543206 on Build GRCh38
rs746910885 Chr.2: 232543075-232543075 on Build GRCh38
rs372184553 Chr.2: 232543086-232543086 on Build GRCh38
rs752281775 Chr.2: 232543224-232543224 on Build GRCh38
rs370116501 Chr.2: 232542909-232542909 on Build GRCh38
rs747062636 Chr.2: 232543011-232543013 on Build GRCh38
rs778730695 Chr.2: 232543007-232543008 on Build GRCh38
rs557680782 Chr.2: 232543227-232543227 on Build GRCh38
rs767367706 Chr.2: 232543175-232543175 on Build GRCh38
rs767144116 Chr.2: 232542859-232542862 on Build GRCh38
rs745549857 Chr.2: 232542877-232542877 on Build GRCh38
rs765407839 Chr.2: 232542993-232542993 on Build GRCh38
rs1014694994 Chr.2: 232543195-232543195 on Build GRCh38
rs1029276370 Chr.2: 232542896-232542896 on Build GRCh38
rs779573860 Chr.2: 232542942-232542942 on Build GRCh38
rs750873458 Chr.2: 232542939-232542939 on Build GRCh38
rs754467232 Chr.2: 232542948-232542948 on Build GRCh38
rs768145897 Chr.2: 232542897-232542897 on Build GRCh38
rs761688091 Chr.2: 232542908-232542908 on Build GRCh38
rs780972486 Chr.2: 232543236-232543236 on Build GRCh38
rs558357870 Chr.2: 232543158-232543158 on Build GRCh38
rs200224934 Chr.2: 232542937-232542937 on Build GRCh38
rs535129324 Chr.2: 232543140-232543140 on Build GRCh38
rs940761016 Chr.2: 232543213-232543213 on Build GRCh38
rs369008934 Chr.2: 232542901-232542901 on Build GRCh38
rs550604447 Chr.2: 232542844-232542844 on Build GRCh38
rs565093373 Chr.2: 232542793-232542793 on Build GRCh38
rs778211410 Chr.2: 232542861-232542861 on Build GRCh38
rs202023568 Chr.2: 232542869-232542870 on Build GRCh38
rs747716114 Chr.2: 232543044-232543044 on Build GRCh38
rs1031178915 Chr.2: 232542808-232542808 on Build GRCh38
rs148066463 Chr.2: 232542938-232542938 on Build GRCh38
rs764180899 Chr.2: 232542932-232542932 on Build GRCh38
rs750890018 Chr.2: 232543012-232543012 on Build GRCh38
rs765437470 Chr.2: 232542915-232542915 on Build GRCh38
rs578137720 Chr.2: 232543179-232543179 on Build GRCh38
rs906953180 Chr.2: 232543143-232543143 on Build GRCh38
rs202064907 Chr.2: 232542879-232542879 on Build GRCh38
rs764114084 Chr.2: 232542994-232542994 on Build GRCh38
rs1027773442 Chr.2: 232542791-232542791 on Build GRCh38
rs1050425905 Chr.2: 232543050-232543050 on Build GRCh38
rs367938116 Chr.2: 232543083-232543083 on Build GRCh38
rs767503038 Chr.2: 232543030-232543031 on Build GRCh38
rs368225965 Chr.2: 232543099-232543099 on Build GRCh38
rs754891684 Chr.2: 232543147-232543147 on Build GRCh38
rs566102476 Chr.2: 232543194-232543194 on Build GRCh38
rs139998228 Chr.2: 232543014-232543014 on Build GRCh38
rs769617058 Chr.2: 232542878-232542878 on Build GRCh38
rs757198225 Chr.2: 232542880-232542881 on Build GRCh38
rs761969308 Chr.2: 232543009-232543009 on Build GRCh38
rs777692827 Chr.2: 232543066-232543066 on Build GRCh38
rs200473548 Chr.2: 232542899-232542899 on Build GRCh38
rs748812705 Chr.2: 232542892-232542892 on Build GRCh38
rs758491822 Chr.2: 232542842-232542842 on Build GRCh38
rs757713343 Chr.2: 232542874-232542874 on Build GRCh38
rs762950375 Chr.2: 232542927-232542927 on Build GRCh38
rs948000256 Chr.2: 232543149-232543149 on Build GRCh38
rs967551582 Chr.2: 232543062-232543062 on Build GRCh38
rs1005462691 Chr.2: 232542806-232542806 on Build GRCh38
rs576014872 Chr.2: 232542773-232542773 on Build GRCh38

View Full Product Details