Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232765681-232766180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
UniGene
Hs.467338
Gene Chromosome Location
Chr.2: 232765802-232776565 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

SNP Detail

SNP ID Chromosome Location
rs960685260 Chr.2: 232766038-232766038 on Build GRCh38
rs746826106 Chr.2: 232765863-232765863 on Build GRCh38
rs1018589645 Chr.2: 232766035-232766035 on Build GRCh38
rs76965238 Chr.2: 232765868-232765868 on Build GRCh38
rs978318637 Chr.2: 232766005-232766005 on Build GRCh38
rs921747206 Chr.2: 232765844-232765844 on Build GRCh38
rs967872871 Chr.2: 232766142-232766142 on Build GRCh38
rs199920802 Chr.2: 232765881-232765881 on Build GRCh38
rs1056657698 Chr.2: 232765800-232765800 on Build GRCh38
rs186922943 Chr.2: 232765920-232765920 on Build GRCh38
rs74645467 Chr.2: 232765744-232765744 on Build GRCh38
rs897149263 Chr.2: 232765948-232765948 on Build GRCh38
rs1012480479 Chr.2: 232765938-232765938 on Build GRCh38
rs74838457 Chr.2: 232765821-232765821 on Build GRCh38
rs923980084 Chr.2: 232765876-232765876 on Build GRCh38
rs935334877 Chr.2: 232765904-232765904 on Build GRCh38
rs994584652 Chr.2: 232765962-232765962 on Build GRCh38
rs1022433298 Chr.2: 232765984-232765984 on Build GRCh38
rs776379309 Chr.2: 232765947-232765947 on Build GRCh38
rs978287489 Chr.2: 232765747-232765747 on Build GRCh38
rs201779513 Chr.2: 232765743-232765743 on Build GRCh38
rs553930244 Chr.2: 232765914-232765914 on Build GRCh38
rs947908643 Chr.2: 232765911-232765911 on Build GRCh38
rs950384773 Chr.2: 232765753-232765753 on Build GRCh38
rs751364011 Chr.2: 232766010-232766010 on Build GRCh38
rs770531736 Chr.2: 232765940-232765940 on Build GRCh38
rs74547374 Chr.2: 232765867-232765867 on Build GRCh38
rs76728154 Chr.2: 232765848-232765848 on Build GRCh38
rs764743784 Chr.2: 232765956-232765956 on Build GRCh38
rs182197159 Chr.2: 232766050-232766050 on Build GRCh38
rs77237820 Chr.2: 232765966-232765966 on Build GRCh38
rs974676104 Chr.2: 232766109-232766109 on Build GRCh38
rs79510296 Chr.2: 232765854-232765854 on Build GRCh38
rs142756645 Chr.2: 232765928-232765928 on Build GRCh38
rs745319511 Chr.2: 232765955-232765955 on Build GRCh38
rs1004969203 Chr.2: 232766006-232766006 on Build GRCh38
rs559960188 Chr.2: 232766128-232766128 on Build GRCh38
rs568786303 Chr.2: 232765730-232765730 on Build GRCh38
rs77407851 Chr.2: 232765815-232765815 on Build GRCh38
rs975001255 Chr.2: 232765797-232765797 on Build GRCh38
rs76055053 Chr.2: 232765739-232765739 on Build GRCh38
rs762949156 Chr.2: 232765967-232765967 on Build GRCh38
rs79767537 Chr.2: 232765832-232765832 on Build GRCh38
rs77197088 Chr.2: 232765841-232765841 on Build GRCh38
rs983100165 Chr.2: 232765759-232765759 on Build GRCh38
rs1028867223 Chr.2: 232766111-232766111 on Build GRCh38
rs375274164 Chr.2: 232765979-232765979 on Build GRCh38
rs556916615 Chr.2: 232765949-232765949 on Build GRCh38
rs181444211 Chr.2: 232765765-232765765 on Build GRCh38
rs529652402 Chr.2: 232766040-232766040 on Build GRCh38
rs568106009 Chr.2: 232765864-232765864 on Build GRCh38
rs1036245901 Chr.2: 232765941-232765941 on Build GRCh38
rs78445597 Chr.2: 232765847-232765847 on Build GRCh38
rs776619856 Chr.2: 232765870-232765870 on Build GRCh38
rs968156060 Chr.2: 232765986-232765986 on Build GRCh38
rs911806648 Chr.2: 232765763-232765763 on Build GRCh38
rs76809839 Chr.2: 232765816-232765816 on Build GRCh38
rs924216122 Chr.2: 232766016-232766016 on Build GRCh38
rs191606103 Chr.2: 232766039-232766039 on Build GRCh38
rs797001702 Chr.2: 232765742-232765742 on Build GRCh38
rs554578441 Chr.2: 232765798-232765798 on Build GRCh38
rs1056437805 Chr.2: 232765905-232765905 on Build GRCh38

View Full Product Details