Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233434688-233435187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
UniGene
Hs.471675
Gene Chromosome Location
Chr.2: 233354507-233472098 on Build GRCh38
Transcripts
NM_003648.2 NM_152879.2 XM_005246107.3 XM_005246108.1 XM_011512028.2 XM_011512029.2 XM_011512030.2 XM_011512031.2 XM_011512032.2 XM_011512033.2 XM_011512035.1 XM_011512037.2 XM_011512039.2 XM_011512040.2 XM_017005138.1 XM_017005139.1 XM_017005140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3049952 c.471G>A p.M157I Chr.2: 233434786-233434786 on Build GRCh38
4092476 c.513C>T p.H171H Chr.2: 233434828-233434828 on Build GRCh38
5509886 c.501C>T p.Y167Y Chr.2: 233434816-233434816 on Build GRCh38
6356067 c.434C>T p.S145L Chr.2: 233434881-233434881 on Build GRCh38
4092477 c.514G>A p.A172T Chr.2: 233434829-233434829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574256020 Chr.2: 233434942-233434942 on Build GRCh38
rs774531099 Chr.2: 233435068-233435068 on Build GRCh38
rs774933426 Chr.2: 233434882-233434882 on Build GRCh38
rs758874885 Chr.2: 233434920-233434920 on Build GRCh38
rs202176763 Chr.2: 233434772-233434772 on Build GRCh38
rs757649244 Chr.2: 233434936-233434936 on Build GRCh38
rs578036955 Chr.2: 233434856-233434856 on Build GRCh38
rs528850359 Chr.2: 233435158-233435158 on Build GRCh38
rs143682023 Chr.2: 233434831-233434831 on Build GRCh38
rs1045919869 Chr.2: 233434997-233434997 on Build GRCh38
rs192660394 Chr.2: 233434935-233434935 on Build GRCh38
rs564058408 Chr.2: 233435067-233435067 on Build GRCh38
rs1049782672 Chr.2: 233434894-233434894 on Build GRCh38
rs372316084 Chr.2: 233434759-233434759 on Build GRCh38
rs977724333 Chr.2: 233435103-233435104 on Build GRCh38
rs770550183 Chr.2: 233434848-233434848 on Build GRCh38
rs777005748 Chr.2: 233434906-233434906 on Build GRCh38
rs769776662 Chr.2: 233434837-233434837 on Build GRCh38
rs116803002 Chr.2: 233434744-233434744 on Build GRCh38
rs150294529 Chr.2: 233434828-233434828 on Build GRCh38
rs747381293 Chr.2: 233434843-233434843 on Build GRCh38
rs369790670 Chr.2: 233434885-233434885 on Build GRCh38
rs187318580 Chr.2: 233435009-233435009 on Build GRCh38
rs755598405 Chr.2: 233434720-233434720 on Build GRCh38
rs543302103 Chr.2: 233434983-233434983 on Build GRCh38
rs914387468 Chr.2: 233435062-233435062 on Build GRCh38
rs545595388 Chr.2: 233435041-233435041 on Build GRCh38
rs985292524 Chr.2: 233435162-233435162 on Build GRCh38
rs1001704463 Chr.2: 233435012-233435012 on Build GRCh38
rs776057203 Chr.2: 233434769-233434769 on Build GRCh38
rs765434987 Chr.2: 233434913-233434913 on Build GRCh38
rs761662091 Chr.2: 233434897-233434897 on Build GRCh38
rs760440640 Chr.2: 233435142-233435142 on Build GRCh38
rs779558587 Chr.2: 233434724-233434724 on Build GRCh38
rs962006085 Chr.2: 233435123-233435123 on Build GRCh38
rs772575218 Chr.2: 233434747-233434747 on Build GRCh38
rs183575237 Chr.2: 233435021-233435021 on Build GRCh38
rs989959809 Chr.2: 233435024-233435024 on Build GRCh38
rs543070012 Chr.2: 233435129-233435129 on Build GRCh38
rs113434947 Chr.2: 233434945-233434945 on Build GRCh38
rs763817416 Chr.2: 233434795-233434795 on Build GRCh38
rs779966904 Chr.2: 233434857-233434857 on Build GRCh38
rs775498035 Chr.2: 233434869-233434869 on Build GRCh38
rs59249585 Chr.2: 233435096-233435096 on Build GRCh38
rs386352297 Chr.2: 233434787-233434787 on Build GRCh38
rs377274788 Chr.2: 233434804-233434804 on Build GRCh38
rs140466615 Chr.2: 233434878-233434878 on Build GRCh38
rs187747217 Chr.2: 233434912-233434912 on Build GRCh38
rs775414474 Chr.2: 233434782-233434782 on Build GRCh38
rs1010450268 Chr.2: 233434979-233434979 on Build GRCh38
rs778146685 Chr.2: 233434758-233434758 on Build GRCh38
rs1010418889 Chr.2: 233434739-233434739 on Build GRCh38
rs544210728 Chr.2: 233434967-233434967 on Build GRCh38
rs759036718 Chr.2: 233434770-233434770 on Build GRCh38
rs1014946664 Chr.2: 233435108-233435108 on Build GRCh38
rs111813835 Chr.2: 233434895-233434895 on Build GRCh38
rs758931999 Chr.2: 233434836-233434836 on Build GRCh38
rs972648687 Chr.2: 233435135-233435135 on Build GRCh38
rs760848357 Chr.2: 233434810-233434810 on Build GRCh38
rs558000890 Chr.2: 233434816-233434816 on Build GRCh38
rs78059682 Chr.2: 233434998-233434998 on Build GRCh38
rs201646438 Chr.2: 233434817-233434817 on Build GRCh38
rs764651439 Chr.2: 233434922-233434922 on Build GRCh38
rs112861398 Chr.2: 233434914-233434914 on Build GRCh38
rs879759729 Chr.2: 233434931-233434931 on Build GRCh38
rs779077238 Chr.2: 233434829-233434829 on Build GRCh38
rs767150603 Chr.2: 233434905-233434905 on Build GRCh38
rs188259019 Chr.2: 233435154-233435154 on Build GRCh38

View Full Product Details