Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233437299-233437805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
UniGene
Hs.471675
Gene Chromosome Location
Chr.2: 233354507-233472098 on Build GRCh38
Transcripts
NM_003648.2 NM_152879.2 XM_005246107.3 XM_005246108.1 XM_011512028.2 XM_011512029.2 XM_011512030.2 XM_011512031.2 XM_011512032.2 XM_011512033.2 XM_011512035.1 XM_011512037.2 XM_011512039.2 XM_011512040.2 XM_017005138.1 XM_017005139.1 XM_017005140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3579249 c.881C>T p.S294L Chr.2: 233437438-233437438 on Build GRCh38
3838880 c.896C>T p.T299M Chr.2: 233437453-233437453 on Build GRCh38
1018010 c.912C>T p.I304I Chr.2: 233437469-233437469 on Build GRCh38
6303574 c.738G>T p.L246L Chr.2: 233437427-233437427 on Build GRCh38
6303573 c.870G>T p.L290L Chr.2: 233437427-233437427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979741262 Chr.2: 233437515-233437515 on Build GRCh38
rs779848067 Chr.2: 233437352-233437352 on Build GRCh38
rs980905093 Chr.2: 233437774-233437774 on Build GRCh38
rs994264463 Chr.2: 233437451-233437451 on Build GRCh38
rs566580453 Chr.2: 233437349-233437349 on Build GRCh38
rs763067374 Chr.2: 233437709-233437709 on Build GRCh38
rs767344808 Chr.2: 233437347-233437347 on Build GRCh38
rs574751730 Chr.2: 233437568-233437568 on Build GRCh38
rs879074274 Chr.2: 233437398-233437398 on Build GRCh38
rs1045904029 Chr.2: 233437605-233437605 on Build GRCh38
rs920505407 Chr.2: 233437536-233437536 on Build GRCh38
rs372440528 Chr.2: 233437362-233437362 on Build GRCh38
rs763732532 Chr.2: 233437329-233437329 on Build GRCh38
rs766126986 Chr.2: 233437432-233437432 on Build GRCh38
rs768303020 Chr.2: 233437513-233437513 on Build GRCh38
rs983288073 Chr.2: 233437565-233437565 on Build GRCh38
rs891252989 Chr.2: 233437658-233437658 on Build GRCh38
rs927496586 Chr.2: 233437782-233437782 on Build GRCh38
rs774063864 Chr.2: 233437391-233437391 on Build GRCh38
rs950562154 Chr.2: 233437541-233437541 on Build GRCh38
rs757295350 Chr.2: 233437469-233437469 on Build GRCh38
rs1038996306 Chr.2: 233437628-233437628 on Build GRCh38
rs182015878 Chr.2: 233437439-233437439 on Build GRCh38
rs35862746 Chr.2: 233437554-233437555 on Build GRCh38
rs547547879 Chr.2: 233437343-233437343 on Build GRCh38
rs111916674 Chr.2: 233437706-233437706 on Build GRCh38
rs113702627 Chr.2: 233437520-233437520 on Build GRCh38
rs186510261 Chr.2: 233437655-233437655 on Build GRCh38
rs760577714 Chr.2: 233437518-233437518 on Build GRCh38
rs1008305241 Chr.2: 233437664-233437664 on Build GRCh38
rs934969693 Chr.2: 233437535-233437535 on Build GRCh38
rs554938784 Chr.2: 233437558-233437558 on Build GRCh38
rs189705564 Chr.2: 233437773-233437773 on Build GRCh38
rs750347550 Chr.2: 233437419-233437419 on Build GRCh38
rs376698642 Chr.2: 233437493-233437493 on Build GRCh38
rs753696369 Chr.2: 233437522-233437522 on Build GRCh38
rs921640178 Chr.2: 233437641-233437641 on Build GRCh38
rs774153689 Chr.2: 233437502-233437502 on Build GRCh38
rs758587372 Chr.2: 233437357-233437357 on Build GRCh38
rs747164359 Chr.2: 233437460-233437460 on Build GRCh38
rs562999062 Chr.2: 233437703-233437703 on Build GRCh38
rs775961540 Chr.2: 233437378-233437378 on Build GRCh38
rs546023520 Chr.2: 233437679-233437679 on Build GRCh38
rs770307616 Chr.2: 233437486-233437486 on Build GRCh38
rs766988422 Chr.2: 233437415-233437415 on Build GRCh38
rs143203690 Chr.2: 233437368-233437368 on Build GRCh38
rs777007059 Chr.2: 233437360-233437360 on Build GRCh38
rs987806495 Chr.2: 233437754-233437754 on Build GRCh38
rs763757604 Chr.2: 233437533-233437533 on Build GRCh38
rs906007684 Chr.2: 233437609-233437609 on Build GRCh38
rs756037747 Chr.2: 233437425-233437425 on Build GRCh38
rs979387341 Chr.2: 233437733-233437733 on Build GRCh38
rs746365249 Chr.2: 233437387-233437387 on Build GRCh38
rs775573428 Chr.2: 233437388-233437388 on Build GRCh38
rs117447187 Chr.2: 233437624-233437624 on Build GRCh38
rs55683548 Chr.2: 233437358-233437358 on Build GRCh38
rs1012336334 Chr.2: 233437743-233437743 on Build GRCh38
rs141457099 Chr.2: 233437457-233437457 on Build GRCh38
rs917628164 Chr.2: 233437571-233437571 on Build GRCh38
rs540455922 Chr.2: 233437608-233437608 on Build GRCh38
rs771532022 Chr.2: 233437359-233437359 on Build GRCh38
rs746396816 Chr.2: 233437476-233437476 on Build GRCh38
rs766322342 Chr.2: 233437521-233437521 on Build GRCh38
rs898532550 Chr.2: 233437753-233437753 on Build GRCh38
rs537835303 Chr.2: 233437475-233437475 on Build GRCh38
rs758673209 Chr.2: 233437453-233437453 on Build GRCh38
rs754368032 Chr.2: 233437776-233437776 on Build GRCh38
rs369085595 Chr.2: 233437507-233437507 on Build GRCh38
rs1018132399 Chr.2: 233437665-233437665 on Build GRCh38
rs759327316 Chr.2: 233437524-233437524 on Build GRCh38
rs946375370 Chr.2: 233437644-233437644 on Build GRCh38
rs973357506 Chr.2: 233437482-233437482 on Build GRCh38
rs764950943 Chr.2: 233437527-233437527 on Build GRCh38
rs375495360 Chr.2: 233437348-233437348 on Build GRCh38
rs896613975 Chr.2: 233437728-233437728 on Build GRCh38
rs113809759 Chr.2: 233437769-233437769 on Build GRCh38
rs761090914 Chr.2: 233437688-233437688 on Build GRCh38
rs898502167 Chr.2: 233437634-233437634 on Build GRCh38
rs924991345 Chr.2: 233437526-233437526 on Build GRCh38
rs531621583 Chr.2: 233437764-233437764 on Build GRCh38
rs761405794 Chr.2: 233437394-233437394 on Build GRCh38
rs767245913 Chr.2: 233437635-233437635 on Build GRCh38
rs773869249 Chr.2: 233437590-233437590 on Build GRCh38
rs1000161763 Chr.2: 233437689-233437689 on Build GRCh38
rs527912860 Chr.2: 233437342-233437342 on Build GRCh38
rs1024137908 Chr.2: 233437744-233437744 on Build GRCh38
rs748399563 Chr.2: 233437354-233437354 on Build GRCh38
rs761476841 Chr.2: 233437503-233437503 on Build GRCh38

View Full Product Details