Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233445516-233446014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
UniGene
Hs.471675
Gene Chromosome Location
Chr.2: 233354507-233472098 on Build GRCh38
Transcripts
NM_003648.2 NM_152879.2 XM_005246107.3 XM_005246108.1 XM_011512028.2 XM_011512029.2 XM_011512030.2 XM_011512031.2 XM_011512032.2 XM_011512033.2 XM_011512035.1 XM_011512037.2 XM_011512039.2 XM_011512040.2 XM_017005138.1 XM_017005139.1 XM_017005140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4677187 c.1272C>T p.D424D Chr.2: 233445700-233445700 on Build GRCh38
6362244 c.1126T>C p.S376P Chr.2: 233445686-233445686 on Build GRCh38
3838882 c.1259C>T p.S420L Chr.2: 233445687-233445687 on Build GRCh38
4001545 c.1246C>T p.L416L Chr.2: 233445674-233445674 on Build GRCh38
1631899 c.1218C>T p.L406L Chr.2: 233445646-233445646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777022600 Chr.2: 233445739-233445739 on Build GRCh38
rs374682653 Chr.2: 233445598-233445598 on Build GRCh38
rs761661468 Chr.2: 233445611-233445611 on Build GRCh38
rs769555743 Chr.2: 233445753-233445753 on Build GRCh38
rs555452983 Chr.2: 233445886-233445886 on Build GRCh38
rs73995952 Chr.2: 233445601-233445601 on Build GRCh38
rs1010254777 Chr.2: 233445984-233445984 on Build GRCh38
rs1013757442 Chr.2: 233445960-233445960 on Build GRCh38
rs775674058 Chr.2: 233445603-233445603 on Build GRCh38
rs775208893 Chr.2: 233445760-233445760 on Build GRCh38
rs891833493 Chr.2: 233445977-233445977 on Build GRCh38
rs751447542 Chr.2: 233445617-233445617 on Build GRCh38
rs757017390 Chr.2: 233445638-233445638 on Build GRCh38
rs557687267 Chr.2: 233445727-233445727 on Build GRCh38
rs186011630 Chr.2: 233445581-233445581 on Build GRCh38
rs1031149588 Chr.2: 233445565-233445565 on Build GRCh38
rs773630709 Chr.2: 233445584-233445584 on Build GRCh38
rs752854423 Chr.2: 233445793-233445793 on Build GRCh38
rs2305542 Chr.2: 233445599-233445599 on Build GRCh38
rs1043104731 Chr.2: 233445820-233445820 on Build GRCh38
rs773036311 Chr.2: 233445712-233445712 on Build GRCh38
rs1023926553 Chr.2: 233445963-233445963 on Build GRCh38
rs371361990 Chr.2: 233445781-233445781 on Build GRCh38
rs1021415105 Chr.2: 233445539-233445539 on Build GRCh38
rs749508553 Chr.2: 233445643-233445643 on Build GRCh38
rs940752845 Chr.2: 233445673-233445673 on Build GRCh38
rs934235177 Chr.2: 233445786-233445786 on Build GRCh38
rs563182761 Chr.2: 233445538-233445538 on Build GRCh38
rs764125308 Chr.2: 233445616-233445616 on Build GRCh38
rs772070434 Chr.2: 233445694-233445694 on Build GRCh38
rs767258853 Chr.2: 233445787-233445787 on Build GRCh38
rs139149715 Chr.2: 233445642-233445642 on Build GRCh38
rs370688469 Chr.2: 233445559-233445559 on Build GRCh38
rs762558406 Chr.2: 233445766-233445766 on Build GRCh38
rs201416623 Chr.2: 233445788-233445788 on Build GRCh38
rs566007321 Chr.2: 233445594-233445594 on Build GRCh38
rs752231437 Chr.2: 233445608-233445608 on Build GRCh38
rs911388690 Chr.2: 233445790-233445790 on Build GRCh38
rs1049892215 Chr.2: 233445548-233445548 on Build GRCh38
rs909591459 Chr.2: 233445695-233445695 on Build GRCh38
rs2305541 Chr.2: 233445674-233445674 on Build GRCh38
rs779016452 Chr.2: 233445654-233445654 on Build GRCh38
rs779110299 Chr.2: 233445791-233445791 on Build GRCh38
rs759158509 Chr.2: 233445748-233445748 on Build GRCh38
rs978315678 Chr.2: 233445560-233445560 on Build GRCh38
rs902959057 Chr.2: 233445841-233445841 on Build GRCh38
rs543375647 Chr.2: 233445985-233445985 on Build GRCh38
rs773601075 Chr.2: 233445700-233445700 on Build GRCh38
rs374652128 Chr.2: 233445782-233445782 on Build GRCh38
rs948118354 Chr.2: 233445619-233445619 on Build GRCh38
rs998980413 Chr.2: 233445864-233445864 on Build GRCh38
rs760609109 Chr.2: 233445929-233445929 on Build GRCh38
rs1054900825 Chr.2: 233445587-233445587 on Build GRCh38
rs936124353 Chr.2: 233445884-233445884 on Build GRCh38
rs763640486 Chr.2: 233445771-233445771 on Build GRCh38
rs764877759 Chr.2: 233445604-233445604 on Build GRCh38
rs909326979 Chr.2: 233445659-233445659 on Build GRCh38
rs144742201 Chr.2: 233445646-233445646 on Build GRCh38
rs1000244544 Chr.2: 233445575-233445575 on Build GRCh38
rs748697746 Chr.2: 233445579-233445579 on Build GRCh38
rs747497481 Chr.2: 233445715-233445715 on Build GRCh38
rs1054529141 Chr.2: 233445902-233445902 on Build GRCh38
rs760948494 Chr.2: 233445593-233445593 on Build GRCh38
rs572015493 Chr.2: 233445903-233445903 on Build GRCh38

View Full Product Details