Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233743787-233744297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB3
Entrez Gene ID
414061
Gene Name
DnaJ heat shock protein family (Hsp40) member B3
Gene Aliases
HCG3
UniGene
Hs.725533
Gene Chromosome Location
Chr.2: 233742750-233744015 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A10
Entrez Gene ID
54575
Gene Name
UDP glucuronosyltransferase family 1 member A10
Gene Aliases
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233636477-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A3
Entrez Gene ID
54659
Gene Name
UDP glucuronosyltransferase family 1 member A3
Gene Aliases
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233729127-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A4
Entrez Gene ID
54657
Gene Name
UDP glucuronosyltransferase family 1 member A4
Gene Aliases
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233718792-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A5
Entrez Gene ID
54579
Gene Name
UDP glucuronosyltransferase family 1 member A5
Gene Aliases
UDPGT, UDPGT 1-5, UGT1E
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233712992-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A6
Entrez Gene ID
54578
Gene Name
UDP glucuronosyltransferase family 1 member A6
Gene Aliases
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233691675-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233681938-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A8
Entrez Gene ID
54576
Gene Name
UDP glucuronosyltransferase family 1 member A8
Gene Aliases
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233617645-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A9
Entrez Gene ID
54600
Gene Name
UDP glucuronosyltransferase family 1 member A9
Gene Aliases
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233671898-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

SNP Detail

SNP ID Chromosome Location
rs981482683 Chr.2: 233743999-233743999 on Build GRCh38
rs749381543 Chr.2: 233743838-233743838 on Build GRCh38
rs17864701 Chr.2: 233744071-233744071 on Build GRCh38
rs576185082 Chr.2: 233744132-233744132 on Build GRCh38
rs371252305 Chr.2: 233743837-233743837 on Build GRCh38
rs776574080 Chr.2: 233743905-233743905 on Build GRCh38
rs964815042 Chr.2: 233743813-233743813 on Build GRCh38
rs1002862654 Chr.2: 233744129-233744129 on Build GRCh38
rs770873874 Chr.2: 233743852-233743852 on Build GRCh38
rs958752952 Chr.2: 233744159-233744159 on Build GRCh38
rs1039365766 Chr.2: 233744056-233744056 on Build GRCh38
rs369107156 Chr.2: 233743863-233743863 on Build GRCh38
rs535474047 Chr.2: 233744149-233744149 on Build GRCh38
rs1033622103 Chr.2: 233743983-233743983 on Build GRCh38
rs927230635 Chr.2: 233743932-233743932 on Build GRCh38
rs982618462 Chr.2: 233744202-233744202 on Build GRCh38
rs775471911 Chr.2: 233743963-233743963 on Build GRCh38
rs529549093 Chr.2: 233743835-233743835 on Build GRCh38
rs759074098 Chr.2: 233743853-233743853 on Build GRCh38
rs376911193 Chr.2: 233743890-233743890 on Build GRCh38
rs771949487 Chr.2: 233743847-233743847 on Build GRCh38
rs766652895 Chr.2: 233743936-233743936 on Build GRCh38
rs745723342 Chr.2: 233743960-233743960 on Build GRCh38
rs982904755 Chr.2: 233744048-233744048 on Build GRCh38
rs761038539 Chr.2: 233743935-233743935 on Build GRCh38
rs764939970 Chr.2: 233743822-233743822 on Build GRCh38
rs537609237 Chr.2: 233744108-233744108 on Build GRCh38
rs770691045 Chr.2: 233743902-233743902 on Build GRCh38
rs763731795 Chr.2: 233743923-233743923 on Build GRCh38
rs1020380348 Chr.2: 233744117-233744117 on Build GRCh38
rs779162939 Chr.2: 233743882-233743882 on Build GRCh38
rs115067532 Chr.2: 233744055-233744055 on Build GRCh38
rs150741507 Chr.2: 233744054-233744054 on Build GRCh38
rs541267003 Chr.2: 233744012-233744012 on Build GRCh38
rs768671945 Chr.2: 233743841-233743841 on Build GRCh38
rs535005289 Chr.2: 233744069-233744069 on Build GRCh38
rs374542514 Chr.2: 233744023-233744023 on Build GRCh38
rs367793428 Chr.2: 233744035-233744035 on Build GRCh38
rs763783144 Chr.2: 233743869-233743869 on Build GRCh38
rs139116240 Chr.2: 233744094-233744094 on Build GRCh38
rs907310484 Chr.2: 233744124-233744124 on Build GRCh38
rs1054717922 Chr.2: 233744074-233744074 on Build GRCh38
rs772404116 Chr.2: 233743889-233743889 on Build GRCh38
rs571958396 Chr.2: 233744167-233744167 on Build GRCh38
rs975143580 Chr.2: 233744268-233744268 on Build GRCh38
rs564002534 Chr.2: 233744210-233744210 on Build GRCh38
rs974356538 Chr.2: 233744164-233744164 on Build GRCh38
rs773166523 Chr.2: 233743848-233743848 on Build GRCh38
rs773777598 Chr.2: 233744022-233744022 on Build GRCh38
rs532921168 Chr.2: 233744024-233744024 on Build GRCh38
rs558062868 Chr.2: 233743944-233743944 on Build GRCh38
rs779214397 Chr.2: 233743846-233743846 on Build GRCh38
rs1026910418 Chr.2: 233744183-233744183 on Build GRCh38
rs867047175 Chr.2: 233743870-233743870 on Build GRCh38
rs371617871 Chr.2: 233743895-233743895 on Build GRCh38
rs757559777 Chr.2: 233743824-233743824 on Build GRCh38
rs769255302 Chr.2: 233743915-233743915 on Build GRCh38
rs1010440918 Chr.2: 233744112-233744112 on Build GRCh38
rs780594755 Chr.2: 233743875-233743875 on Build GRCh38
rs371517697 Chr.2: 233743959-233743959 on Build GRCh38
rs62191918 Chr.2: 233743836-233743836 on Build GRCh38
rs943816403 Chr.2: 233744235-233744235 on Build GRCh38
rs747210262 Chr.2: 233743958-233743958 on Build GRCh38
rs893316227 Chr.2: 233743988-233743988 on Build GRCh38
rs753368953 Chr.2: 233743820-233743820 on Build GRCh38
rs536470196 Chr.2: 233743921-233743921 on Build GRCh38
rs577435617 Chr.2: 233744231-233744231 on Build GRCh38
rs569722093 Chr.2: 233743832-233743832 on Build GRCh38
rs746819060 Chr.2: 233743891-233743891 on Build GRCh38
rs756502626 Chr.2: 233743872-233743872 on Build GRCh38
rs773932018 Chr.2: 233743927-233743927 on Build GRCh38
rs184316279 Chr.2: 233743956-233743956 on Build GRCh38
rs774501603 Chr.2: 233743864-233743864 on Build GRCh38
rs1040596623 Chr.2: 233743965-233743965 on Build GRCh38
rs752795670 Chr.2: 233743955-233743955 on Build GRCh38
rs374192063 Chr.2: 233743919-233743919 on Build GRCh38
rs555064194 Chr.2: 233744160-233744160 on Build GRCh38
rs988813505 Chr.2: 233743865-233743865 on Build GRCh38
rs752406673 Chr.2: 233743823-233743823 on Build GRCh38
rs755016650 Chr.2: 233743881-233743881 on Build GRCh38
rs189644754 Chr.2: 233743962-233743962 on Build GRCh38
rs60469444 Chr.2: 233743994-233743994 on Build GRCh38
rs186714678 Chr.2: 233744068-233744068 on Build GRCh38
rs375309664 Chr.2: 233743821-233743821 on Build GRCh38
rs754327674 Chr.2: 233743938-233743938 on Build GRCh38
rs368834284 Chr.2: 233743991-233743991 on Build GRCh38
rs765761325 Chr.2: 233743810-233743810 on Build GRCh38
rs373345166 Chr.2: 233743883-233743883 on Build GRCh38
rs774957959 Chr.2: 233743916-233743916 on Build GRCh38
rs201448352 Chr.2: 233743914-233743914 on Build GRCh38
rs79211437 Chr.2: 233744030-233744030 on Build GRCh38
rs541433916 Chr.2: 233743990-233743990 on Build GRCh38
rs754311427 Chr.2: 233743880-233743880 on Build GRCh38
rs556316328 Chr.2: 233744115-233744115 on Build GRCh38
rs540976533 Chr.2: 233744197-233744197 on Build GRCh38
rs375874968 Chr.2: 233743851-233743851 on Build GRCh38
rs556044106 Chr.2: 233743839-233743839 on Build GRCh38
rs764670005 Chr.2: 233743856-233743856 on Build GRCh38
rs76063448 Chr.2: 233743996-233743996 on Build GRCh38
rs576123874 Chr.2: 233743987-233743987 on Build GRCh38
rs900800021 Chr.2: 233743978-233743978 on Build GRCh38
rs181200285 Chr.2: 233744063-233744063 on Build GRCh38
rs6735370 Chr.2: 233743982-233743982 on Build GRCh38
rs929829035 Chr.2: 233744036-233744036 on Build GRCh38
rs767946429 Chr.2: 233743812-233743813 on Build GRCh38
rs772396813 Chr.2: 233744020-233744020 on Build GRCh38

View Full Product Details