Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 235750262-235750758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP1
Entrez Gene ID
116987
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
AGAP-1, CENTG2, GGAP1, cnt-g2
UniGene
Hs.435039
Gene Chromosome Location
Chr.2: 235494089-236131800 on Build GRCh38
Transcripts
NM_001037131.2 NM_001244888.1 NM_014914.4 XM_005246059.4 XM_006712234.3 XM_006712235.3 XM_006712237.3 XM_006712239.3 XM_011510547.2 XM_011510548.2 XM_011510549.2 XM_017003282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5006823 c.672C>T p.D224D Chr.2: 235750487-235750487 on Build GRCh38
1182180 c.584C>T p.A195V Chr.2: 235750399-235750399 on Build GRCh38
3579485 c.625G>A p.E209K Chr.2: 235750440-235750440 on Build GRCh38
6596012 c.542C>T p.A181V Chr.2: 235750357-235750357 on Build GRCh38
574311 c.618G>A p.T206T Chr.2: 235750433-235750433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113426089 Chr.2: 235750627-235750627 on Build GRCh38
rs942722376 Chr.2: 235750308-235750308 on Build GRCh38
rs750862874 Chr.2: 235750394-235750394 on Build GRCh38
rs753030538 Chr.2: 235750416-235750416 on Build GRCh38
rs970418764 Chr.2: 235750688-235750688 on Build GRCh38
rs757013176 Chr.2: 235750533-235750533 on Build GRCh38
rs142215390 Chr.2: 235750414-235750414 on Build GRCh38
rs766843373 Chr.2: 235750496-235750496 on Build GRCh38
rs568109133 Chr.2: 235750594-235750594 on Build GRCh38
rs76079209 Chr.2: 235750725-235750725 on Build GRCh38
rs569411849 Chr.2: 235750289-235750289 on Build GRCh38
rs780530427 Chr.2: 235750521-235750522 on Build GRCh38
rs573666540 Chr.2: 235750573-235750573 on Build GRCh38
rs367559858 Chr.2: 235750392-235750392 on Build GRCh38
rs760978302 Chr.2: 235750495-235750495 on Build GRCh38
rs148699717 Chr.2: 235750391-235750391 on Build GRCh38
rs143860937 Chr.2: 235750305-235750305 on Build GRCh38
rs139143512 Chr.2: 235750433-235750433 on Build GRCh38
rs149931123 Chr.2: 235750445-235750445 on Build GRCh38
rs774797485 Chr.2: 235750695-235750695 on Build GRCh38
rs773578859 Chr.2: 235750487-235750487 on Build GRCh38
rs556115042 Chr.2: 235750716-235750716 on Build GRCh38
rs759075492 Chr.2: 235750379-235750379 on Build GRCh38
rs185110377 Chr.2: 235750595-235750595 on Build GRCh38
rs994658768 Chr.2: 235750617-235750617 on Build GRCh38
rs867165100 Chr.2: 235750552-235750552 on Build GRCh38
rs777665614 Chr.2: 235750432-235750432 on Build GRCh38
rs373359973 Chr.2: 235750373-235750373 on Build GRCh38
rs555737698 Chr.2: 235750528-235750528 on Build GRCh38
rs142209544 Chr.2: 235750415-235750415 on Build GRCh38
rs375137075 Chr.2: 235750315-235750315 on Build GRCh38
rs112666282 Chr.2: 235750466-235750466 on Build GRCh38
rs201118697 Chr.2: 235750525-235750525 on Build GRCh38
rs576969179 Chr.2: 235750385-235750385 on Build GRCh38
rs374400240 Chr.2: 235750532-235750532 on Build GRCh38
rs553839639 Chr.2: 235750599-235750599 on Build GRCh38
rs552465885 Chr.2: 235750499-235750499 on Build GRCh38
rs769351419 Chr.2: 235750454-235750454 on Build GRCh38
rs968250082 Chr.2: 235750675-235750675 on Build GRCh38
rs923654950 Chr.2: 235750295-235750296 on Build GRCh38
rs770319108 Chr.2: 235750375-235750375 on Build GRCh38
rs143378661 Chr.2: 235750389-235750389 on Build GRCh38
rs375739545 Chr.2: 235750382-235750382 on Build GRCh38
rs368968053 Chr.2: 235750616-235750616 on Build GRCh38
rs1000543358 Chr.2: 235750720-235750720 on Build GRCh38
rs935195058 Chr.2: 235750299-235750299 on Build GRCh38
rs540369692 Chr.2: 235750358-235750358 on Build GRCh38
rs201642711 Chr.2: 235750497-235750497 on Build GRCh38
rs767569690 Chr.2: 235750402-235750402 on Build GRCh38
rs776154968 Chr.2: 235750376-235750376 on Build GRCh38
rs773013695 Chr.2: 235750363-235750363 on Build GRCh38
rs1044975277 Chr.2: 235750584-235750584 on Build GRCh38
rs752940419 Chr.2: 235750518-235750518 on Build GRCh38
rs947995823 Chr.2: 235750549-235750549 on Build GRCh38
rs542549618 Chr.2: 235750524-235750524 on Build GRCh38
rs182163679 Chr.2: 235750457-235750457 on Build GRCh38
rs755204272 Chr.2: 235750338-235750338 on Build GRCh38
rs151207771 Chr.2: 235750539-235750539 on Build GRCh38
rs985694363 Chr.2: 235750449-235750449 on Build GRCh38
rs1001130472 Chr.2: 235750604-235750604 on Build GRCh38
rs757618879 Chr.2: 235750314-235750314 on Build GRCh38
rs559207404 Chr.2: 235750288-235750288 on Build GRCh38
rs972855842 Chr.2: 235750425-235750425 on Build GRCh38
rs1007032736 Chr.2: 235750652-235750652 on Build GRCh38
rs77558625 Chr.2: 235750323-235750323 on Build GRCh38
rs189435652 Chr.2: 235750658-235750658 on Build GRCh38
rs756142948 Chr.2: 235750324-235750324 on Build GRCh38
rs374045675 Chr.2: 235750400-235750400 on Build GRCh38
rs373272693 Chr.2: 235750329-235750329 on Build GRCh38
rs753915768 Chr.2: 235750406-235750406 on Build GRCh38
rs553582318 Chr.2: 235750359-235750359 on Build GRCh38
rs200368421 Chr.2: 235750498-235750498 on Build GRCh38
rs895067411 Chr.2: 235750651-235750651 on Build GRCh38
rs560372408 Chr.2: 235750439-235750439 on Build GRCh38
rs767954745 Chr.2: 235750322-235750322 on Build GRCh38
rs371113525 Chr.2: 235750336-235750336 on Build GRCh38
rs922649441 Chr.2: 235750592-235750592 on Build GRCh38
rs200056479 Chr.2: 235750386-235750386 on Build GRCh38
rs377320529 Chr.2: 235750332-235750332 on Build GRCh38
rs772299964 Chr.2: 235750473-235750473 on Build GRCh38
rs909110825 Chr.2: 235750591-235750591 on Build GRCh38
rs1024647766 Chr.2: 235750684-235750684 on Build GRCh38
rs537859776 Chr.2: 235750293-235750293 on Build GRCh38
rs758267171 Chr.2: 235750417-235750417 on Build GRCh38
rs1000783207 Chr.2: 235750387-235750387 on Build GRCh38

View Full Product Details