Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 235799284-235799792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP1
Entrez Gene ID
116987
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
AGAP-1, CENTG2, GGAP1, cnt-g2
UniGene
Hs.435039
Gene Chromosome Location
Chr.2: 235494089-236131800 on Build GRCh38
Transcripts
NM_001037131.2 NM_001244888.1 NM_014914.4 XM_005246059.4 XM_006712234.3 XM_006712235.3 XM_006712237.3 XM_006712239.3 XM_011510547.2 XM_011510548.2 XM_011510549.2 XM_017003282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1018235 c.828C>T p.S276S Chr.2: 235799393-235799393 on Build GRCh38
1018236 c.922C>T p.R308C Chr.2: 235799487-235799487 on Build GRCh38
6290830 c.915G>C p.T305T Chr.2: 235799480-235799480 on Build GRCh38
5946566 c.860G>T p.S287I Chr.2: 235799425-235799425 on Build GRCh38
6291292 c.909G>A p.T303T Chr.2: 235799474-235799474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765674769 Chr.2: 235799319-235799319 on Build GRCh38
rs762329781 Chr.2: 235799483-235799483 on Build GRCh38
rs774442441 Chr.2: 235799380-235799380 on Build GRCh38
rs753181390 Chr.2: 235799328-235799328 on Build GRCh38
rs754181254 Chr.2: 235799506-235799506 on Build GRCh38
rs778991201 Chr.2: 235799447-235799447 on Build GRCh38
rs754790245 Chr.2: 235799514-235799514 on Build GRCh38
rs527561815 Chr.2: 235799738-235799738 on Build GRCh38
rs73998921 Chr.2: 235799561-235799561 on Build GRCh38
rs772010019 Chr.2: 235799534-235799534 on Build GRCh38
rs760137870 Chr.2: 235799469-235799469 on Build GRCh38
rs149256350 Chr.2: 235799473-235799473 on Build GRCh38
rs552661511 Chr.2: 235799749-235799749 on Build GRCh38
rs144632180 Chr.2: 235799412-235799412 on Build GRCh38
rs939001308 Chr.2: 235799540-235799540 on Build GRCh38
rs112968637 Chr.2: 235799445-235799445 on Build GRCh38
rs762087448 Chr.2: 235799559-235799559 on Build GRCh38
rs998252713 Chr.2: 235799737-235799737 on Build GRCh38
rs1031705967 Chr.2: 235799681-235799681 on Build GRCh38
rs201953851 Chr.2: 235799369-235799369 on Build GRCh38
rs745954616 Chr.2: 235799434-235799434 on Build GRCh38
rs1032154598 Chr.2: 235799443-235799443 on Build GRCh38
rs375623335 Chr.2: 235799552-235799552 on Build GRCh38
rs549232765 Chr.2: 235799555-235799555 on Build GRCh38
rs898684496 Chr.2: 235799596-235799596 on Build GRCh38
rs267599279 Chr.2: 235799457-235799457 on Build GRCh38
rs1028306481 Chr.2: 235799405-235799405 on Build GRCh38
rs572016525 Chr.2: 235799616-235799616 on Build GRCh38
rs965328894 Chr.2: 235799718-235799718 on Build GRCh38
rs886884617 Chr.2: 235799323-235799323 on Build GRCh38
rs757248532 Chr.2: 235799428-235799428 on Build GRCh38
rs965444369 Chr.2: 235799485-235799485 on Build GRCh38
rs866950982 Chr.2: 235799406-235799406 on Build GRCh38
rs750448988 Chr.2: 235799487-235799487 on Build GRCh38
rs1022518069 Chr.2: 235799719-235799719 on Build GRCh38
rs776589933 Chr.2: 235799688-235799688 on Build GRCh38
rs891408919 Chr.2: 235799639-235799639 on Build GRCh38
rs372775296 Chr.2: 235799549-235799549 on Build GRCh38
rs561623752 Chr.2: 235799711-235799711 on Build GRCh38
rs760425326 Chr.2: 235799408-235799408 on Build GRCh38
rs552995872 Chr.2: 235799409-235799409 on Build GRCh38
rs760804235 Chr.2: 235799570-235799570 on Build GRCh38
rs748157624 Chr.2: 235799450-235799450 on Build GRCh38
rs575180547 Chr.2: 235799463-235799463 on Build GRCh38
rs761395219 Chr.2: 235799390-235799390 on Build GRCh38
rs1028398853 Chr.2: 235799759-235799759 on Build GRCh38
rs757374729 Chr.2: 235799351-235799351 on Build GRCh38
rs763294074 Chr.2: 235799474-235799474 on Build GRCh38
rs753970882 Chr.2: 235799364-235799364 on Build GRCh38
rs770933913 Chr.2: 235799562-235799562 on Build GRCh38
rs536214577 Chr.2: 235799708-235799708 on Build GRCh38
rs777579052 Chr.2: 235799539-235799539 on Build GRCh38
rs749074252 Chr.2: 235799312-235799312 on Build GRCh38
rs141716710 Chr.2: 235799494-235799494 on Build GRCh38
rs529253565 Chr.2: 235799548-235799548 on Build GRCh38
rs551994813 Chr.2: 235799607-235799607 on Build GRCh38
rs1054863712 Chr.2: 235799566-235799566 on Build GRCh38
rs926568815 Chr.2: 235799536-235799536 on Build GRCh38
rs146005443 Chr.2: 235799497-235799497 on Build GRCh38
rs1038908122 Chr.2: 235799578-235799578 on Build GRCh38
rs1003260240 Chr.2: 235799332-235799332 on Build GRCh38
rs550583166 Chr.2: 235799663-235799663 on Build GRCh38
rs867329900 Chr.2: 235799403-235799404 on Build GRCh38
rs2292708 Chr.2: 235799522-235799522 on Build GRCh38
rs567164264 Chr.2: 235799692-235799692 on Build GRCh38
rs895001999 Chr.2: 235799567-235799567 on Build GRCh38
rs752124359 Chr.2: 235799423-235799423 on Build GRCh38
rs535948190 Chr.2: 235799398-235799398 on Build GRCh38
rs114931553 Chr.2: 235799362-235799362 on Build GRCh38
rs372555700 Chr.2: 235799335-235799335 on Build GRCh38
rs369405118 Chr.2: 235799488-235799488 on Build GRCh38
rs573706698 Chr.2: 235799394-235799394 on Build GRCh38
rs189491188 Chr.2: 235799464-235799464 on Build GRCh38
rs997106060 Chr.2: 235799393-235799393 on Build GRCh38
rs537320978 Chr.2: 235799654-235799654 on Build GRCh38
rs982745709 Chr.2: 235799480-235799480 on Build GRCh38
rs758857198 Chr.2: 235799334-235799334 on Build GRCh38
rs778903333 Chr.2: 235799516-235799516 on Build GRCh38
rs768645780 Chr.2: 235799379-235799379 on Build GRCh38
rs771462060 Chr.2: 235799458-235799458 on Build GRCh38
rs770552808 Chr.2: 235799472-235799472 on Build GRCh38
rs764529810 Chr.2: 235799416-235799416 on Build GRCh38
rs781379317 Chr.2: 235799429-235799429 on Build GRCh38
rs1015563900 Chr.2: 235799717-235799717 on Build GRCh38
rs562548507 Chr.2: 235799360-235799360 on Build GRCh38
rs559839614 Chr.2: 235799556-235799556 on Build GRCh38
rs369599695 Chr.2: 235799414-235799414 on Build GRCh38
rs954394727 Chr.2: 235799763-235799763 on Build GRCh38
rs564880147 Chr.2: 235799446-235799446 on Build GRCh38
rs1028568180 Chr.2: 235799627-235799627 on Build GRCh38
rs767939918 Chr.2: 235799484-235799484 on Build GRCh38
rs371029126 Chr.2: 235799477-235799477 on Build GRCh38

View Full Product Details