Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237341874-237342373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
UniGene
Hs.233240
Gene Chromosome Location
Chr.2: 237324012-237434822 on Build GRCh38
Transcripts
NM_004369.3 NM_057166.4 NM_057167.3 XM_005246065.1 XM_005246066.1 XM_006712253.1 XM_011510574.1 XM_017003303.1 XM_017003304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406565 c.7765+2T>C p.? Chr.2: 237342063-237342063 on Build GRCh38
720110 c.7757T>C p.V2586A Chr.2: 237342073-237342073 on Build GRCh38
98216 c.7741G>A p.V2581I Chr.2: 237342089-237342089 on Build GRCh38
1406566 c.7717G>T p.D2573Y Chr.2: 237342113-237342113 on Build GRCh38
4788004 c.7765+2T>G p.? Chr.2: 237342063-237342063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145581705 Chr.2: 237342075-237342075 on Build GRCh38
rs765388693 Chr.2: 237342007-237342007 on Build GRCh38
rs764193290 Chr.2: 237342163-237342163 on Build GRCh38
rs376637667 Chr.2: 237342039-237342039 on Build GRCh38
rs773416790 Chr.2: 237342044-237342044 on Build GRCh38
rs930558902 Chr.2: 237342320-237342320 on Build GRCh38
rs112386121 Chr.2: 237341939-237341939 on Build GRCh38
rs543795014 Chr.2: 237341937-237341938 on Build GRCh38
rs779667683 Chr.2: 237342121-237342121 on Build GRCh38
rs574975784 Chr.2: 237342177-237342177 on Build GRCh38
rs1015920317 Chr.2: 237342284-237342284 on Build GRCh38
rs886043891 Chr.2: 237342102-237342102 on Build GRCh38
rs794727598 Chr.2: 237342124-237342124 on Build GRCh38
rs992035159 Chr.2: 237342269-237342269 on Build GRCh38
rs980856765 Chr.2: 237341957-237341957 on Build GRCh38
rs761150742 Chr.2: 237342048-237342048 on Build GRCh38
rs59232721 Chr.2: 237342196-237342196 on Build GRCh38
rs567192927 Chr.2: 237342182-237342182 on Build GRCh38
rs758506738 Chr.2: 237341980-237341982 on Build GRCh38
rs200456255 Chr.2: 237342135-237342135 on Build GRCh38
rs548961023 Chr.2: 237341943-237341943 on Build GRCh38
rs886043033 Chr.2: 237342076-237342076 on Build GRCh38
rs766781765 Chr.2: 237342049-237342049 on Build GRCh38
rs780964113 Chr.2: 237342143-237342143 on Build GRCh38
rs960587236 Chr.2: 237341972-237341972 on Build GRCh38
rs1041006651 Chr.2: 237342139-237342139 on Build GRCh38
rs992187883 Chr.2: 237342153-237342153 on Build GRCh38
rs756140209 Chr.2: 237342117-237342117 on Build GRCh38
rs745738184 Chr.2: 237342107-237342107 on Build GRCh38
rs143631346 Chr.2: 237342145-237342145 on Build GRCh38
rs765245863 Chr.2: 237342073-237342073 on Build GRCh38
rs889053037 Chr.2: 237342344-237342344 on Build GRCh38
rs140021275 Chr.2: 237342082-237342082 on Build GRCh38
rs528635041 Chr.2: 237341922-237341922 on Build GRCh38
rs886055804 Chr.2: 237342139-237342139 on Build GRCh38
rs780850057 Chr.2: 237342086-237342086 on Build GRCh38
rs144765607 Chr.2: 237342000-237342000 on Build GRCh38
rs971999657 Chr.2: 237342172-237342172 on Build GRCh38
rs140635358 Chr.2: 237341940-237341940 on Build GRCh38
rs776994369 Chr.2: 237342144-237342144 on Build GRCh38
rs146872525 Chr.2: 237342160-237342160 on Build GRCh38
rs758582935 Chr.2: 237342081-237342081 on Build GRCh38
rs34137614 Chr.2: 237342268-237342269 on Build GRCh38
rs777263519 Chr.2: 237342050-237342050 on Build GRCh38
rs755632634 Chr.2: 237342258-237342258 on Build GRCh38
rs1009122752 Chr.2: 237342344-237342345 on Build GRCh38
rs1022266665 Chr.2: 237342157-237342157 on Build GRCh38
rs751294512 Chr.2: 237342084-237342084 on Build GRCh38
rs919559147 Chr.2: 237342311-237342311 on Build GRCh38
rs995197003 Chr.2: 237341946-237341946 on Build GRCh38
rs750270909 Chr.2: 237342206-237342206 on Build GRCh38
rs1050802138 Chr.2: 237342330-237342330 on Build GRCh38
rs557375944 Chr.2: 237341998-237341998 on Build GRCh38
rs1033018695 Chr.2: 237342141-237342141 on Build GRCh38
rs879912909 Chr.2: 237342266-237342266 on Build GRCh38
rs34668305 Chr.2: 237341959-237341959 on Build GRCh38
rs961356271 Chr.2: 237342306-237342306 on Build GRCh38
rs201479636 Chr.2: 237342128-237342128 on Build GRCh38
rs908480457 Chr.2: 237342006-237342006 on Build GRCh38
rs1009592477 Chr.2: 237341903-237341903 on Build GRCh38
rs973963137 Chr.2: 237342309-237342309 on Build GRCh38
rs760040712 Chr.2: 237342071-237342071 on Build GRCh38
rs201840494 Chr.2: 237342016-237342016 on Build GRCh38
rs956895139 Chr.2: 237341901-237341901 on Build GRCh38
rs1009561726 Chr.2: 237342154-237342154 on Build GRCh38
rs1009082593 Chr.2: 237342122-237342122 on Build GRCh38
rs377202620 Chr.2: 237342013-237342013 on Build GRCh38
rs747330663 Chr.2: 237342136-237342136 on Build GRCh38
rs763071900 Chr.2: 237342164-237342164 on Build GRCh38
rs188020704 Chr.2: 237342195-237342195 on Build GRCh38

View Full Product Details