Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238447079-238447578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB1
Entrez Gene ID
51665
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
ASB-1
UniGene
Hs.516788
Gene Chromosome Location
Chr.2: 238426741-238452250 on Build GRCh38
Transcripts
NM_001040445.1 XM_005246080.1

SNP Detail

SNP ID Chromosome Location
rs139458604 Chr.2: 238447501-238447501 on Build GRCh38
rs543250406 Chr.2: 238447558-238447558 on Build GRCh38
rs958859787 Chr.2: 238447196-238447196 on Build GRCh38
rs930763208 Chr.2: 238447274-238447274 on Build GRCh38
rs1053602561 Chr.2: 238447264-238447264 on Build GRCh38
rs913239413 Chr.2: 238447443-238447443 on Build GRCh38
rs1035332946 Chr.2: 238447139-238447139 on Build GRCh38
rs2340870 Chr.2: 238447385-238447385 on Build GRCh38
rs566514638 Chr.2: 238447123-238447123 on Build GRCh38
rs772273713 Chr.2: 238447185-238447185 on Build GRCh38
rs919314145 Chr.2: 238447258-238447258 on Build GRCh38
rs937528531 Chr.2: 238447498-238447498 on Build GRCh38
rs577636953 Chr.2: 238447398-238447398 on Build GRCh38
rs775130388 Chr.2: 238447446-238447446 on Build GRCh38
rs927412157 Chr.2: 238447480-238447480 on Build GRCh38
rs889151196 Chr.2: 238447295-238447295 on Build GRCh38
rs1035952202 Chr.2: 238447330-238447330 on Build GRCh38
rs773316768 Chr.2: 238447226-238447226 on Build GRCh38
rs991457134 Chr.2: 238447197-238447197 on Build GRCh38
rs947472977 Chr.2: 238447468-238447468 on Build GRCh38
rs538720391 Chr.2: 238447415-238447415 on Build GRCh38
rs967085358 Chr.2: 238447349-238447349 on Build GRCh38
rs1036204848 Chr.2: 238447257-238447257 on Build GRCh38
rs149702312 Chr.2: 238447526-238447526 on Build GRCh38
rs749853114 Chr.2: 238447112-238447112 on Build GRCh38
rs952652173 Chr.2: 238447182-238447182 on Build GRCh38
rs949827271 Chr.2: 238447255-238447255 on Build GRCh38
rs971867385 Chr.2: 238447416-238447416 on Build GRCh38
rs917085953 Chr.2: 238447241-238447241 on Build GRCh38
rs1049514379 Chr.2: 238447287-238447287 on Build GRCh38
rs776322300 Chr.2: 238447439-238447439 on Build GRCh38
rs1025796368 Chr.2: 238447357-238447357 on Build GRCh38
rs555221830 Chr.2: 238447447-238447447 on Build GRCh38
rs919115023 Chr.2: 238447441-238447441 on Build GRCh38
rs477041 Chr.2: 238447354-238447354 on Build GRCh38
rs1040359455 Chr.2: 238447545-238447545 on Build GRCh38
rs952522054 Chr.2: 238447379-238447379 on Build GRCh38
rs1021089840 Chr.2: 238447337-238447337 on Build GRCh38
rs896093664 Chr.2: 238447557-238447557 on Build GRCh38
rs778366144 Chr.2: 238447143-238447143 on Build GRCh38
rs1001801999 Chr.2: 238447124-238447124 on Build GRCh38
rs902072646 Chr.2: 238447553-238447553 on Build GRCh38
rs947531258 Chr.2: 238447301-238447301 on Build GRCh38
rs1009653951 Chr.2: 238447326-238447326 on Build GRCh38
rs1043167206 Chr.2: 238447477-238447477 on Build GRCh38
rs534028618 Chr.2: 238447129-238447129 on Build GRCh38
rs910539110 Chr.2: 238447499-238447499 on Build GRCh38
rs1008216317 Chr.2: 238447304-238447304 on Build GRCh38
rs188757308 Chr.2: 238447388-238447388 on Build GRCh38
rs779111519 Chr.2: 238447155-238447155 on Build GRCh38
rs746754209 Chr.2: 238447309-238447309 on Build GRCh38
rs988859275 Chr.2: 238447437-238447437 on Build GRCh38
rs182248338 Chr.2: 238447165-238447165 on Build GRCh38
rs746053297 Chr.2: 238447386-238447386 on Build GRCh38
rs999979801 Chr.2: 238447353-238447353 on Build GRCh38
rs185587812 Chr.2: 238447305-238447305 on Build GRCh38
rs911124533 Chr.2: 238447223-238447223 on Build GRCh38
rs942590993 Chr.2: 238447232-238447232 on Build GRCh38
rs1040667001 Chr.2: 238447308-238447308 on Build GRCh38
rs891915934 Chr.2: 238447291-238447291 on Build GRCh38
rs954869789 Chr.2: 238447417-238447417 on Build GRCh38
rs906145936 Chr.2: 238447119-238447119 on Build GRCh38

View Full Product Details