Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238449357-238449874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB1
Entrez Gene ID
51665
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
ASB-1
UniGene
Hs.516788
Gene Chromosome Location
Chr.2: 238426741-238452250 on Build GRCh38
Transcripts
NM_001040445.1 XM_005246080.1

SNP Detail

SNP ID Chromosome Location
rs759901883 Chr.2: 238449651-238449651 on Build GRCh38
rs2340869 Chr.2: 238449470-238449470 on Build GRCh38
rs983335396 Chr.2: 238449697-238449697 on Build GRCh38
rs559419212 Chr.2: 238449661-238449661 on Build GRCh38
rs1008434950 Chr.2: 238449616-238449616 on Build GRCh38
rs907812232 Chr.2: 238449498-238449498 on Build GRCh38
rs757342404 Chr.2: 238449790-238449790 on Build GRCh38
rs1029803256 Chr.2: 238449601-238449601 on Build GRCh38
rs530116513 Chr.2: 238449699-238449699 on Build GRCh38
rs75472009 Chr.2: 238449715-238449715 on Build GRCh38
rs771796246 Chr.2: 238449542-238449542 on Build GRCh38
rs956436390 Chr.2: 238449637-238449637 on Build GRCh38
rs1049844192 Chr.2: 238449577-238449577 on Build GRCh38
rs932687990 Chr.2: 238449763-238449763 on Build GRCh38
rs776040374 Chr.2: 238449703-238449703 on Build GRCh38
rs540636 Chr.2: 238449700-238449700 on Build GRCh38
rs1024767398 Chr.2: 238449654-238449654 on Build GRCh38
rs751570122 Chr.2: 238449780-238449780 on Build GRCh38
rs548429153 Chr.2: 238449507-238449511 on Build GRCh38
rs780492586 Chr.2: 238449506-238449507 on Build GRCh38
rs575689536 Chr.2: 238449467-238449467 on Build GRCh38
rs989445080 Chr.2: 238449393-238449393 on Build GRCh38
rs959081137 Chr.2: 238449397-238449397 on Build GRCh38
rs1038789353 Chr.2: 238449750-238449750 on Build GRCh38
rs1031075109 Chr.2: 238449635-238449635 on Build GRCh38
rs969314837 Chr.2: 238449679-238449679 on Build GRCh38
rs1050268200 Chr.2: 238449768-238449768 on Build GRCh38
rs941606352 Chr.2: 238449531-238449531 on Build GRCh38
rs755490681 Chr.2: 238449646-238449647 on Build GRCh38
rs1038580531 Chr.2: 238449540-238449540 on Build GRCh38
rs763425231 Chr.2: 238449708-238449708 on Build GRCh38
rs200257921 Chr.2: 238449465-238449465 on Build GRCh38
rs1042529647 Chr.2: 238449827-238449827 on Build GRCh38
rs554232931 Chr.2: 238449451-238449451 on Build GRCh38
rs768425493 Chr.2: 238449643-238449646 on Build GRCh38
rs891233334 Chr.2: 238449791-238449791 on Build GRCh38
rs956818919 Chr.2: 238449390-238449390 on Build GRCh38
rs989561080 Chr.2: 238449640-238449640 on Build GRCh38
rs887150726 Chr.2: 238449521-238449521 on Build GRCh38
rs1044934583 Chr.2: 238449452-238449452 on Build GRCh38
rs146798748 Chr.2: 238449835-238449835 on Build GRCh38
rs908624561 Chr.2: 238449735-238449735 on Build GRCh38
rs1022185526 Chr.2: 238449666-238449666 on Build GRCh38
rs73092608 Chr.2: 238449384-238449384 on Build GRCh38
rs568533897 Chr.2: 238449642-238449643 on Build GRCh38
rs183146890 Chr.2: 238449639-238449639 on Build GRCh38
rs902699531 Chr.2: 238449847-238449847 on Build GRCh38
rs866673462 Chr.2: 238449602-238449602 on Build GRCh38
rs532224094 Chr.2: 238449631-238449631 on Build GRCh38
rs915056002 Chr.2: 238449432-238449432 on Build GRCh38
rs944190166 Chr.2: 238449795-238449795 on Build GRCh38
rs530997231 Chr.2: 238449718-238449718 on Build GRCh38
rs58365110 Chr.2: 238449811-238449811 on Build GRCh38
rs941336696 Chr.2: 238449736-238449736 on Build GRCh38
rs996940183 Chr.2: 238449576-238449576 on Build GRCh38
rs187075452 Chr.2: 238449713-238449713 on Build GRCh38
rs947837369 Chr.2: 238449438-238449438 on Build GRCh38
rs565064862 Chr.2: 238449500-238449500 on Build GRCh38
rs530298373 Chr.2: 238449826-238449826 on Build GRCh38

View Full Product Details