Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240560728-240561230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1 NM_001033575.1

Gene Symbol
DUSP28
Entrez Gene ID
285193
Gene Name
dual specificity phosphatase 28
Gene Aliases
DUSP26, VHP
UniGene
Hs.369297
Gene Chromosome Location
Chr.2: 240560054-240564014 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1 NM_001033575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679730 c.238G>A p.A80T Chr.2: 240560922-240560922 on Build GRCh38
5354243 c.379G>A p.A127T Chr.2: 240561063-240561063 on Build GRCh38
6090827 c.66C>G p.V22V Chr.2: 240560750-240560750 on Build GRCh38
6157407 c.329C>T p.S110L Chr.2: 240561013-240561013 on Build GRCh38
5502650 c.98C>T p.A33V Chr.2: 240560782-240560782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990440994 Chr.2: 240560951-240560951 on Build GRCh38
rs1007823553 Chr.2: 240560966-240560966 on Build GRCh38
rs560521658 Chr.2: 240560865-240560865 on Build GRCh38
rs771443330 Chr.2: 240561101-240561101 on Build GRCh38
rs916164512 Chr.2: 240560972-240560972 on Build GRCh38
rs754396712 Chr.2: 240561000-240561000 on Build GRCh38
rs982096102 Chr.2: 240561016-240561016 on Build GRCh38
rs900417524 Chr.2: 240560877-240560877 on Build GRCh38
rs542332202 Chr.2: 240560815-240560815 on Build GRCh38
rs756447114 Chr.2: 240560804-240560804 on Build GRCh38
rs758640402 Chr.2: 240561009-240561009 on Build GRCh38
rs781763546 Chr.2: 240560786-240560786 on Build GRCh38
rs1003154166 Chr.2: 240561153-240561153 on Build GRCh38
rs907902179 Chr.2: 240561019-240561019 on Build GRCh38
rs997353268 Chr.2: 240560886-240560886 on Build GRCh38
rs777744662 Chr.2: 240560963-240560963 on Build GRCh38
rs1036393246 Chr.2: 240561156-240561156 on Build GRCh38
rs765590102 Chr.2: 240561005-240561005 on Build GRCh38
rs571403854 Chr.2: 240561064-240561064 on Build GRCh38
rs868495061 Chr.2: 240560943-240560943 on Build GRCh38
rs763623217 Chr.2: 240560980-240560980 on Build GRCh38
rs780192561 Chr.2: 240561084-240561084 on Build GRCh38
rs754428063 Chr.2: 240561093-240561093 on Build GRCh38
rs114887011 Chr.2: 240560869-240560869 on Build GRCh38
rs755399446 Chr.2: 240560934-240560934 on Build GRCh38
rs957972647 Chr.2: 240560948-240560948 on Build GRCh38
rs933196842 Chr.2: 240561020-240561020 on Build GRCh38
rs780978886 Chr.2: 240561032-240561032 on Build GRCh38
rs1032543680 Chr.2: 240561040-240561040 on Build GRCh38
rs553554660 Chr.2: 240560769-240560769 on Build GRCh38
rs796295010 Chr.2: 240561075-240561091 on Build GRCh38
rs760505115 Chr.2: 240560753-240560753 on Build GRCh38
rs754471524 Chr.2: 240561133-240561133 on Build GRCh38
rs371287024 Chr.2: 240561134-240561134 on Build GRCh38
rs776672030 Chr.2: 240560970-240560970 on Build GRCh38
rs111241666 Chr.2: 240561147-240561147 on Build GRCh38
rs532600172 Chr.2: 240560858-240560881 on Build GRCh38
rs902421764 Chr.2: 240561099-240561099 on Build GRCh38
rs935202834 Chr.2: 240561127-240561127 on Build GRCh38
rs774507085 Chr.2: 240560874-240560874 on Build GRCh38
rs547966255 Chr.2: 240560995-240560995 on Build GRCh38
rs756146137 Chr.2: 240561083-240561083 on Build GRCh38
rs756613284 Chr.2: 240560913-240560913 on Build GRCh38
rs1039013938 Chr.2: 240561012-240561012 on Build GRCh38
rs764424498 Chr.2: 240560876-240560877 on Build GRCh38
rs781430049 Chr.2: 240561078-240561078 on Build GRCh38
rs920653506 Chr.2: 240561150-240561150 on Build GRCh38
rs752233645 Chr.2: 240560778-240560778 on Build GRCh38
rs754117685 Chr.2: 240560924-240560924 on Build GRCh38
rs544076250 Chr.2: 240560891-240560891 on Build GRCh38
rs13407858 Chr.2: 240560872-240560872 on Build GRCh38
rs755273079 Chr.2: 240561003-240561003 on Build GRCh38
rs889715010 Chr.2: 240560952-240560952 on Build GRCh38
rs767063982 Chr.2: 240561062-240561062 on Build GRCh38
rs933290295 Chr.2: 240561093-240561093 on Build GRCh38
rs753036166 Chr.2: 240561073-240561073 on Build GRCh38
rs746914734 Chr.2: 240560967-240560967 on Build GRCh38
rs1036859605 Chr.2: 240560914-240560914 on Build GRCh38
rs770754483 Chr.2: 240560969-240560969 on Build GRCh38
rs950957973 Chr.2: 240561170-240561170 on Build GRCh38
rs757425889 Chr.2: 240561025-240561025 on Build GRCh38
rs10933615 Chr.2: 240561091-240561091 on Build GRCh38
rs1023839799 Chr.2: 240561110-240561110 on Build GRCh38
rs761212547 Chr.2: 240561057-240561057 on Build GRCh38
rs768988257 Chr.2: 240561190-240561193 on Build GRCh38
rs948925868 Chr.2: 240560823-240560823 on Build GRCh38
rs747836528 Chr.2: 240561096-240561096 on Build GRCh38
rs1053588622 Chr.2: 240561128-240561128 on Build GRCh38
rs758876457 Chr.2: 240561025-240561043 on Build GRCh38
rs1015181376 Chr.2: 240561142-240561142 on Build GRCh38
rs201793117 Chr.2: 240561039-240561039 on Build GRCh38
rs906236717 Chr.2: 240561152-240561152 on Build GRCh38
rs978263411 Chr.2: 240560818-240560818 on Build GRCh38
rs777055749 Chr.2: 240561063-240561063 on Build GRCh38
rs867347548 Chr.2: 240561028-240561028 on Build GRCh38
rs565477343 Chr.2: 240560918-240560918 on Build GRCh38
rs1011925554 Chr.2: 240561140-240561140 on Build GRCh38
rs750953578 Chr.2: 240560792-240560792 on Build GRCh38
rs571854171 Chr.2: 240561013-240561013 on Build GRCh38
rs542291327 Chr.2: 240561129-240561129 on Build GRCh38
rs960942830 Chr.2: 240560962-240560962 on Build GRCh38
rs1004026924 Chr.2: 240560765-240560765 on Build GRCh38
rs1023226275 Chr.2: 240561051-240561051 on Build GRCh38
rs748219279 Chr.2: 240560855-240560855 on Build GRCh38
rs760226423 Chr.2: 240561121-240561121 on Build GRCh38
rs1016404757 Chr.2: 240561180-240561180 on Build GRCh38
rs1019261311 Chr.2: 240560968-240560968 on Build GRCh38
rs886210827 Chr.2: 240561095-240561111 on Build GRCh38
rs575983369 Chr.2: 240561196-240561196 on Build GRCh38
rs991108921 Chr.2: 240560854-240560854 on Build GRCh38
rs564591872 Chr.2: 240560863-240560863 on Build GRCh38
rs74753884 Chr.2: 240561190-240561190 on Build GRCh38
rs766787898 Chr.2: 240560999-240560999 on Build GRCh38
rs1045835139 Chr.2: 240560824-240560824 on Build GRCh38
rs929351915 Chr.2: 240560832-240560832 on Build GRCh38
rs769649960 Chr.2: 240561041-240561041 on Build GRCh38
rs973443326 Chr.2: 240561148-240561148 on Build GRCh38
rs772729836 Chr.2: 240561102-240561102 on Build GRCh38
rs776958679 Chr.2: 240560898-240560909 on Build GRCh38
rs537091038 Chr.2: 240561043-240561043 on Build GRCh38
rs758023880 Chr.2: 240560779-240560779 on Build GRCh38
rs983634963 Chr.2: 240561079-240561079 on Build GRCh38
rs761424683 Chr.2: 240560996-240560996 on Build GRCh38
rs769769611 Chr.2: 240560975-240560975 on Build GRCh38
rs752876567 Chr.2: 240561008-240561008 on Build GRCh38
rs768174886 Chr.2: 240561049-240561049 on Build GRCh38
rs759401384 Chr.2: 240560971-240560971 on Build GRCh38
rs571769896 Chr.2: 240560810-240560810 on Build GRCh38
rs940800288 Chr.2: 240560895-240560895 on Build GRCh38
rs556419229 Chr.2: 240561048-240561048 on Build GRCh38
rs1027945442 Chr.2: 240560778-240560778 on Build GRCh38
rs764953267 Chr.2: 240560888-240560888 on Build GRCh38
rs750878881 Chr.2: 240560903-240560903 on Build GRCh38
rs751867093 Chr.2: 240561076-240561076 on Build GRCh38
rs762296334 Chr.2: 240560929-240560949 on Build GRCh38
rs763529689 Chr.2: 240560893-240560893 on Build GRCh38
rs772048349 Chr.2: 240560749-240560749 on Build GRCh38
rs1030136409 Chr.2: 240560901-240560901 on Build GRCh38
rs774117553 Chr.2: 240561053-240561053 on Build GRCh38
rs779143635 Chr.2: 240560849-240560849 on Build GRCh38
rs768842256 Chr.2: 240560841-240560841 on Build GRCh38
rs966879537 Chr.2: 240560816-240560816 on Build GRCh38
rs749489665 Chr.2: 240560814-240560814 on Build GRCh38
rs765645674 Chr.2: 240560955-240560959 on Build GRCh38
rs201249226 Chr.2: 240561074-240561074 on Build GRCh38
rs773089505 Chr.2: 240560750-240560750 on Build GRCh38
rs977795360 Chr.2: 240560774-240560774 on Build GRCh38
rs866376996 Chr.2: 240560831-240560831 on Build GRCh38
rs889680518 Chr.2: 240561058-240561058 on Build GRCh38
rs770718310 Chr.2: 240560873-240560873 on Build GRCh38
rs530184793 Chr.2: 240561011-240561011 on Build GRCh38
rs538932275 Chr.2: 240561070-240561070 on Build GRCh38
rs928770386 Chr.2: 240560982-240560982 on Build GRCh38
rs767410576 Chr.2: 240560747-240560747 on Build GRCh38
rs1051616309 Chr.2: 240561030-240561030 on Build GRCh38
rs772072452 Chr.2: 240560949-240560949 on Build GRCh38
rs963475709 Chr.2: 240561205-240561205 on Build GRCh38
rs762702910 Chr.2: 240560979-240560979 on Build GRCh38
rs112543214 Chr.2: 240560766-240560766 on Build GRCh38
rs759454501 Chr.2: 240560884-240560884 on Build GRCh38
rs1047739442 Chr.2: 240560847-240560847 on Build GRCh38
rs768550670 Chr.2: 240561085-240561085 on Build GRCh38
rs984033615 Chr.2: 240561172-240561172 on Build GRCh38
rs745764545 Chr.2: 240561082-240561082 on Build GRCh38
rs766793961 Chr.2: 240561063-240561064 on Build GRCh38
rs767086994 Chr.2: 240561189-240561190 on Build GRCh38
rs766100450 Chr.2: 240560761-240560761 on Build GRCh38
rs761928968 Chr.2: 240560746-240560746 on Build GRCh38
rs373908074 Chr.2: 240560941-240560941 on Build GRCh38
rs775703352 Chr.2: 240560760-240560761 on Build GRCh38
rs924913056 Chr.2: 240560782-240560782 on Build GRCh38
rs566287242 Chr.2: 240561010-240561010 on Build GRCh38
rs762539689 Chr.2: 240560892-240560892 on Build GRCh38
rs772193176 Chr.2: 240560862-240560862 on Build GRCh38
rs370148084 Chr.2: 240561069-240561069 on Build GRCh38
rs866357062 Chr.2: 240561038-240561038 on Build GRCh38
rs775229532 Chr.2: 240560976-240560976 on Build GRCh38
rs751142968 Chr.2: 240561076-240561091 on Build GRCh38
rs750977042 Chr.2: 240561008-240561009 on Build GRCh38
rs974754904 Chr.2: 240561087-240561087 on Build GRCh38

View Full Product Details