Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240589260-240589761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN10
Entrez Gene ID
11132
Gene Name
calpain 10
Gene Aliases
CANP10, NIDDM1
UniGene
Hs.728234
Gene Chromosome Location
Chr.2: 240586716-240599109 on Build GRCh38
Transcripts
NM_023083.3 NM_023085.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157402 c.180A>T p.P60P Chr.2: 240589381-240589381 on Build GRCh38
259924 c.237C>T p.C79C Chr.2: 240589438-240589438 on Build GRCh38
3579899 c.170C>T p.P57L Chr.2: 240589371-240589371 on Build GRCh38
2909635 c.273+4C>T p.? Chr.2: 240589478-240589478 on Build GRCh38
1692255 c.194T>G p.V65G Chr.2: 240589395-240589395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189777246 Chr.2: 240589557-240589557 on Build GRCh38
rs768069970 Chr.2: 240589515-240589515 on Build GRCh38
rs1038103693 Chr.2: 240589431-240589431 on Build GRCh38
rs1023231247 Chr.2: 240589489-240589489 on Build GRCh38
rs936074337 Chr.2: 240589732-240589732 on Build GRCh38
rs559918880 Chr.2: 240589536-240589536 on Build GRCh38
rs1035485692 Chr.2: 240589665-240589665 on Build GRCh38
rs539306792 Chr.2: 240589294-240589294 on Build GRCh38
rs761473089 Chr.2: 240589470-240589470 on Build GRCh38
rs558390096 Chr.2: 240589678-240589678 on Build GRCh38
rs759781981 Chr.2: 240589475-240589475 on Build GRCh38
rs553784127 Chr.2: 240589572-240589572 on Build GRCh38
rs142380588 Chr.2: 240589658-240589658 on Build GRCh38
rs758637318 Chr.2: 240589490-240589490 on Build GRCh38
rs763206424 Chr.2: 240589332-240589332 on Build GRCh38
rs1009140284 Chr.2: 240589505-240589505 on Build GRCh38
rs566234266 Chr.2: 240589311-240589311 on Build GRCh38
rs753287892 Chr.2: 240589522-240589522 on Build GRCh38
rs982732066 Chr.2: 240589566-240589566 on Build GRCh38
rs764779084 Chr.2: 240589575-240589575 on Build GRCh38
rs778941616 Chr.2: 240589421-240589421 on Build GRCh38
rs12620571 Chr.2: 240589662-240589662 on Build GRCh38
rs3792263 Chr.2: 240589731-240589731 on Build GRCh38
rs758590960 Chr.2: 240589438-240589438 on Build GRCh38
rs376516872 Chr.2: 240589329-240589329 on Build GRCh38
rs911869925 Chr.2: 240589576-240589576 on Build GRCh38
rs1050748679 Chr.2: 240589580-240589580 on Build GRCh38
rs1053605474 Chr.2: 240589658-240589659 on Build GRCh38
rs201581519 Chr.2: 240589478-240589478 on Build GRCh38
rs944729963 Chr.2: 240589679-240589679 on Build GRCh38
rs749938829 Chr.2: 240589602-240589602 on Build GRCh38
rs372834354 Chr.2: 240589491-240589492 on Build GRCh38
rs773980754 Chr.2: 240589467-240589467 on Build GRCh38
rs756667300 Chr.2: 240589388-240589388 on Build GRCh38
rs778512049 Chr.2: 240589358-240589358 on Build GRCh38
rs200982375 Chr.2: 240589447-240589447 on Build GRCh38
rs1022398827 Chr.2: 240589530-240589530 on Build GRCh38
rs781112808 Chr.2: 240589442-240589442 on Build GRCh38
rs199590817 Chr.2: 240589304-240589304 on Build GRCh38
rs980837355 Chr.2: 240589664-240589664 on Build GRCh38
rs781061292 Chr.2: 240589516-240589516 on Build GRCh38
rs139462940 Chr.2: 240589370-240589370 on Build GRCh38
rs571223554 Chr.2: 240589624-240589624 on Build GRCh38
rs762318743 Chr.2: 240589378-240589378 on Build GRCh38
rs142113962 Chr.2: 240589441-240589441 on Build GRCh38
rs61210947 Chr.2: 240589705-240589705 on Build GRCh38
rs762634427 Chr.2: 240589457-240589457 on Build GRCh38
rs748435887 Chr.2: 240589429-240589429 on Build GRCh38
rs939291631 Chr.2: 240589558-240589558 on Build GRCh38
rs767047251 Chr.2: 240589393-240589393 on Build GRCh38
rs527538258 Chr.2: 240589545-240589545 on Build GRCh38
rs977435106 Chr.2: 240589694-240589694 on Build GRCh38
rs560852120 Chr.2: 240589555-240589555 on Build GRCh38
rs957933760 Chr.2: 240589482-240589482 on Build GRCh38
rs899631968 Chr.2: 240589574-240589574 on Build GRCh38
rs55976923 Chr.2: 240589551-240589551 on Build GRCh38
rs113172458 Chr.2: 240589599-240589599 on Build GRCh38
rs941280000 Chr.2: 240589727-240589727 on Build GRCh38
rs745593937 Chr.2: 240589443-240589443 on Build GRCh38
rs536480834 Chr.2: 240589663-240589663 on Build GRCh38
rs969450839 Chr.2: 240589532-240589532 on Build GRCh38
rs778037172 Chr.2: 240589509-240589509 on Build GRCh38
rs774660762 Chr.2: 240589596-240589596 on Build GRCh38
rs1005767229 Chr.2: 240589418-240589418 on Build GRCh38
rs779259579 Chr.2: 240589315-240589315 on Build GRCh38
rs970430968 Chr.2: 240589300-240589300 on Build GRCh38
rs751154296 Chr.2: 240589383-240589383 on Build GRCh38
rs941206037 Chr.2: 240589571-240589571 on Build GRCh38
rs908462860 Chr.2: 240589718-240589718 on Build GRCh38
rs145259832 Chr.2: 240589444-240589444 on Build GRCh38
rs754246897 Chr.2: 240589394-240589394 on Build GRCh38
rs55893551 Chr.2: 240589510-240589510 on Build GRCh38
rs753055677 Chr.2: 240589479-240589479 on Build GRCh38
rs1031257543 Chr.2: 240589639-240589639 on Build GRCh38
rs924116181 Chr.2: 240589307-240589307 on Build GRCh38
rs764730986 Chr.2: 240589361-240589361 on Build GRCh38
rs771126731 Chr.2: 240589362-240589362 on Build GRCh38
rs776397701 Chr.2: 240589340-240589340 on Build GRCh38
rs745847820 Chr.2: 240589518-240589518 on Build GRCh38
rs201282500 Chr.2: 240589331-240589331 on Build GRCh38
rs56351903 Chr.2: 240589661-240589661 on Build GRCh38
rs948039711 Chr.2: 240589343-240589343 on Build GRCh38
rs1011889395 Chr.2: 240589486-240589486 on Build GRCh38
rs763690414 Chr.2: 240589382-240589382 on Build GRCh38
rs768242411 Chr.2: 240589462-240589462 on Build GRCh38
rs769885507 Chr.2: 240589309-240589309 on Build GRCh38
rs202048700 Chr.2: 240589323-240589323 on Build GRCh38
rs532861903 Chr.2: 240589660-240589660 on Build GRCh38
rs1047360120 Chr.2: 240589408-240589408 on Build GRCh38
rs142471367 Chr.2: 240589349-240589349 on Build GRCh38
rs762035846 Chr.2: 240589472-240589472 on Build GRCh38
rs908431005 Chr.2: 240589567-240589567 on Build GRCh38
rs534278931 Chr.2: 240589414-240589414 on Build GRCh38
rs780534696 Chr.2: 240589295-240589295 on Build GRCh38
rs369553350 Chr.2: 240589556-240589556 on Build GRCh38
rs758480249 Chr.2: 240589483-240589484 on Build GRCh38
rs766922732 Chr.2: 240589471-240589471 on Build GRCh38
rs750375555 Chr.2: 240589479-240589479 on Build GRCh38
rs70961717 Chr.2: 240589523-240589523 on Build GRCh38

View Full Product Details