Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240886436-240886935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
-
UniGene
Hs.193745
Gene Chromosome Location
Chr.2: 240886048-240896889 on Build GRCh38
Transcripts
NM_001085437.2 NM_001282921.1 NM_024861.3 XM_011511877.1

SNP Detail

SNP ID Chromosome Location
rs557280649 Chr.2: 240886481-240886481 on Build GRCh38
rs770580232 Chr.2: 240886807-240886807 on Build GRCh38
rs753515372 Chr.2: 240886834-240886834 on Build GRCh38
rs13397515 Chr.2: 240886633-240886633 on Build GRCh38
rs533599038 Chr.2: 240886789-240886789 on Build GRCh38
rs767184043 Chr.2: 240886551-240886551 on Build GRCh38
rs56008219 Chr.2: 240886736-240886736 on Build GRCh38
rs748877633 Chr.2: 240886783-240886783 on Build GRCh38
rs36014033 Chr.2: 240886523-240886523 on Build GRCh38
rs894605572 Chr.2: 240886561-240886561 on Build GRCh38
rs757584605 Chr.2: 240886513-240886513 on Build GRCh38
rs754811285 Chr.2: 240886847-240886847 on Build GRCh38
rs527711446 Chr.2: 240886598-240886598 on Build GRCh38
rs1036648439 Chr.2: 240886897-240886897 on Build GRCh38
rs764388882 Chr.2: 240886508-240886508 on Build GRCh38
rs977761507 Chr.2: 240886623-240886623 on Build GRCh38
rs550754514 Chr.2: 240886483-240886483 on Build GRCh38
rs578184007 Chr.2: 240886522-240886522 on Build GRCh38
rs551775763 Chr.2: 240886554-240886554 on Build GRCh38
rs1030638160 Chr.2: 240886876-240886876 on Build GRCh38
rs903440982 Chr.2: 240886826-240886826 on Build GRCh38
rs193117565 Chr.2: 240886704-240886704 on Build GRCh38
rs915982908 Chr.2: 240886831-240886831 on Build GRCh38
rs975777084 Chr.2: 240886665-240886665 on Build GRCh38
rs879194339 Chr.2: 240886721-240886722 on Build GRCh38
rs1022316944 Chr.2: 240886577-240886577 on Build GRCh38
rs33998315 Chr.2: 240886509-240886509 on Build GRCh38
rs373381052 Chr.2: 240886501-240886501 on Build GRCh38
rs1011732639 Chr.2: 240886571-240886571 on Build GRCh38
rs1056309967 Chr.2: 240886740-240886740 on Build GRCh38
rs921690196 Chr.2: 240886669-240886669 on Build GRCh38
rs965714413 Chr.2: 240886895-240886895 on Build GRCh38
rs907229435 Chr.2: 240886889-240886889 on Build GRCh38
rs566541368 Chr.2: 240886754-240886754 on Build GRCh38
rs555334337 Chr.2: 240886875-240886875 on Build GRCh38
rs928593998 Chr.2: 240886727-240886727 on Build GRCh38
rs953359492 Chr.2: 240886678-240886678 on Build GRCh38
rs940712752 Chr.2: 240886512-240886512 on Build GRCh38
rs1054430751 Chr.2: 240886499-240886499 on Build GRCh38
rs894582068 Chr.2: 240886504-240886504 on Build GRCh38
rs11680484 Chr.2: 240886721-240886721 on Build GRCh38
rs1049479459 Chr.2: 240886495-240886495 on Build GRCh38
rs560602451 Chr.2: 240886529-240886529 on Build GRCh38
rs948829708 Chr.2: 240886836-240886836 on Build GRCh38
rs965786115 Chr.2: 240886661-240886661 on Build GRCh38
rs994693378 Chr.2: 240886556-240886556 on Build GRCh38
rs189354778 Chr.2: 240886639-240886639 on Build GRCh38
rs13422330 Chr.2: 240886722-240886722 on Build GRCh38
rs752815080 Chr.2: 240886472-240886472 on Build GRCh38
rs80340507 Chr.2: 240886699-240886699 on Build GRCh38
rs567216521 Chr.2: 240886555-240886555 on Build GRCh38
rs573038017 Chr.2: 240886712-240886712 on Build GRCh38
rs529157941 Chr.2: 240886696-240886696 on Build GRCh38
rs1012196605 Chr.2: 240886815-240886815 on Build GRCh38
rs1043222180 Chr.2: 240886821-240886821 on Build GRCh38

View Full Product Details