Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 884994-885490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444271 c.679C>G p.L227V Chr.20: 885234-885234 on Build GRCh38
6599624 c.717C>T p.R239R Chr.20: 885196-885196 on Build GRCh38
7355426 c.728G>A p.G243D Chr.20: 885185-885185 on Build GRCh38
6599625 c.720C>A p.G240G Chr.20: 885193-885193 on Build GRCh38
444272 c.678G>A p.T226T Chr.20: 885235-885235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771933656 Chr.20: 885208-885208 on Build GRCh38
rs984445755 Chr.20: 885330-885330 on Build GRCh38
rs759073338 Chr.20: 885135-885135 on Build GRCh38
rs398088198 Chr.20: 885025-885025 on Build GRCh38
rs764893973 Chr.20: 885235-885235 on Build GRCh38
rs767753323 Chr.20: 885279-885279 on Build GRCh38
rs371525435 Chr.20: 885236-885236 on Build GRCh38
rs759237922 Chr.20: 885233-885233 on Build GRCh38
rs538984668 Chr.20: 885201-885201 on Build GRCh38
rs774314576 Chr.20: 885095-885095 on Build GRCh38
rs766002178 Chr.20: 885124-885124 on Build GRCh38
rs746887158 Chr.20: 885303-885303 on Build GRCh38
rs781438375 Chr.20: 885183-885183 on Build GRCh38
rs138750068 Chr.20: 885412-885412 on Build GRCh38
rs745582719 Chr.20: 885332-885332 on Build GRCh38
rs776383392 Chr.20: 885232-885232 on Build GRCh38
rs763482667 Chr.20: 885353-885353 on Build GRCh38
rs781131096 Chr.20: 885336-885336 on Build GRCh38
rs770519444 Chr.20: 885220-885220 on Build GRCh38
rs939710052 Chr.20: 885038-885038 on Build GRCh38
rs760252535 Chr.20: 885123-885123 on Build GRCh38
rs528391316 Chr.20: 885349-885349 on Build GRCh38
rs763557392 Chr.20: 885244-885244 on Build GRCh38
rs535869468 Chr.20: 885410-885410 on Build GRCh38
rs903202079 Chr.20: 885256-885256 on Build GRCh38
rs55955946 Chr.20: 885022-885022 on Build GRCh38
rs138501858 Chr.20: 885265-885265 on Build GRCh38
rs780095528 Chr.20: 885198-885198 on Build GRCh38
rs750717096 Chr.20: 885282-885282 on Build GRCh38
rs771917022 Chr.20: 885108-885108 on Build GRCh38
rs140682745 Chr.20: 885178-885178 on Build GRCh38
rs757118130 Chr.20: 885056-885056 on Build GRCh38
rs866845691 Chr.20: 885117-885117 on Build GRCh38
rs780116086 Chr.20: 885286-885286 on Build GRCh38
rs755455470 Chr.20: 885041-885041 on Build GRCh38
rs398081184 Chr.20: 885024-885024 on Build GRCh38
rs747253117 Chr.20: 885068-885068 on Build GRCh38
rs758960561 Chr.20: 885174-885174 on Build GRCh38
rs62641665 Chr.20: 885253-885253 on Build GRCh38
rs777722836 Chr.20: 885301-885301 on Build GRCh38
rs911075471 Chr.20: 885025-885026 on Build GRCh38
rs749311870 Chr.20: 885290-885290 on Build GRCh38
rs866552914 Chr.20: 885313-885313 on Build GRCh38
rs368820037 Chr.20: 885073-885073 on Build GRCh38
rs73892515 Chr.20: 885384-885384 on Build GRCh38
rs955505144 Chr.20: 885015-885015 on Build GRCh38
rs758356780 Chr.20: 885295-885295 on Build GRCh38
rs753325845 Chr.20: 885032-885032 on Build GRCh38
rs937778858 Chr.20: 885356-885356 on Build GRCh38
rs919891447 Chr.20: 885047-885047 on Build GRCh38
rs150174277 Chr.20: 885318-885318 on Build GRCh38
rs543557667 Chr.20: 885090-885090 on Build GRCh38
rs951582542 Chr.20: 885387-885387 on Build GRCh38
rs908177302 Chr.20: 885333-885333 on Build GRCh38
rs774398590 Chr.20: 885239-885239 on Build GRCh38
rs374785828 Chr.20: 885284-885284 on Build GRCh38
rs558718250 Chr.20: 885350-885350 on Build GRCh38
rs753302256 Chr.20: 885162-885162 on Build GRCh38
rs952622950 Chr.20: 885319-885319 on Build GRCh38
rs746047404 Chr.20: 885186-885186 on Build GRCh38
rs753140834 Chr.20: 885043-885043 on Build GRCh38
rs753308902 Chr.20: 885134-885134 on Build GRCh38
rs1047545770 Chr.20: 885067-885067 on Build GRCh38
rs200211515 Chr.20: 885026-885026 on Build GRCh38
rs894640199 Chr.20: 885390-885390 on Build GRCh38
rs773059233 Chr.20: 885121-885121 on Build GRCh38
rs564934817 Chr.20: 885354-885354 on Build GRCh38
rs748154330 Chr.20: 885203-885203 on Build GRCh38
rs143483356 Chr.20: 885091-885091 on Build GRCh38
rs757681930 Chr.20: 885278-885278 on Build GRCh38
rs566917313 Chr.20: 885386-885386 on Build GRCh38
rs1043717534 Chr.20: 885418-885418 on Build GRCh38
rs758869977 Chr.20: 885061-885061 on Build GRCh38
rs891025270 Chr.20: 885287-885287 on Build GRCh38
rs974089060 Chr.20: 885379-885379 on Build GRCh38
rs908210500 Chr.20: 885034-885034 on Build GRCh38
rs746772044 Chr.20: 885216-885216 on Build GRCh38
rs778257398 Chr.20: 885064-885064 on Build GRCh38
rs201829261 Chr.20: 885157-885157 on Build GRCh38
rs894721531 Chr.20: 885228-885228 on Build GRCh38
rs761716632 Chr.20: 885097-885097 on Build GRCh38
rs552328978 Chr.20: 885338-885338 on Build GRCh38
rs141812904 Chr.20: 885450-885450 on Build GRCh38
rs768613578 Chr.20: 885092-885092 on Build GRCh38
rs778347681 Chr.20: 885175-885175 on Build GRCh38
rs1028608928 Chr.20: 885291-885291 on Build GRCh38
rs1019642548 Chr.20: 885270-885270 on Build GRCh38
rs375064442 Chr.20: 885209-885209 on Build GRCh38
rs769911824 Chr.20: 885188-885188 on Build GRCh38
rs866232392 Chr.20: 885269-885269 on Build GRCh38
rs530585551 Chr.20: 885383-885383 on Build GRCh38
rs767992580 Chr.20: 885358-885358 on Build GRCh38
rs750125090 Chr.20: 885030-885030 on Build GRCh38

View Full Product Details