Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2795508-2796008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM1
Entrez Gene ID
56265
Gene Name
carboxypeptidase X (M14 family), member 1
Gene Aliases
CPX1, CPXM
UniGene
Hs.659346
Gene Chromosome Location
Chr.20: 2794069-2800646 on Build GRCh38
Transcripts
NM_001184699.1 NM_019609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6302833 c.1704G>A p.W568* Chr.20: 2795615-2795615 on Build GRCh38
5391751 c.1487A>G p.N496S Chr.20: 2795832-2795832 on Build GRCh38
5857568 c.1538C>T p.T513I Chr.20: 2795781-2795781 on Build GRCh38
5391750 c.1530G>A p.M510I Chr.20: 2795789-2795789 on Build GRCh38
390815 c.1633G>T p.D545Y Chr.20: 2795686-2795686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368108932 Chr.20: 2795979-2795979 on Build GRCh38
rs765326895 Chr.20: 2795570-2795570 on Build GRCh38
rs768695193 Chr.20: 2795726-2795726 on Build GRCh38
rs1027598497 Chr.20: 2795624-2795624 on Build GRCh38
rs767317980 Chr.20: 2795565-2795565 on Build GRCh38
rs371931784 Chr.20: 2795942-2795942 on Build GRCh38
rs147765286 Chr.20: 2795603-2795603 on Build GRCh38
rs1004343442 Chr.20: 2795780-2795780 on Build GRCh38
rs370337829 Chr.20: 2795858-2795858 on Build GRCh38
rs777608220 Chr.20: 2795709-2795709 on Build GRCh38
rs746438167 Chr.20: 2795565-2795565 on Build GRCh38
rs764676972 Chr.20: 2795982-2795982 on Build GRCh38
rs761705901 Chr.20: 2795556-2795556 on Build GRCh38
rs765850112 Chr.20: 2795813-2795813 on Build GRCh38
rs746791045 Chr.20: 2795716-2795716 on Build GRCh38
rs367560871 Chr.20: 2795905-2795905 on Build GRCh38
rs936559798 Chr.20: 2795698-2795698 on Build GRCh38
rs141526809 Chr.20: 2795677-2795677 on Build GRCh38
rs747229395 Chr.20: 2795882-2795882 on Build GRCh38
rs374663889 Chr.20: 2795609-2795609 on Build GRCh38
rs116267300 Chr.20: 2795756-2795756 on Build GRCh38
rs759523234 Chr.20: 2795723-2795723 on Build GRCh38
rs191472527 Chr.20: 2795626-2795626 on Build GRCh38
rs1033135068 Chr.20: 2795564-2795565 on Build GRCh38
rs774231916 Chr.20: 2795555-2795555 on Build GRCh38
rs760577440 Chr.20: 2795568-2795568 on Build GRCh38
rs1053862808 Chr.20: 2795689-2795689 on Build GRCh38
rs746182609 Chr.20: 2795974-2795974 on Build GRCh38
rs749150924 Chr.20: 2795954-2795954 on Build GRCh38
rs115394567 Chr.20: 2795564-2795564 on Build GRCh38
rs535070271 Chr.20: 2795798-2795798 on Build GRCh38
rs373765291 Chr.20: 2795890-2795890 on Build GRCh38
rs777022114 Chr.20: 2795970-2795970 on Build GRCh38
rs756605193 Chr.20: 2795627-2795627 on Build GRCh38
rs768482754 Chr.20: 2795551-2795551 on Build GRCh38
rs548761906 Chr.20: 2795932-2795932 on Build GRCh38
rs758603446 Chr.20: 2795589-2795589 on Build GRCh38
rs915331226 Chr.20: 2795920-2795920 on Build GRCh38
rs200029209 Chr.20: 2795648-2795648 on Build GRCh38
rs750813847 Chr.20: 2795684-2795684 on Build GRCh38
rs771686733 Chr.20: 2795703-2795703 on Build GRCh38
rs200677998 Chr.20: 2795938-2795938 on Build GRCh38
rs781010186 Chr.20: 2795774-2795774 on Build GRCh38
rs751145338 Chr.20: 2795832-2795832 on Build GRCh38
rs373345558 Chr.20: 2795791-2795791 on Build GRCh38
rs145475367 Chr.20: 2795729-2795729 on Build GRCh38
rs757667358 Chr.20: 2795606-2795606 on Build GRCh38
rs758008735 Chr.20: 2795764-2795764 on Build GRCh38
rs775442271 Chr.20: 2795713-2795714 on Build GRCh38
rs755980214 Chr.20: 2795862-2795862 on Build GRCh38
rs371160930 Chr.20: 2795916-2795916 on Build GRCh38
rs538584869 Chr.20: 2795647-2795647 on Build GRCh38
rs766944887 Chr.20: 2795743-2795743 on Build GRCh38
rs770028632 Chr.20: 2795977-2795977 on Build GRCh38
rs763022497 Chr.20: 2795672-2795672 on Build GRCh38
rs202103486 Chr.20: 2795757-2795757 on Build GRCh38
rs746357181 Chr.20: 2795612-2795612 on Build GRCh38
rs780955082 Chr.20: 2795856-2795856 on Build GRCh38
rs147866857 Chr.20: 2795642-2795642 on Build GRCh38
rs145736623 Chr.20: 2795602-2795602 on Build GRCh38
rs543432133 Chr.20: 2795767-2795767 on Build GRCh38
rs779889342 Chr.20: 2795867-2795867 on Build GRCh38
rs764257542 Chr.20: 2795673-2795673 on Build GRCh38
rs368549089 Chr.20: 2795852-2795852 on Build GRCh38
rs761805318 Chr.20: 2795783-2795783 on Build GRCh38
rs759980275 Chr.20: 2795910-2795910 on Build GRCh38
rs752205258 Chr.20: 2795762-2795762 on Build GRCh38
rs905042088 Chr.20: 2795534-2795534 on Build GRCh38
rs370747954 Chr.20: 2795824-2795824 on Build GRCh38
rs755008775 Chr.20: 2795957-2795957 on Build GRCh38
rs775487980 Chr.20: 2795662-2795662 on Build GRCh38
rs376564008 Chr.20: 2795831-2795831 on Build GRCh38
rs149777086 Chr.20: 2795641-2795641 on Build GRCh38
rs756975018 Chr.20: 2795851-2795851 on Build GRCh38
rs778615093 Chr.20: 2795692-2795692 on Build GRCh38
rs769783313 Chr.20: 2795778-2795778 on Build GRCh38
rs549143936 Chr.20: 2795530-2795530 on Build GRCh38
rs372249479 Chr.20: 2795730-2795730 on Build GRCh38
rs751132169 Chr.20: 2795933-2795933 on Build GRCh38
rs748821645 Chr.20: 2795628-2795628 on Build GRCh38
rs754320160 Chr.20: 2795803-2795803 on Build GRCh38
rs763184113 Chr.20: 2795734-2795735 on Build GRCh38
rs766818424 Chr.20: 2795795-2795795 on Build GRCh38
rs1001086455 Chr.20: 2795540-2795540 on Build GRCh38
rs376373857 Chr.20: 2795965-2795965 on Build GRCh38
rs763360005 Chr.20: 2795981-2795981 on Build GRCh38
rs41309351 Chr.20: 2795881-2795881 on Build GRCh38
rs1019250052 Chr.20: 2795586-2795586 on Build GRCh38
rs201609174 Chr.20: 2795633-2795633 on Build GRCh38
rs756588229 Chr.20: 2795687-2795687 on Build GRCh38
rs748210749 Chr.20: 2795880-2795880 on Build GRCh38
rs199961287 Chr.20: 2795941-2795941 on Build GRCh38
rs369924880 Chr.20: 2795812-2795812 on Build GRCh38
rs776576915 Chr.20: 2795721-2795721 on Build GRCh38
rs559989068 Chr.20: 2795857-2795857 on Build GRCh38
rs751762428 Chr.20: 2795674-2795674 on Build GRCh38
rs771334283 Chr.20: 2795904-2795904 on Build GRCh38
rs141160474 Chr.20: 2795765-2795765 on Build GRCh38
rs767165149 Chr.20: 2795940-2795940 on Build GRCh38
rs773697213 Chr.20: 2795572-2795572 on Build GRCh38
rs6115674 Chr.20: 2795590-2795590 on Build GRCh38
rs778560289 Chr.20: 2795758-2795758 on Build GRCh38
rs753359247 Chr.20: 2795691-2795691 on Build GRCh38
rs769992463 Chr.20: 2795678-2795680 on Build GRCh38
rs763559979 Chr.20: 2795930-2795930 on Build GRCh38
rs369243896 Chr.20: 2795777-2795777 on Build GRCh38
rs374518251 Chr.20: 2795919-2795919 on Build GRCh38
rs770784652 Chr.20: 2795720-2795720 on Build GRCh38
rs746775795 Chr.20: 2795766-2795766 on Build GRCh38
rs781760114 Chr.20: 2795555-2795556 on Build GRCh38
rs1011528614 Chr.20: 2795529-2795529 on Build GRCh38
rs549828285 Chr.20: 2795948-2795948 on Build GRCh38
rs147693328 Chr.20: 2795731-2795731 on Build GRCh38
rs780596511 Chr.20: 2795924-2795924 on Build GRCh38
rs752838139 Chr.20: 2795575-2795575 on Build GRCh38
rs767923118 Chr.20: 2795681-2795681 on Build GRCh38
rs201409887 Chr.20: 2795688-2795688 on Build GRCh38
rs747803655 Chr.20: 2795699-2795699 on Build GRCh38
rs943982703 Chr.20: 2795952-2795952 on Build GRCh38
rs781462252 Chr.20: 2795610-2795610 on Build GRCh38
rs577150634 Chr.20: 2795785-2795785 on Build GRCh38
rs373422048 Chr.20: 2795825-2795825 on Build GRCh38
rs565163190 Chr.20: 2795784-2795784 on Build GRCh38
rs571190562 Chr.20: 2795964-2795964 on Build GRCh38
rs1016393952 Chr.20: 2795830-2795830 on Build GRCh38
rs768357907 Chr.20: 2795630-2795630 on Build GRCh38
rs376401302 Chr.20: 2795794-2795794 on Build GRCh38
rs774433970 Chr.20: 2795781-2795781 on Build GRCh38

View Full Product Details