Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2796408-2796901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM1
Entrez Gene ID
56265
Gene Name
carboxypeptidase X (M14 family), member 1
Gene Aliases
CPX1, CPXM
UniGene
Hs.659346
Gene Chromosome Location
Chr.20: 2794069-2800646 on Build GRCh38
Transcripts
NM_001184699.1 NM_019609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846564 c.940G>C p.E314Q Chr.20: 2796632-2796632 on Build GRCh38
4290592 c.1020G>A p.S340S Chr.20: 2796552-2796552 on Build GRCh38
1025469 c.948C>T p.C316C Chr.20: 2796624-2796624 on Build GRCh38
5366053 c.1058T>C p.V353A Chr.20: 2796431-2796431 on Build GRCh38
6266061 c.959C>A p.T320N Chr.20: 2796613-2796613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779698196 Chr.20: 2796634-2796634 on Build GRCh38
rs751923365 Chr.20: 2796472-2796472 on Build GRCh38
rs766563846 Chr.20: 2796637-2796637 on Build GRCh38
rs776705207 Chr.20: 2796453-2796453 on Build GRCh38
rs764512354 Chr.20: 2796469-2796469 on Build GRCh38
rs774081235 Chr.20: 2796436-2796436 on Build GRCh38
rs765395063 Chr.20: 2796459-2796459 on Build GRCh38
rs749908669 Chr.20: 2796511-2796511 on Build GRCh38
rs767978402 Chr.20: 2796624-2796624 on Build GRCh38
rs764232811 Chr.20: 2796613-2796613 on Build GRCh38
rs921071351 Chr.20: 2796742-2796742 on Build GRCh38
rs754455123 Chr.20: 2796789-2796789 on Build GRCh38
rs778713002 Chr.20: 2796543-2796543 on Build GRCh38
rs976628333 Chr.20: 2796741-2796741 on Build GRCh38
rs1010621442 Chr.20: 2796810-2796810 on Build GRCh38
rs559802149 Chr.20: 2796670-2796670 on Build GRCh38
rs1002382493 Chr.20: 2796480-2796480 on Build GRCh38
rs774418507 Chr.20: 2796683-2796683 on Build GRCh38
rs1046609512 Chr.20: 2796757-2796757 on Build GRCh38
rs750933007 Chr.20: 2796628-2796628 on Build GRCh38
rs771741874 Chr.20: 2796647-2796647 on Build GRCh38
rs773155636 Chr.20: 2796610-2796610 on Build GRCh38
rs372756171 Chr.20: 2796800-2796800 on Build GRCh38
rs6051399 Chr.20: 2796673-2796673 on Build GRCh38
rs201086152 Chr.20: 2796429-2796429 on Build GRCh38
rs1985525 Chr.20: 2796813-2796813 on Build GRCh38
rs372839676 Chr.20: 2796455-2796455 on Build GRCh38
rs932477514 Chr.20: 2796650-2796650 on Build GRCh38
rs754594614 Chr.20: 2796638-2796638 on Build GRCh38
rs377013053 Chr.20: 2796652-2796652 on Build GRCh38
rs762139486 Chr.20: 2796688-2796688 on Build GRCh38
rs867520539 Chr.20: 2796540-2796540 on Build GRCh38
rs188787150 Chr.20: 2796816-2796816 on Build GRCh38
rs906674609 Chr.20: 2796474-2796474 on Build GRCh38
rs61731570 Chr.20: 2796432-2796432 on Build GRCh38
rs762109961 Chr.20: 2796623-2796623 on Build GRCh38
rs1033417191 Chr.20: 2796490-2796490 on Build GRCh38
rs752893225 Chr.20: 2796466-2796466 on Build GRCh38
rs576568240 Chr.20: 2796827-2796827 on Build GRCh38
rs747898613 Chr.20: 2796552-2796552 on Build GRCh38
rs772003992 Chr.20: 2796598-2796598 on Build GRCh38
rs753069583 Chr.20: 2796840-2796840 on Build GRCh38
rs763146002 Chr.20: 2796612-2796612 on Build GRCh38
rs768252824 Chr.20: 2796538-2796538 on Build GRCh38
rs371403963 Chr.20: 2796669-2796669 on Build GRCh38
rs773288274 Chr.20: 2796530-2796530 on Build GRCh38
rs767769814 Chr.20: 2796689-2796689 on Build GRCh38
rs542464997 Chr.20: 2796697-2796697 on Build GRCh38
rs895418144 Chr.20: 2796483-2796483 on Build GRCh38
rs983988906 Chr.20: 2796773-2796773 on Build GRCh38
rs201255371 Chr.20: 2796521-2796521 on Build GRCh38
rs1025108999 Chr.20: 2796470-2796470 on Build GRCh38
rs760853354 Chr.20: 2796553-2796553 on Build GRCh38
rs888094293 Chr.20: 2796427-2796427 on Build GRCh38
rs185072826 Chr.20: 2796788-2796788 on Build GRCh38
rs768875938 Chr.20: 2796676-2796676 on Build GRCh38
rs375803414 Chr.20: 2796501-2796501 on Build GRCh38
rs771917825 Chr.20: 2796445-2796445 on Build GRCh38
rs954008259 Chr.20: 2796761-2796761 on Build GRCh38
rs138560626 Chr.20: 2796611-2796611 on Build GRCh38
rs778460841 Chr.20: 2796639-2796639 on Build GRCh38
rs751915961 Chr.20: 2796616-2796616 on Build GRCh38
rs376062269 Chr.20: 2796680-2796680 on Build GRCh38
rs200844270 Chr.20: 2796684-2796684 on Build GRCh38
rs370103504 Chr.20: 2796515-2796515 on Build GRCh38
rs756880482 Chr.20: 2796851-2796851 on Build GRCh38
rs765702872 Chr.20: 2796820-2796820 on Build GRCh38
rs761975322 Chr.20: 2796467-2796467 on Build GRCh38
rs1019043669 Chr.20: 2796618-2796618 on Build GRCh38
rs201677011 Chr.20: 2796442-2796442 on Build GRCh38
rs747926278 Chr.20: 2796641-2796641 on Build GRCh38
rs748944255 Chr.20: 2796537-2796537 on Build GRCh38
rs757718654 Chr.20: 2796489-2796489 on Build GRCh38

View Full Product Details