Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3082556-3083078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVP
Entrez Gene ID
551
Gene Name
arginine vasopressin
Gene Aliases
ADH, ARVP, AVP-NPII, AVRP, VP
UniGene
Hs.89648
Gene Chromosome Location
Chr.20: 3082555-3093521 on Build GRCh38
Transcripts
NM_000490.4 XM_011529267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4808064 c.237C>T p.N79N Chr.20: 3083062-3083062 on Build GRCh38
3693426 c.230A>T p.E77V Chr.20: 3083069-3083069 on Build GRCh38
222396 c.440T>G p.V147G Chr.20: 3082685-3082685 on Build GRCh38
6525588 c.475G>T p.A159S Chr.20: 3082650-3082650 on Build GRCh38
4642425 c.221G>A p.R74H Chr.20: 3083078-3083078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932498498 Chr.20: 3082943-3082944 on Build GRCh38
rs1015300508 Chr.20: 3082790-3082790 on Build GRCh38
rs948121208 Chr.20: 3082865-3082865 on Build GRCh38
rs748643601 Chr.20: 3082974-3082974 on Build GRCh38
rs534290018 Chr.20: 3082751-3082751 on Build GRCh38
rs1051747 Chr.20: 3082621-3082621 on Build GRCh38
rs1056437967 Chr.20: 3082619-3082619 on Build GRCh38
rs756246762 Chr.20: 3082926-3082926 on Build GRCh38
rs762655956 Chr.20: 3082945-3082946 on Build GRCh38
rs121964882 Chr.20: 3083037-3083037 on Build GRCh38
rs1041251942 Chr.20: 3082709-3082709 on Build GRCh38
rs760345436 Chr.20: 3082650-3082650 on Build GRCh38
rs918195268 Chr.20: 3082620-3082620 on Build GRCh38
rs5195 Chr.20: 3083054-3083054 on Build GRCh38
rs896887866 Chr.20: 3082793-3082793 on Build GRCh38
rs971361345 Chr.20: 3082632-3082632 on Build GRCh38
rs780084300 Chr.20: 3082946-3082946 on Build GRCh38
rs763977037 Chr.20: 3083030-3083030 on Build GRCh38
rs121964891 Chr.20: 3083024-3083024 on Build GRCh38
rs1009997849 Chr.20: 3082722-3082722 on Build GRCh38
rs911455353 Chr.20: 3082590-3082590 on Build GRCh38
rs925274371 Chr.20: 3082947-3082947 on Build GRCh38
rs121964885 Chr.20: 3083022-3083022 on Build GRCh38
rs941539399 Chr.20: 3082592-3082592 on Build GRCh38
rs772747008 Chr.20: 3082978-3082978 on Build GRCh38
rs549545938 Chr.20: 3082760-3082760 on Build GRCh38
rs121964889 Chr.20: 3082788-3082788 on Build GRCh38
rs951507234 Chr.20: 3082858-3082858 on Build GRCh38
rs917977387 Chr.20: 3082864-3082864 on Build GRCh38
rs1051746 Chr.20: 3082623-3082623 on Build GRCh38
rs948461947 Chr.20: 3082915-3082938 on Build GRCh38
rs753996905 Chr.20: 3083050-3083050 on Build GRCh38
rs765910191 Chr.20: 3082665-3082665 on Build GRCh38
rs917410096 Chr.20: 3082602-3082618 on Build GRCh38
rs926643351 Chr.20: 3082680-3082680 on Build GRCh38
rs765019311 Chr.20: 3082716-3082716 on Build GRCh38
rs986119822 Chr.20: 3082589-3082589 on Build GRCh38
rs568253217 Chr.20: 3082764-3082764 on Build GRCh38
rs1000504167 Chr.20: 3082707-3082707 on Build GRCh38
rs932588483 Chr.20: 3082757-3082757 on Build GRCh38
rs565894445 Chr.20: 3082611-3082611 on Build GRCh38
rs914512433 Chr.20: 3082873-3082873 on Build GRCh38
rs1044716891 Chr.20: 3082692-3082692 on Build GRCh38
rs746457360 Chr.20: 3082995-3082995 on Build GRCh38
rs775308351 Chr.20: 3083021-3083021 on Build GRCh38
rs765002935 Chr.20: 3083017-3083017 on Build GRCh38
rs74315383 Chr.20: 3082779-3082779 on Build GRCh38
rs974234418 Chr.20: 3082599-3082599 on Build GRCh38
rs1033920744 Chr.20: 3082857-3082857 on Build GRCh38
rs769735284 Chr.20: 3082925-3082925 on Build GRCh38
rs759192862 Chr.20: 3082704-3082704 on Build GRCh38
rs121964884 Chr.20: 3083005-3083005 on Build GRCh38
rs751535373 Chr.20: 3083034-3083034 on Build GRCh38
rs1049648051 Chr.20: 3082758-3082758 on Build GRCh38
rs970904958 Chr.20: 3082832-3082832 on Build GRCh38
rs1005281728 Chr.20: 3082609-3082609 on Build GRCh38
rs121964890 Chr.20: 3083039-3083039 on Build GRCh38
rs535682482 Chr.20: 3082980-3082980 on Build GRCh38
rs758387902 Chr.20: 3082762-3082762 on Build GRCh38
rs888325542 Chr.20: 3082946-3082947 on Build GRCh38
rs13041122 Chr.20: 3082588-3082588 on Build GRCh38
rs1054332322 Chr.20: 3082708-3082708 on Build GRCh38
rs906725769 Chr.20: 3083033-3083033 on Build GRCh38
rs13041150 Chr.20: 3082642-3082642 on Build GRCh38
rs930095239 Chr.20: 3082612-3082612 on Build GRCh38
rs762842372 Chr.20: 3083026-3083026 on Build GRCh38
rs1028376714 Chr.20: 3082594-3082594 on Build GRCh38
rs757686786 Chr.20: 3082949-3082950 on Build GRCh38
rs936650522 Chr.20: 3082691-3082691 on Build GRCh38
rs751309418 Chr.20: 3082948-3082949 on Build GRCh38
rs867198716 Chr.20: 3082775-3082775 on Build GRCh38
rs13041148 Chr.20: 3082636-3082636 on Build GRCh38
rs766551044 Chr.20: 3083047-3083047 on Build GRCh38
rs566930108 Chr.20: 3082703-3082703 on Build GRCh38
rs963031797 Chr.20: 3082815-3082815 on Build GRCh38
rs121964886 Chr.20: 3083012-3083012 on Build GRCh38
rs759299430 Chr.20: 3083014-3083014 on Build GRCh38
rs1051744 Chr.20: 3082769-3082769 on Build GRCh38
rs974288244 Chr.20: 3082827-3082827 on Build GRCh38
rs767559658 Chr.20: 3082785-3082785 on Build GRCh38
rs756084110 Chr.20: 3083035-3083035 on Build GRCh38
rs776428607 Chr.20: 3082742-3082743 on Build GRCh38
rs1018338454 Chr.20: 3082794-3082794 on Build GRCh38
rs775089446 Chr.20: 3082942-3082943 on Build GRCh38
rs991251917 Chr.20: 3082869-3082869 on Build GRCh38
rs57591338 Chr.20: 3082610-3082610 on Build GRCh38
rs776370930 Chr.20: 3082685-3082685 on Build GRCh38
rs992218608 Chr.20: 3082859-3082859 on Build GRCh38
rs755297710 Chr.20: 3082962-3082962 on Build GRCh38
rs1045097290 Chr.20: 3082874-3082874 on Build GRCh38
rs1051748 Chr.20: 3082617-3082617 on Build GRCh38
rs779482841 Chr.20: 3082973-3082973 on Build GRCh38
rs958558398 Chr.20: 3082855-3082855 on Build GRCh38
rs13041277 Chr.20: 3082660-3082660 on Build GRCh38
rs968999163 Chr.20: 3082899-3082899 on Build GRCh38
rs891637899 Chr.20: 3083016-3083016 on Build GRCh38
rs200820983 Chr.20: 3082941-3082942 on Build GRCh38
rs918762576 Chr.20: 3082838-3082838 on Build GRCh38
rs890908130 Chr.20: 3083028-3083028 on Build GRCh38
rs763810447 Chr.20: 3082947-3082948 on Build GRCh38
rs981472398 Chr.20: 3082843-3082843 on Build GRCh38

View Full Product Details