Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3259915-3260414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf194
Entrez Gene ID
25943
Gene Name
chromosome 20 open reading frame 194
Gene Aliases
-
UniGene
Hs.516853
Gene Chromosome Location
Chr.20: 3249302-3407662 on Build GRCh38
Transcripts
NM_001009984.2 XM_005260684.3 XM_005260687.4 XM_011529208.2 XM_017027783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495025 c.2884A>G p.K962E Chr.20: 3260018-3260018 on Build GRCh38
1731477 c.2946delC p.L983fs*33 Chr.20: 3259956-3259956 on Build GRCh38
3545512 c.2880A>G p.E960E Chr.20: 3260022-3260022 on Build GRCh38
5436109 c.2965G>A p.V989M Chr.20: 3259937-3259937 on Build GRCh38
7102004 c.2900C>T p.S967L Chr.20: 3260002-3260002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767876865 Chr.20: 3260039-3260039 on Build GRCh38
rs376312499 Chr.20: 3260259-3260259 on Build GRCh38
rs6051686 Chr.20: 3260207-3260207 on Build GRCh38
rs1033478341 Chr.20: 3260111-3260111 on Build GRCh38
rs757685587 Chr.20: 3260026-3260026 on Build GRCh38
rs182286470 Chr.20: 3259973-3259973 on Build GRCh38
rs751818329 Chr.20: 3260009-3260009 on Build GRCh38
rs547015265 Chr.20: 3260062-3260062 on Build GRCh38
rs989818945 Chr.20: 3260147-3260147 on Build GRCh38
rs369010167 Chr.20: 3259947-3259947 on Build GRCh38
rs947357596 Chr.20: 3260155-3260155 on Build GRCh38
rs866546003 Chr.20: 3260181-3260181 on Build GRCh38
rs62208077 Chr.20: 3260312-3260312 on Build GRCh38
rs771882764 Chr.20: 3259961-3259961 on Build GRCh38
rs932890163 Chr.20: 3260196-3260196 on Build GRCh38
rs6115832 Chr.20: 3260164-3260164 on Build GRCh38
rs1003829279 Chr.20: 3260232-3260232 on Build GRCh38
rs376988253 Chr.20: 3260079-3260079 on Build GRCh38
rs6084317 Chr.20: 3260078-3260078 on Build GRCh38
rs906256429 Chr.20: 3260248-3260248 on Build GRCh38
rs1048467463 Chr.20: 3260346-3260346 on Build GRCh38
rs773976624 Chr.20: 3260096-3260096 on Build GRCh38
rs6051687 Chr.20: 3260286-3260286 on Build GRCh38
rs915193209 Chr.20: 3260154-3260154 on Build GRCh38
rs1031307695 Chr.20: 3260288-3260288 on Build GRCh38
rs1049108088 Chr.20: 3260201-3260201 on Build GRCh38
rs773574489 Chr.20: 3260345-3260346 on Build GRCh38
rs895622409 Chr.20: 3260231-3260231 on Build GRCh38
rs375229902 Chr.20: 3260113-3260113 on Build GRCh38
rs527336883 Chr.20: 3260279-3260279 on Build GRCh38
rs773243278 Chr.20: 3259964-3259964 on Build GRCh38
rs748811298 Chr.20: 3260055-3260055 on Build GRCh38
rs1057425457 Chr.20: 3260209-3260209 on Build GRCh38
rs778676304 Chr.20: 3260061-3260061 on Build GRCh38
rs754629105 Chr.20: 3260059-3260059 on Build GRCh38
rs978405040 Chr.20: 3259943-3259943 on Build GRCh38
rs940283428 Chr.20: 3260316-3260316 on Build GRCh38
rs759321443 Chr.20: 3260106-3260106 on Build GRCh38
rs1022136609 Chr.20: 3260388-3260388 on Build GRCh38
rs901807470 Chr.20: 3260126-3260126 on Build GRCh38
rs76776062 Chr.20: 3260369-3260369 on Build GRCh38
rs372426469 Chr.20: 3259960-3259960 on Build GRCh38
rs558772984 Chr.20: 3260124-3260124 on Build GRCh38
rs572308676 Chr.20: 3260175-3260175 on Build GRCh38
rs538688796 Chr.20: 3260068-3260068 on Build GRCh38
rs900049860 Chr.20: 3260066-3260066 on Build GRCh38
rs989355127 Chr.20: 3260264-3260264 on Build GRCh38
rs1018726109 Chr.20: 3260117-3260117 on Build GRCh38
rs1011149315 Chr.20: 3260359-3260359 on Build GRCh38
rs1041813666 Chr.20: 3260008-3260008 on Build GRCh38
rs771971232 Chr.20: 3260065-3260065 on Build GRCh38
rs567509039 Chr.20: 3260308-3260308 on Build GRCh38
rs561819153 Chr.20: 3260107-3260107 on Build GRCh38
rs763098856 Chr.20: 3259997-3259997 on Build GRCh38
rs755670492 Chr.20: 3260045-3260045 on Build GRCh38
rs888687006 Chr.20: 3260315-3260315 on Build GRCh38
rs984577510 Chr.20: 3260357-3260357 on Build GRCh38
rs6139052 Chr.20: 3260324-3260324 on Build GRCh38
rs6051685 Chr.20: 3260077-3260077 on Build GRCh38
rs539337015 Chr.20: 3260356-3260356 on Build GRCh38
rs113661467 Chr.20: 3260256-3260256 on Build GRCh38
rs556622629 Chr.20: 3259942-3259942 on Build GRCh38
rs912690115 Chr.20: 3260011-3260011 on Build GRCh38
rs966651505 Chr.20: 3260271-3260271 on Build GRCh38
rs759609254 Chr.20: 3259972-3259972 on Build GRCh38
rs539171583 Chr.20: 3260080-3260080 on Build GRCh38
rs944078904 Chr.20: 3260287-3260287 on Build GRCh38
rs760623654 Chr.20: 3260097-3260097 on Build GRCh38
rs1015277395 Chr.20: 3260105-3260105 on Build GRCh38
rs1003266187 Chr.20: 3260249-3260249 on Build GRCh38
rs1009991402 Chr.20: 3260114-3260114 on Build GRCh38
rs543153576 Chr.20: 3260091-3260091 on Build GRCh38
rs190357840 Chr.20: 3260166-3260166 on Build GRCh38
rs28440122 Chr.20: 3259975-3259975 on Build GRCh38
rs573930463 Chr.20: 3260128-3260128 on Build GRCh38
rs773149876 Chr.20: 3260011-3260011 on Build GRCh38
rs1057162370 Chr.20: 3260383-3260387 on Build GRCh38
rs751033843 Chr.20: 3260042-3260042 on Build GRCh38
rs1004048288 Chr.20: 3260204-3260204 on Build GRCh38
rs894934264 Chr.20: 3260358-3260358 on Build GRCh38
rs955693448 Chr.20: 3260295-3260295 on Build GRCh38
rs779651147 Chr.20: 3260047-3260047 on Build GRCh38
rs369960186 Chr.20: 3259979-3259979 on Build GRCh38
rs182598026 Chr.20: 3260309-3260309 on Build GRCh38
rs906223679 Chr.20: 3260329-3260329 on Build GRCh38
rs186904559 Chr.20: 3259998-3259998 on Build GRCh38
rs900016190 Chr.20: 3260195-3260195 on Build GRCh38
rs1022631647 Chr.20: 3260270-3260270 on Build GRCh38
rs1015244972 Chr.20: 3260246-3260246 on Build GRCh38
rs772637794 Chr.20: 3260143-3260143 on Build GRCh38
rs868857645 Chr.20: 3260314-3260314 on Build GRCh38

View Full Product Details