Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3659771-3660267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA4
Entrez Gene ID
64096
Gene Name
GDNF family receptor alpha 4
Gene Aliases
-
UniGene
Hs.302025
Gene Chromosome Location
Chr.20: 3659292-3663399 on Build GRCh38
Transcripts
NM_022139.3 NM_145762.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723805 c.802G>T p.E268* Chr.20: 3660175-3660175 on Build GRCh38
1726226 c.850C>T p.R284C Chr.20: 3659959-3659959 on Build GRCh38
5724366 c.785A>G p.N262S Chr.20: 3660192-3660192 on Build GRCh38
6373778 c.763C>T p.P255S Chr.20: 3660214-3660214 on Build GRCh38
3405055 c.744C>A p.A248A Chr.20: 3660233-3660233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919030306 Chr.20: 3660232-3660232 on Build GRCh38
rs753006911 Chr.20: 3660021-3660021 on Build GRCh38
rs375585615 Chr.20: 3660036-3660036 on Build GRCh38
rs545460535 Chr.20: 3660195-3660195 on Build GRCh38
rs765456420 Chr.20: 3659930-3659930 on Build GRCh38
rs111644613 Chr.20: 3660112-3660112 on Build GRCh38
rs755123531 Chr.20: 3659925-3659925 on Build GRCh38
rs553182220 Chr.20: 3660121-3660121 on Build GRCh38
rs73893068 Chr.20: 3659833-3659833 on Build GRCh38
rs142300828 Chr.20: 3660218-3660218 on Build GRCh38
rs371337814 Chr.20: 3660156-3660156 on Build GRCh38
rs576933084 Chr.20: 3660096-3660096 on Build GRCh38
rs559099783 Chr.20: 3659801-3659801 on Build GRCh38
rs746166317 Chr.20: 3659937-3659937 on Build GRCh38
rs998061212 Chr.20: 3660208-3660208 on Build GRCh38
rs150162133 Chr.20: 3660220-3660220 on Build GRCh38
rs893889707 Chr.20: 3659845-3659845 on Build GRCh38
rs1036615847 Chr.20: 3659794-3659794 on Build GRCh38
rs548029145 Chr.20: 3659827-3659827 on Build GRCh38
rs776630288 Chr.20: 3660001-3660001 on Build GRCh38
rs201707979 Chr.20: 3659909-3659909 on Build GRCh38
rs1028540707 Chr.20: 3660110-3660110 on Build GRCh38
rs759631725 Chr.20: 3660003-3660003 on Build GRCh38
rs374283828 Chr.20: 3660222-3660222 on Build GRCh38
rs372535900 Chr.20: 3659807-3659807 on Build GRCh38
rs758825232 Chr.20: 3659937-3659937 on Build GRCh38
rs1042506706 Chr.20: 3660081-3660081 on Build GRCh38
rs775360551 Chr.20: 3659904-3659904 on Build GRCh38
rs915763108 Chr.20: 3660074-3660074 on Build GRCh38
rs759582850 Chr.20: 3660226-3660226 on Build GRCh38
rs373372326 Chr.20: 3659959-3659959 on Build GRCh38
rs1010047838 Chr.20: 3660073-3660073 on Build GRCh38
rs773161894 Chr.20: 3660224-3660224 on Build GRCh38
rs756510486 Chr.20: 3660108-3660109 on Build GRCh38
rs73611721 Chr.20: 3659872-3659872 on Build GRCh38
rs1036277934 Chr.20: 3659809-3659809 on Build GRCh38
rs764226732 Chr.20: 3660243-3660243 on Build GRCh38
rs765401400 Chr.20: 3660013-3660013 on Build GRCh38
rs780916766 Chr.20: 3659969-3659969 on Build GRCh38
rs74740423 Chr.20: 3659921-3659921 on Build GRCh38
rs1046272890 Chr.20: 3660014-3660014 on Build GRCh38
rs867124936 Chr.20: 3659926-3659926 on Build GRCh38
rs377368591 Chr.20: 3659972-3659972 on Build GRCh38
rs745574549 Chr.20: 3659898-3659898 on Build GRCh38
rs953167413 Chr.20: 3659928-3659928 on Build GRCh38
rs919068307 Chr.20: 3659812-3659812 on Build GRCh38
rs767443200 Chr.20: 3660000-3660000 on Build GRCh38
rs763306194 Chr.20: 3660240-3660240 on Build GRCh38
rs2853208 Chr.20: 3660176-3660176 on Build GRCh38
rs679393 Chr.20: 3659852-3659852 on Build GRCh38
rs1031278963 Chr.20: 3659893-3659893 on Build GRCh38
rs926542542 Chr.20: 3660028-3660028 on Build GRCh38
rs769709083 Chr.20: 3659899-3659899 on Build GRCh38
rs563073540 Chr.20: 3660166-3660166 on Build GRCh38
rs78068372 Chr.20: 3659994-3659994 on Build GRCh38
rs919068467 Chr.20: 3659985-3659985 on Build GRCh38
rs755723552 Chr.20: 3660181-3660181 on Build GRCh38
rs752767048 Chr.20: 3660182-3660183 on Build GRCh38
rs769514460 Chr.20: 3659979-3659979 on Build GRCh38
rs774142324 Chr.20: 3659997-3659997 on Build GRCh38
rs769968310 Chr.20: 3660233-3660233 on Build GRCh38
rs370637798 Chr.20: 3659891-3659891 on Build GRCh38
rs558807968 Chr.20: 3660064-3660064 on Build GRCh38
rs774384904 Chr.20: 3659919-3659919 on Build GRCh38
rs1046324862 Chr.20: 3659814-3659814 on Build GRCh38
rs544069656 Chr.20: 3659923-3659923 on Build GRCh38
rs998373249 Chr.20: 3660106-3660106 on Build GRCh38
rs948115902 Chr.20: 3660077-3660077 on Build GRCh38
rs115995912 Chr.20: 3660098-3660098 on Build GRCh38
rs141342254 Chr.20: 3659965-3659965 on Build GRCh38
rs115789290 Chr.20: 3659922-3659922 on Build GRCh38
rs775564801 Chr.20: 3660234-3660234 on Build GRCh38
rs73893069 Chr.20: 3660126-3660126 on Build GRCh38
rs776404687 Chr.20: 3659892-3659892 on Build GRCh38
rs1045079777 Chr.20: 3659856-3659856 on Build GRCh38
rs752953333 Chr.20: 3659934-3659934 on Build GRCh38
rs768332292 Chr.20: 3660189-3660189 on Build GRCh38
rs778363482 Chr.20: 3659870-3659870 on Build GRCh38
rs763896844 Chr.20: 3659912-3659912 on Build GRCh38
rs635674 Chr.20: 3659820-3659820 on Build GRCh38
rs544481488 Chr.20: 3660185-3660185 on Build GRCh38
rs762822745 Chr.20: 3659907-3659907 on Build GRCh38
rs535023669 Chr.20: 3659890-3659890 on Build GRCh38
rs574392801 Chr.20: 3660123-3660123 on Build GRCh38
rs766802635 Chr.20: 3659996-3659996 on Build GRCh38
rs1054188235 Chr.20: 3660045-3660045 on Build GRCh38
rs373027287 Chr.20: 3659868-3659868 on Build GRCh38
rs145198176 Chr.20: 3659984-3659984 on Build GRCh38
rs1021536912 Chr.20: 3659853-3659853 on Build GRCh38
rs1056923550 Chr.20: 3659828-3659828 on Build GRCh38
rs963040290 Chr.20: 3659991-3659991 on Build GRCh38
rs749026491 Chr.20: 3660187-3660187 on Build GRCh38

View Full Product Details