Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65389245-65389744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC6
Entrez Gene ID
9829
Gene Name
DnaJ heat shock protein family (Hsp40) member C6
Gene Aliases
DJC6, PARK19
UniGene
Hs.647643
Gene Chromosome Location
Chr.1: 65264694-65415869 on Build GRCh38
Transcripts
NM_001256864.1 NM_001256865.1 NM_014787.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
681476 c.1258T>C p.Y420H Chr.1: 65389588-65389588 on Build GRCh38
5899815 c.1262G>A p.G421E Chr.1: 65389592-65389592 on Build GRCh38
3491787 c.1348T>G p.S450A Chr.1: 65389410-65389410 on Build GRCh38
6240518 c.1468+1G>A p.? Chr.1: 65389628-65389628 on Build GRCh38
426592 c.1260C>A p.Y420* Chr.1: 65389590-65389590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763701367 Chr.1: 65389673-65389673 on Build GRCh38
rs778250210 Chr.1: 65389301-65389301 on Build GRCh38
rs976248784 Chr.1: 65389447-65389447 on Build GRCh38
rs144003387 Chr.1: 65389393-65389393 on Build GRCh38
rs760541769 Chr.1: 65389478-65389478 on Build GRCh38
rs769980617 Chr.1: 65389458-65389458 on Build GRCh38
rs758966341 Chr.1: 65389517-65389517 on Build GRCh38
rs78141380 Chr.1: 65389615-65389615 on Build GRCh38
rs759463088 Chr.1: 65389657-65389657 on Build GRCh38
rs568329793 Chr.1: 65389619-65389619 on Build GRCh38
rs914394613 Chr.1: 65389697-65389697 on Build GRCh38
rs367817786 Chr.1: 65389469-65389469 on Build GRCh38
rs374833232 Chr.1: 65389314-65389314 on Build GRCh38
rs148673423 Chr.1: 65389436-65389436 on Build GRCh38
rs746624365 Chr.1: 65389700-65389700 on Build GRCh38
rs372678776 Chr.1: 65389516-65389516 on Build GRCh38
rs753102342 Chr.1: 65389383-65389383 on Build GRCh38
rs768950366 Chr.1: 65389601-65389601 on Build GRCh38
rs771595238 Chr.1: 65389655-65389655 on Build GRCh38
rs775839311 Chr.1: 65389461-65389461 on Build GRCh38
rs563941772 Chr.1: 65389287-65389287 on Build GRCh38
rs761693558 Chr.1: 65389368-65389368 on Build GRCh38
rs1043737289 Chr.1: 65389284-65389284 on Build GRCh38
rs770826929 Chr.1: 65389649-65389649 on Build GRCh38
rs372634207 Chr.1: 65389527-65389527 on Build GRCh38
rs142136708 Chr.1: 65389382-65389382 on Build GRCh38
rs769881010 Chr.1: 65389339-65389339 on Build GRCh38
rs757677534 Chr.1: 65389544-65389544 on Build GRCh38
rs767476436 Chr.1: 65389371-65389371 on Build GRCh38
rs778651457 Chr.1: 65389271-65389271 on Build GRCh38
rs528270090 Chr.1: 65389558-65389558 on Build GRCh38
rs757982796 Chr.1: 65389528-65389528 on Build GRCh38
rs753239343 Chr.1: 65389379-65389379 on Build GRCh38
rs763771047 Chr.1: 65389538-65389538 on Build GRCh38
rs748491396 Chr.1: 65389274-65389274 on Build GRCh38
rs1022699921 Chr.1: 65389591-65389591 on Build GRCh38
rs765168146 Chr.1: 65389664-65389664 on Build GRCh38
rs766371179 Chr.1: 65389377-65389377 on Build GRCh38
rs745350696 Chr.1: 65389428-65389428 on Build GRCh38
rs781745301 Chr.1: 65389448-65389448 on Build GRCh38
rs756700209 Chr.1: 65389574-65389574 on Build GRCh38
rs750256177 Chr.1: 65389372-65389372 on Build GRCh38
rs764787385 Chr.1: 65389521-65389521 on Build GRCh38
rs778494257 Chr.1: 65389493-65389493 on Build GRCh38
rs138800944 Chr.1: 65389381-65389381 on Build GRCh38
rs1044896664 Chr.1: 65389529-65389529 on Build GRCh38
rs762494027 Chr.1: 65389670-65389670 on Build GRCh38
rs531245733 Chr.1: 65389696-65389696 on Build GRCh38
rs752416727 Chr.1: 65389416-65389416 on Build GRCh38
rs758752612 Chr.1: 65389420-65389420 on Build GRCh38
rs778057827 Chr.1: 65389421-65389421 on Build GRCh38
rs766206503 Chr.1: 65389479-65389479 on Build GRCh38
rs749462312 Chr.1: 65389462-65389462 on Build GRCh38
rs202109326 Chr.1: 65389412-65389412 on Build GRCh38
rs772468365 Chr.1: 65389280-65389280 on Build GRCh38
rs775892105 Chr.1: 65389344-65389344 on Build GRCh38
rs745312421 Chr.1: 65389567-65389567 on Build GRCh38
rs745902496 Chr.1: 65389451-65389451 on Build GRCh38
rs61758427 Chr.1: 65389328-65389328 on Build GRCh38
rs1003128731 Chr.1: 65389581-65389581 on Build GRCh38
rs376004077 Chr.1: 65389652-65389652 on Build GRCh38
rs761176258 Chr.1: 65389507-65389507 on Build GRCh38
rs1049298126 Chr.1: 65389511-65389511 on Build GRCh38
rs368276625 Chr.1: 65389562-65389562 on Build GRCh38
rs759679738 Chr.1: 65389332-65389332 on Build GRCh38
rs769003540 Chr.1: 65389464-65389464 on Build GRCh38
rs751844844 Chr.1: 65389541-65389541 on Build GRCh38
rs747408305 Chr.1: 65389425-65389425 on Build GRCh38
rs202225808 Chr.1: 65389552-65389552 on Build GRCh38
rs774297201 Chr.1: 65389595-65389596 on Build GRCh38
rs878918182 Chr.1: 65389677-65389677 on Build GRCh38
rs139447717 Chr.1: 65389590-65389590 on Build GRCh38
rs189783499 Chr.1: 65389376-65389376 on Build GRCh38
rs956148381 Chr.1: 65389680-65389680 on Build GRCh38
rs774945685 Chr.1: 65389667-65389667 on Build GRCh38
rs555883956 Chr.1: 65389710-65389710 on Build GRCh38
rs372053013 Chr.1: 65389579-65389579 on Build GRCh38

View Full Product Details