Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67128332-67128874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf141
Entrez Gene ID
400757
Gene Name
chromosome 1 open reading frame 141
Gene Aliases
-
UniGene
Hs.666621
Gene Chromosome Location
Chr.1: 67092176-67141646 on Build GRCh38
Transcripts
NM_001276351.1 NM_001276352.1 NR_075077.1 XM_011541465.2 XM_011541466.2 XM_011541467.1 XM_011541472.1 XM_011541473.2 XM_011541475.1 XM_017001276.1 XM_017001277.1

SNP Detail

SNP ID Chromosome Location
rs376887015 Chr.1: 67128828-67128828 on Build GRCh38
rs911076922 Chr.1: 67128601-67128601 on Build GRCh38
rs986164904 Chr.1: 67128591-67128591 on Build GRCh38
rs1034744812 Chr.1: 67128773-67128773 on Build GRCh38
rs1047738458 Chr.1: 67128390-67128390 on Build GRCh38
rs932038035 Chr.1: 67128574-67128574 on Build GRCh38
rs976950756 Chr.1: 67128767-67128767 on Build GRCh38
rs899438616 Chr.1: 67128384-67128384 on Build GRCh38
rs768404477 Chr.1: 67128693-67128693 on Build GRCh38
rs553173973 Chr.1: 67128706-67128706 on Build GRCh38
rs377607017 Chr.1: 67128662-67128662 on Build GRCh38
rs970875101 Chr.1: 67128463-67128463 on Build GRCh38
rs989139373 Chr.1: 67128480-67128480 on Build GRCh38
rs760308243 Chr.1: 67128358-67128377 on Build GRCh38
rs918527247 Chr.1: 67128771-67128771 on Build GRCh38
rs200034463 Chr.1: 67128381-67128381 on Build GRCh38
rs541580098 Chr.1: 67128799-67128799 on Build GRCh38
rs1008944111 Chr.1: 67128402-67128402 on Build GRCh38
rs1046564436 Chr.1: 67128356-67128357 on Build GRCh38
rs928107102 Chr.1: 67128356-67128377 on Build GRCh38
rs965190528 Chr.1: 67128755-67128755 on Build GRCh38
rs117531913 Chr.1: 67128820-67128820 on Build GRCh38
rs796636743 Chr.1: 67128369-67128389 on Build GRCh38
rs1012441607 Chr.1: 67128441-67128441 on Build GRCh38
rs1040998637 Chr.1: 67128615-67128615 on Build GRCh38
rs1003707153 Chr.1: 67128674-67128674 on Build GRCh38
rs911967695 Chr.1: 67128744-67128744 on Build GRCh38
rs112415517 Chr.1: 67128389-67128389 on Build GRCh38
rs111998730 Chr.1: 67128369-67128369 on Build GRCh38
rs1023782645 Chr.1: 67128455-67128455 on Build GRCh38
rs556104298 Chr.1: 67128512-67128512 on Build GRCh38
rs989509023 Chr.1: 67128708-67128708 on Build GRCh38
rs943870124 Chr.1: 67128606-67128606 on Build GRCh38
rs572711551 Chr.1: 67128740-67128740 on Build GRCh38
rs897028119 Chr.1: 67128616-67128616 on Build GRCh38
rs200632204 Chr.1: 67128354-67128377 on Build GRCh38
rs927720226 Chr.1: 67128547-67128547 on Build GRCh38
rs991407150 Chr.1: 67128592-67128592 on Build GRCh38
rs12124695 Chr.1: 67128659-67128659 on Build GRCh38
rs746066754 Chr.1: 67128726-67128726 on Build GRCh38
rs1047685984 Chr.1: 67128626-67128626 on Build GRCh38
rs148217962 Chr.1: 67128385-67128385 on Build GRCh38
rs199795044 Chr.1: 67128354-67128369 on Build GRCh38
rs142954688 Chr.1: 67128354-67128373 on Build GRCh38
rs986217252 Chr.1: 67128723-67128723 on Build GRCh38
rs1026741325 Chr.1: 67128400-67128400 on Build GRCh38
rs868577382 Chr.1: 67128386-67128386 on Build GRCh38
rs888152048 Chr.1: 67128440-67128440 on Build GRCh38
rs1048296558 Chr.1: 67128803-67128803 on Build GRCh38
rs796354684 Chr.1: 67128774-67128774 on Build GRCh38
rs1853011 Chr.1: 67128571-67128571 on Build GRCh38
rs576900388 Chr.1: 67128691-67128691 on Build GRCh38
rs1031669008 Chr.1: 67128474-67128474 on Build GRCh38
rs938214739 Chr.1: 67128374-67128374 on Build GRCh38
rs1010838923 Chr.1: 67128708-67128709 on Build GRCh38
rs906247677 Chr.1: 67128842-67128842 on Build GRCh38
rs906740992 Chr.1: 67128666-67128666 on Build GRCh38
rs201961938 Chr.1: 67128377-67128377 on Build GRCh38
rs913575587 Chr.1: 67128493-67128493 on Build GRCh38
rs756181803 Chr.1: 67128696-67128696 on Build GRCh38
rs886486249 Chr.1: 67128401-67128401 on Build GRCh38
rs920831424 Chr.1: 67128605-67128605 on Build GRCh38
rs1022746299 Chr.1: 67128718-67128718 on Build GRCh38
rs958209803 Chr.1: 67128478-67128478 on Build GRCh38
rs930893254 Chr.1: 67128612-67128612 on Build GRCh38
rs956766244 Chr.1: 67128477-67128477 on Build GRCh38
rs900614666 Chr.1: 67128679-67128679 on Build GRCh38
rs981852544 Chr.1: 67128546-67128546 on Build GRCh38
rs117252944 Chr.1: 67128701-67128701 on Build GRCh38
rs1055363074 Chr.1: 67128374-67128382 on Build GRCh38
rs989853237 Chr.1: 67128522-67128522 on Build GRCh38
rs1020920206 Chr.1: 67128709-67128709 on Build GRCh38
rs1048513761 Chr.1: 67128652-67128652 on Build GRCh38
rs953360574 Chr.1: 67128720-67128720 on Build GRCh38
rs142001088 Chr.1: 67128641-67128641 on Build GRCh38
rs865839152 Chr.1: 67128660-67128660 on Build GRCh38
rs982386763 Chr.1: 67128469-67128469 on Build GRCh38
rs72676041 Chr.1: 67128839-67128839 on Build GRCh38

View Full Product Details