Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68137697-68138208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12-AS1
Entrez Gene ID
100289178
Gene Name
GNG12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.677603
Gene Chromosome Location
Chr.1: 67832288-68202987 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
UniGene
Hs.647659
Gene Chromosome Location
Chr.1: 68098459-68233000 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3492053 c.1402C>T p.Q468* Chr.1: 68137894-68137894 on Build GRCh38
3805842 c.1484A>C p.K495T Chr.1: 68137806-68137806 on Build GRCh38
2240450 c.1393G>T p.V465F Chr.1: 68137897-68137897 on Build GRCh38
3805844 c.1449C>G p.V483V Chr.1: 68137841-68137841 on Build GRCh38
5414636 c.1374G>A p.W458* Chr.1: 68137916-68137916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750483989 Chr.1: 68137948-68137948 on Build GRCh38
rs753379698 Chr.1: 68137767-68137767 on Build GRCh38
rs367885134 Chr.1: 68137800-68137800 on Build GRCh38
rs370965179 Chr.1: 68137755-68137755 on Build GRCh38
rs41410344 Chr.1: 68137762-68137762 on Build GRCh38
rs889633615 Chr.1: 68137997-68137997 on Build GRCh38
rs557987531 Chr.1: 68137938-68137938 on Build GRCh38
rs776580033 Chr.1: 68137983-68137983 on Build GRCh38
rs952397370 Chr.1: 68138086-68138086 on Build GRCh38
rs754765677 Chr.1: 68137891-68137891 on Build GRCh38
rs140465138 Chr.1: 68137929-68137929 on Build GRCh38
rs746134565 Chr.1: 68137745-68137745 on Build GRCh38
rs1026343968 Chr.1: 68138077-68138077 on Build GRCh38
rs966003796 Chr.1: 68138067-68138067 on Build GRCh38
rs764816861 Chr.1: 68137912-68137912 on Build GRCh38
rs747919046 Chr.1: 68137892-68137892 on Build GRCh38
rs202246168 Chr.1: 68137730-68137730 on Build GRCh38
rs200884626 Chr.1: 68137751-68137751 on Build GRCh38
rs756171146 Chr.1: 68137950-68137950 on Build GRCh38
rs766409566 Chr.1: 68137904-68137904 on Build GRCh38
rs753014700 Chr.1: 68137798-68137798 on Build GRCh38
rs1831297 Chr.1: 68138048-68138048 on Build GRCh38
rs768640630 Chr.1: 68137800-68137800 on Build GRCh38
rs542708578 Chr.1: 68138092-68138092 on Build GRCh38
rs771268915 Chr.1: 68138167-68138167 on Build GRCh38
rs764940216 Chr.1: 68137782-68137782 on Build GRCh38
rs775725554 Chr.1: 68137867-68137867 on Build GRCh38
rs192499054 Chr.1: 68137962-68137962 on Build GRCh38
rs534138530 Chr.1: 68137974-68137974 on Build GRCh38
rs993481488 Chr.1: 68138068-68138068 on Build GRCh38
rs142571662 Chr.1: 68137829-68137829 on Build GRCh38
rs547376883 Chr.1: 68137753-68137753 on Build GRCh38
rs757739330 Chr.1: 68137817-68137817 on Build GRCh38
rs768981171 Chr.1: 68137960-68137960 on Build GRCh38
rs765541986 Chr.1: 68137856-68137856 on Build GRCh38
rs769915437 Chr.1: 68137834-68137834 on Build GRCh38
rs776656746 Chr.1: 68137744-68137744 on Build GRCh38
rs1052938372 Chr.1: 68137968-68137968 on Build GRCh38
rs1003649495 Chr.1: 68137773-68137773 on Build GRCh38
rs558440498 Chr.1: 68137919-68137919 on Build GRCh38
rs985347096 Chr.1: 68137863-68137863 on Build GRCh38
rs983034 Chr.1: 68137903-68137903 on Build GRCh38
rs749565374 Chr.1: 68137956-68137956 on Build GRCh38
rs932737839 Chr.1: 68138110-68138110 on Build GRCh38
rs754647145 Chr.1: 68137769-68137769 on Build GRCh38
rs749059713 Chr.1: 68137801-68137801 on Build GRCh38
rs745953882 Chr.1: 68137830-68137830 on Build GRCh38
rs916122325 Chr.1: 68137868-68137868 on Build GRCh38
rs771860961 Chr.1: 68137895-68137895 on Build GRCh38
rs747984899 Chr.1: 68137969-68137969 on Build GRCh38
rs778280457 Chr.1: 68137804-68137804 on Build GRCh38
rs139409633 Chr.1: 68137811-68137811 on Build GRCh38
rs780172146 Chr.1: 68137953-68137953 on Build GRCh38
rs1049773189 Chr.1: 68138130-68138130 on Build GRCh38
rs537667082 Chr.1: 68137907-68137907 on Build GRCh38
rs760293011 Chr.1: 68137758-68137758 on Build GRCh38
rs772057198 Chr.1: 68137989-68137989 on Build GRCh38
rs1011531549 Chr.1: 68137988-68137988 on Build GRCh38
rs770958386 Chr.1: 68137731-68137731 on Build GRCh38
rs1430754 Chr.1: 68137749-68137749 on Build GRCh38
rs1008015444 Chr.1: 68138035-68138035 on Build GRCh38
rs1050016932 Chr.1: 68137996-68137996 on Build GRCh38
rs916032651 Chr.1: 68138134-68138134 on Build GRCh38
rs893036769 Chr.1: 68137980-68137980 on Build GRCh38
rs915428245 Chr.1: 68138171-68138171 on Build GRCh38
rs1028181391 Chr.1: 68137734-68137734 on Build GRCh38
rs773115339 Chr.1: 68137898-68137898 on Build GRCh38
rs746798181 Chr.1: 68137963-68137963 on Build GRCh38
rs553801543 Chr.1: 68138028-68138028 on Build GRCh38
rs370568408 Chr.1: 68137937-68137937 on Build GRCh38
rs777772233 Chr.1: 68137961-68137961 on Build GRCh38
rs747155608 Chr.1: 68138091-68138091 on Build GRCh38

View Full Product Details