Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74198080-74198608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
UniGene
Hs.644625
Gene Chromosome Location
Chr.1: 74026015-74198201 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5651417 c.82+5C>T p.? Chr.1: 74198365-74198365 on Build GRCh38
6540543 c.73G>T p.E25* Chr.1: 74198351-74198351 on Build GRCh38
6064418 c.72_73CG>GA p.E25K Chr.1: 74198350-74198351 on Build GRCh38
4538791 c.25G>A p.E9K Chr.1: 74198303-74198303 on Build GRCh38
5440611 c.28G>T p.V10L Chr.1: 74198306-74198306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043601904 Chr.1: 74198307-74198307 on Build GRCh38
rs767611770 Chr.1: 74198244-74198244 on Build GRCh38
rs569881223 Chr.1: 74198186-74198186 on Build GRCh38
rs779800359 Chr.1: 74198263-74198263 on Build GRCh38
rs776043426 Chr.1: 74198467-74198467 on Build GRCh38
rs913633834 Chr.1: 74198563-74198563 on Build GRCh38
rs777721429 Chr.1: 74198254-74198254 on Build GRCh38
rs933635238 Chr.1: 74198553-74198553 on Build GRCh38
rs752502862 Chr.1: 74198250-74198250 on Build GRCh38
rs776193087 Chr.1: 74198204-74198204 on Build GRCh38
rs767517976 Chr.1: 74198347-74198347 on Build GRCh38
rs760683467 Chr.1: 74198450-74198450 on Build GRCh38
rs200584512 Chr.1: 74198348-74198348 on Build GRCh38
rs1011752317 Chr.1: 74198432-74198432 on Build GRCh38
rs781248089 Chr.1: 74198425-74198425 on Build GRCh38
rs764626083 Chr.1: 74198212-74198212 on Build GRCh38
rs200750484 Chr.1: 74198268-74198268 on Build GRCh38
rs201134169 Chr.1: 74198291-74198291 on Build GRCh38
rs766987339 Chr.1: 74198319-74198319 on Build GRCh38
rs745516648 Chr.1: 74198228-74198228 on Build GRCh38
rs149849239 Chr.1: 74198558-74198558 on Build GRCh38
rs983319531 Chr.1: 74198388-74198388 on Build GRCh38
rs758142053 Chr.1: 74198252-74198252 on Build GRCh38
rs748674628 Chr.1: 74198269-74198269 on Build GRCh38
rs745885732 Chr.1: 74198431-74198431 on Build GRCh38
rs746461735 Chr.1: 74198194-74198194 on Build GRCh38
rs940474044 Chr.1: 74198562-74198562 on Build GRCh38
rs199832542 Chr.1: 74198380-74198380 on Build GRCh38
rs77030123 Chr.1: 74198167-74198167 on Build GRCh38
rs529372202 Chr.1: 74198477-74198477 on Build GRCh38
rs778386034 Chr.1: 74198407-74198407 on Build GRCh38
rs76672163 Chr.1: 74198247-74198247 on Build GRCh38
rs755731800 Chr.1: 74198360-74198360 on Build GRCh38
rs750352320 Chr.1: 74198356-74198356 on Build GRCh38
rs758336426 Chr.1: 74198222-74198222 on Build GRCh38
rs565972704 Chr.1: 74198151-74198151 on Build GRCh38
rs201738008 Chr.1: 74198315-74198315 on Build GRCh38
rs916442072 Chr.1: 74198281-74198281 on Build GRCh38
rs771739721 Chr.1: 74198447-74198447 on Build GRCh38
rs186026131 Chr.1: 74198436-74198436 on Build GRCh38
rs773346127 Chr.1: 74198277-74198277 on Build GRCh38
rs766352159 Chr.1: 74198457-74198457 on Build GRCh38
rs199645291 Chr.1: 74198330-74198330 on Build GRCh38
rs147701183 Chr.1: 74198327-74198327 on Build GRCh38
rs763348248 Chr.1: 74198233-74198233 on Build GRCh38
rs543637299 Chr.1: 74198446-74198446 on Build GRCh38
rs760310249 Chr.1: 74198427-74198428 on Build GRCh38
rs751513361 Chr.1: 74198295-74198295 on Build GRCh38
rs770438286 Chr.1: 74198225-74198225 on Build GRCh38
rs200371237 Chr.1: 74198266-74198266 on Build GRCh38
rs759010567 Chr.1: 74198313-74198336 on Build GRCh38
rs752698904 Chr.1: 74198221-74198221 on Build GRCh38
rs1051276580 Chr.1: 74198278-74198278 on Build GRCh38
rs201711657 Chr.1: 74198391-74198391 on Build GRCh38
rs770278938 Chr.1: 74198273-74198273 on Build GRCh38
rs770505202 Chr.1: 74198196-74198196 on Build GRCh38
rs371399271 Chr.1: 74198260-74198260 on Build GRCh38
rs984659437 Chr.1: 74198535-74198535 on Build GRCh38
rs774878969 Chr.1: 74198239-74198239 on Build GRCh38
rs1004513676 Chr.1: 74198182-74198182 on Build GRCh38
rs770968758 Chr.1: 74198324-74198324 on Build GRCh38
rs951611367 Chr.1: 74198246-74198246 on Build GRCh38
rs546653717 Chr.1: 74198122-74198122 on Build GRCh38
rs767295769 Chr.1: 74198297-74198297 on Build GRCh38
rs574465280 Chr.1: 74198265-74198265 on Build GRCh38
rs748925611 Chr.1: 74198302-74198302 on Build GRCh38
rs747123659 Chr.1: 74198316-74198316 on Build GRCh38
rs1023563015 Chr.1: 74198487-74198487 on Build GRCh38
rs930424424 Chr.1: 74198160-74198160 on Build GRCh38
rs771058925 Chr.1: 74198416-74198416 on Build GRCh38
rs1412825 Chr.1: 74198130-74198130 on Build GRCh38
rs755449906 Chr.1: 74198303-74198303 on Build GRCh38
rs749528030 Chr.1: 74198351-74198351 on Build GRCh38
rs761462169 Chr.1: 74198342-74198342 on Build GRCh38
rs975795253 Chr.1: 74198573-74198573 on Build GRCh38
rs148983142 Chr.1: 74198301-74198301 on Build GRCh38
rs201917402 Chr.1: 74198276-74198276 on Build GRCh38
rs763858437 Chr.1: 74198294-74198294 on Build GRCh38
rs778748702 Chr.1: 74198109-74198109 on Build GRCh38
rs886151973 Chr.1: 74198170-74198170 on Build GRCh38
rs770072561 Chr.1: 74198435-74198435 on Build GRCh38
rs770166566 Chr.1: 74198203-74198203 on Build GRCh38
rs991450839 Chr.1: 74198469-74198469 on Build GRCh38
rs200152525 Chr.1: 74198238-74198238 on Build GRCh38
rs139721001 Chr.1: 74198117-74198117 on Build GRCh38
rs201008457 Chr.1: 74198317-74198317 on Build GRCh38
rs753412149 Chr.1: 74198459-74198459 on Build GRCh38
rs751698525 Chr.1: 74198215-74198215 on Build GRCh38
rs1014279846 Chr.1: 74198372-74198372 on Build GRCh38
rs751486239 Chr.1: 74198255-74198255 on Build GRCh38
rs760690123 Chr.1: 74198284-74198284 on Build GRCh38
rs752398689 Chr.1: 74198384-74198386 on Build GRCh38
rs762232739 Chr.1: 74198251-74198255 on Build GRCh38
rs551677653 Chr.1: 74198179-74198179 on Build GRCh38
rs772647312 Chr.1: 74198311-74198311 on Build GRCh38
rs780744071 Chr.1: 74198258-74198258 on Build GRCh38
rs375715390 Chr.1: 74198403-74198403 on Build GRCh38
rs753129487 Chr.1: 74198394-74198394 on Build GRCh38
rs200727460 Chr.1: 74198387-74198387 on Build GRCh38
rs571970784 Chr.1: 74198341-74198341 on Build GRCh38
rs761837940 Chr.1: 74198243-74198243 on Build GRCh38
rs374334766 Chr.1: 74198275-74198275 on Build GRCh38
rs776299899 Chr.1: 74198197-74198197 on Build GRCh38
rs903746972 Chr.1: 74198314-74198314 on Build GRCh38
rs756769942 Chr.1: 74198256-74198256 on Build GRCh38
rs952027028 Chr.1: 74198522-74198522 on Build GRCh38
rs778664916 Chr.1: 74198220-74198220 on Build GRCh38
rs750626015 Chr.1: 74198216-74198216 on Build GRCh38
rs185634617 Chr.1: 74198147-74198147 on Build GRCh38
rs199558076 Chr.1: 74198292-74198292 on Build GRCh38
rs748393876 Chr.1: 74198305-74198305 on Build GRCh38
rs917369416 Chr.1: 74198223-74198223 on Build GRCh38
rs1039988060 Chr.1: 74198184-74198184 on Build GRCh38
rs200343931 Chr.1: 74198287-74198287 on Build GRCh38
rs967402346 Chr.1: 74198106-74198106 on Build GRCh38
rs149824098 Chr.1: 74198279-74198279 on Build GRCh38
rs766611619 Chr.1: 74198286-74198286 on Build GRCh38
rs200593681 Chr.1: 74198409-74198409 on Build GRCh38
rs200292408 Chr.1: 74198231-74198231 on Build GRCh38
rs201671455 Chr.1: 74198235-74198235 on Build GRCh38
rs763544912 Chr.1: 74198210-74198210 on Build GRCh38
rs368869827 Chr.1: 74198159-74198159 on Build GRCh38
rs202083917 Chr.1: 74198259-74198259 on Build GRCh38
rs749968533 Chr.1: 74198300-74198300 on Build GRCh38
rs758848042 Chr.1: 74198472-74198472 on Build GRCh38
rs765972262 Chr.1: 74198361-74198361 on Build GRCh38
rs766521579 Chr.1: 74198249-74198249 on Build GRCh38
rs754777825 Chr.1: 74198218-74198218 on Build GRCh38
rs777631676 Chr.1: 74198224-74198224 on Build GRCh38
rs1050926512 Chr.1: 74198126-74198126 on Build GRCh38
rs4422957 Chr.1: 74198465-74198465 on Build GRCh38
rs200018409 Chr.1: 74198398-74198398 on Build GRCh38
rs945154604 Chr.1: 74198583-74198583 on Build GRCh38
rs922979224 Chr.1: 74198580-74198580 on Build GRCh38
rs779331179 Chr.1: 74198304-74198304 on Build GRCh38
rs758922686 Chr.1: 74198464-74198464 on Build GRCh38
rs181548075 Chr.1: 74198366-74198366 on Build GRCh38
rs773873767 Chr.1: 74198277-74198277 on Build GRCh38

View Full Product Details