Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74569419-74569929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

SNP Detail

SNP ID Chromosome Location
rs1024898810 Chr.1: 74569664-74569664 on Build GRCh38
rs919894697 Chr.1: 74569764-74569764 on Build GRCh38
rs546473017 Chr.1: 74569838-74569838 on Build GRCh38
rs35856741 Chr.1: 74569505-74569506 on Build GRCh38
rs189125992 Chr.1: 74569507-74569507 on Build GRCh38
rs1039768677 Chr.1: 74569864-74569864 on Build GRCh38
rs919188583 Chr.1: 74569727-74569727 on Build GRCh38
rs959061512 Chr.1: 74569603-74569603 on Build GRCh38
rs932658649 Chr.1: 74569794-74569794 on Build GRCh38
rs543111693 Chr.1: 74569500-74569500 on Build GRCh38
rs577316087 Chr.1: 74569505-74569505 on Build GRCh38
rs542634174 Chr.1: 74569568-74569568 on Build GRCh38
rs1021095538 Chr.1: 74569533-74569533 on Build GRCh38
rs902721071 Chr.1: 74569901-74569901 on Build GRCh38
rs534098264 Chr.1: 74569795-74569795 on Build GRCh38
rs1036172698 Chr.1: 74569721-74569721 on Build GRCh38
rs944317998 Chr.1: 74569821-74569821 on Build GRCh38
rs941845263 Chr.1: 74569720-74569720 on Build GRCh38
rs902248392 Chr.1: 74569542-74569542 on Build GRCh38
rs903926770 Chr.1: 74569872-74569872 on Build GRCh38
rs1012137184 Chr.1: 74569661-74569661 on Build GRCh38
rs907728773 Chr.1: 74569725-74569725 on Build GRCh38
rs970525034 Chr.1: 74569670-74569670 on Build GRCh38
rs6659750 Chr.1: 74569804-74569804 on Build GRCh38
rs980539773 Chr.1: 74569555-74569555 on Build GRCh38
rs187630574 Chr.1: 74569551-74569551 on Build GRCh38
rs565023040 Chr.1: 74569848-74569848 on Build GRCh38
rs748537094 Chr.1: 74569608-74569608 on Build GRCh38
rs1008416801 Chr.1: 74569525-74569525 on Build GRCh38
rs1000613627 Chr.1: 74569904-74569904 on Build GRCh38
rs1007418424 Chr.1: 74569858-74569858 on Build GRCh38
rs992264814 Chr.1: 74569480-74569480 on Build GRCh38
rs767492969 Chr.1: 74569562-74569562 on Build GRCh38
rs961463731 Chr.1: 74569732-74569732 on Build GRCh38
rs12728662 Chr.1: 74569556-74569556 on Build GRCh38
rs933238478 Chr.1: 74569788-74569788 on Build GRCh38
rs987550169 Chr.1: 74569521-74569521 on Build GRCh38
rs974600162 Chr.1: 74569752-74569752 on Build GRCh38

View Full Product Details