Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74714419-74714951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZ
Entrez Gene ID
1429
Gene Name
crystallin zeta
Gene Aliases
-
UniGene
Hs.83114
Gene Chromosome Location
Chr.1: 74705482-74733408 on Build GRCh38
Transcripts
NM_001130042.1 NM_001130043.1 NM_001134759.1 NM_001889.3 XM_005270491.4 XM_011540747.1 XM_017000367.1 XM_017000368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805884 c.434G>C p.C145S Chr.1: 74714625-74714625 on Build GRCh38
2241721 c.463C>A p.H155N Chr.1: 74714596-74714596 on Build GRCh38
5824133 c.469G>A p.A157T Chr.1: 74714590-74714590 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370178975 Chr.1: 74714772-74714772 on Build GRCh38
rs960069392 Chr.1: 74714887-74714887 on Build GRCh38
rs761861191 Chr.1: 74714562-74714562 on Build GRCh38
rs781274574 Chr.1: 74714535-74714535 on Build GRCh38
rs781186543 Chr.1: 74714642-74714642 on Build GRCh38
rs368614504 Chr.1: 74714661-74714661 on Build GRCh38
rs377055371 Chr.1: 74714854-74714854 on Build GRCh38
rs34112618 Chr.1: 74714925-74714926 on Build GRCh38
rs759903237 Chr.1: 74714616-74714616 on Build GRCh38
rs894360011 Chr.1: 74714631-74714631 on Build GRCh38
rs776508484 Chr.1: 74714539-74714540 on Build GRCh38
rs938360374 Chr.1: 74714626-74714626 on Build GRCh38
rs768648785 Chr.1: 74714541-74714541 on Build GRCh38
rs58387974 Chr.1: 74714830-74714830 on Build GRCh38
rs539874084 Chr.1: 74714694-74714694 on Build GRCh38
rs147153422 Chr.1: 74714494-74714494 on Build GRCh38
rs753213162 Chr.1: 74714625-74714625 on Build GRCh38
rs772976288 Chr.1: 74714561-74714561 on Build GRCh38
rs914607504 Chr.1: 74714761-74714761 on Build GRCh38
rs149676618 Chr.1: 74714649-74714649 on Build GRCh38
rs1030760291 Chr.1: 74714765-74714765 on Build GRCh38
rs765225008 Chr.1: 74714563-74714564 on Build GRCh38
rs765947574 Chr.1: 74714601-74714601 on Build GRCh38
rs548322101 Chr.1: 74714804-74714804 on Build GRCh38
rs772711684 Chr.1: 74714707-74714707 on Build GRCh38
rs201041656 Chr.1: 74714540-74714540 on Build GRCh38
rs965220561 Chr.1: 74714685-74714685 on Build GRCh38
rs1037119859 Chr.1: 74714872-74714872 on Build GRCh38
rs750352525 Chr.1: 74714572-74714575 on Build GRCh38
rs139512669 Chr.1: 74714627-74714627 on Build GRCh38
rs541278533 Chr.1: 74714899-74714899 on Build GRCh38
rs760423890 Chr.1: 74714564-74714564 on Build GRCh38
rs527342267 Chr.1: 74714450-74714450 on Build GRCh38
rs765762437 Chr.1: 74714617-74714617 on Build GRCh38
rs756145995 Chr.1: 74714739-74714739 on Build GRCh38
rs948710091 Chr.1: 74714785-74714785 on Build GRCh38
rs758455007 Chr.1: 74714738-74714739 on Build GRCh38
rs953632023 Chr.1: 74714770-74714770 on Build GRCh38
rs1026687398 Chr.1: 74714658-74714658 on Build GRCh38
rs758599907 Chr.1: 74714576-74714577 on Build GRCh38
rs774209827 Chr.1: 74714544-74714544 on Build GRCh38
rs74095153 Chr.1: 74714650-74714650 on Build GRCh38
rs977596537 Chr.1: 74714848-74714848 on Build GRCh38
rs79201095 Chr.1: 74714641-74714641 on Build GRCh38
rs6679213 Chr.1: 74714513-74714513 on Build GRCh38
rs749322238 Chr.1: 74714890-74714890 on Build GRCh38
rs781190292 Chr.1: 74714806-74714806 on Build GRCh38
rs771799366 Chr.1: 74714557-74714557 on Build GRCh38
rs74095154 Chr.1: 74714717-74714717 on Build GRCh38
rs776285088 Chr.1: 74714613-74714613 on Build GRCh38
rs574378715 Chr.1: 74714849-74714849 on Build GRCh38
rs889439642 Chr.1: 74714660-74714660 on Build GRCh38
rs990631435 Chr.1: 74714721-74714721 on Build GRCh38
rs758759996 Chr.1: 74714628-74714628 on Build GRCh38
rs748082936 Chr.1: 74714653-74714653 on Build GRCh38
rs35020802 Chr.1: 74714718-74714719 on Build GRCh38
rs773170702 Chr.1: 74714656-74714656 on Build GRCh38
rs1002971389 Chr.1: 74714638-74714638 on Build GRCh38
rs747967831 Chr.1: 74714546-74714546 on Build GRCh38
rs143597042 Chr.1: 74714632-74714632 on Build GRCh38
rs745944096 Chr.1: 74714645-74714645 on Build GRCh38
rs200579063 Chr.1: 74714599-74714599 on Build GRCh38
rs368335432 Chr.1: 74714684-74714684 on Build GRCh38
rs116332406 Chr.1: 74714715-74714715 on Build GRCh38
rs771850610 Chr.1: 74714655-74714655 on Build GRCh38
rs879355360 Chr.1: 74714823-74714823 on Build GRCh38

View Full Product Details