Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75732711-75733254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3492382 c.226C>T p.P76S Chr.1: 75732862-75732862 on Build GRCh38
5128301 c.229-2A>G p.? Chr.1: 75732851-75732851 on Build GRCh38
6452793 c.208G>A p.E70K Chr.1: 75732721-75732721 on Build GRCh38
5987543 c.241C>G p.L81V Chr.1: 75732865-75732865 on Build GRCh38
5673044 c.245G>T p.W82L Chr.1: 75732881-75732881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111647855 Chr.1: 75732960-75732960 on Build GRCh38
rs780692978 Chr.1: 75732814-75732814 on Build GRCh38
rs766134786 Chr.1: 75733197-75733197 on Build GRCh38
rs767649048 Chr.1: 75732896-75732896 on Build GRCh38
rs755862445 Chr.1: 75733115-75733115 on Build GRCh38
rs141043231 Chr.1: 75732904-75732904 on Build GRCh38
rs534657889 Chr.1: 75733156-75733156 on Build GRCh38
rs746655981 Chr.1: 75732808-75732808 on Build GRCh38
rs750526774 Chr.1: 75732781-75732781 on Build GRCh38
rs749859620 Chr.1: 75732865-75732865 on Build GRCh38
rs767630461 Chr.1: 75732780-75732780 on Build GRCh38
rs745371854 Chr.1: 75732840-75732840 on Build GRCh38
rs758213925 Chr.1: 75732795-75732795 on Build GRCh38
rs779749395 Chr.1: 75733017-75733017 on Build GRCh38
rs75792854 Chr.1: 75732966-75732966 on Build GRCh38
rs764020215 Chr.1: 75733091-75733091 on Build GRCh38
rs770293389 Chr.1: 75732861-75732861 on Build GRCh38
rs749508931 Chr.1: 75733196-75733196 on Build GRCh38
rs758476299 Chr.1: 75732874-75732874 on Build GRCh38
rs151126827 Chr.1: 75732867-75732867 on Build GRCh38
rs1057516801 Chr.1: 75732742-75732742 on Build GRCh38
rs555153199 Chr.1: 75733083-75733083 on Build GRCh38
rs776622010 Chr.1: 75733159-75733159 on Build GRCh38
rs772304227 Chr.1: 75733148-75733148 on Build GRCh38
rs398123074 Chr.1: 75732869-75732869 on Build GRCh38
rs901359252 Chr.1: 75732977-75732977 on Build GRCh38
rs115877127 Chr.1: 75733229-75733229 on Build GRCh38
rs749808563 Chr.1: 75733023-75733023 on Build GRCh38
rs762165843 Chr.1: 75732779-75732779 on Build GRCh38
rs774132223 Chr.1: 75733170-75733170 on Build GRCh38
rs930392145 Chr.1: 75732839-75732839 on Build GRCh38
rs750289849 Chr.1: 75733178-75733178 on Build GRCh38
rs748467366 Chr.1: 75733049-75733049 on Build GRCh38
rs763998240 Chr.1: 75732802-75732805 on Build GRCh38
rs546640550 Chr.1: 75732892-75732892 on Build GRCh38
rs763858826 Chr.1: 75732933-75732933 on Build GRCh38
rs756863325 Chr.1: 75732939-75732939 on Build GRCh38
rs778722578 Chr.1: 75732791-75732791 on Build GRCh38
rs773331172 Chr.1: 75733055-75733055 on Build GRCh38
rs755476467 Chr.1: 75733145-75733145 on Build GRCh38
rs773278772 Chr.1: 75732900-75732900 on Build GRCh38
rs2275378 Chr.1: 75732751-75732751 on Build GRCh38
rs778243064 Chr.1: 75733150-75733150 on Build GRCh38
rs11549025 Chr.1: 75732905-75732905 on Build GRCh38
rs886042053 Chr.1: 75732866-75732866 on Build GRCh38
rs769344842 Chr.1: 75733028-75733028 on Build GRCh38
rs752612962 Chr.1: 75732913-75732913 on Build GRCh38
rs998347648 Chr.1: 75733064-75733064 on Build GRCh38
rs374358376 Chr.1: 75732739-75732739 on Build GRCh38
rs887693711 Chr.1: 75733223-75733223 on Build GRCh38
rs780737278 Chr.1: 75732963-75732963 on Build GRCh38
rs1057516480 Chr.1: 75732860-75732860 on Build GRCh38
rs755555817 Chr.1: 75733016-75733016 on Build GRCh38
rs786204566 Chr.1: 75732880-75732881 on Build GRCh38
rs768529713 Chr.1: 75733165-75733165 on Build GRCh38
rs528788578 Chr.1: 75732746-75732746 on Build GRCh38
rs186349479 Chr.1: 75733015-75733015 on Build GRCh38
rs750012444 Chr.1: 75733112-75733112 on Build GRCh38
rs398123073 Chr.1: 75732743-75732743 on Build GRCh38
rs3818855 Chr.1: 75733100-75733100 on Build GRCh38
rs761849768 Chr.1: 75733171-75733171 on Build GRCh38
rs56294347 Chr.1: 75733095-75733095 on Build GRCh38
rs886042055 Chr.1: 75732922-75732922 on Build GRCh38
rs774800083 Chr.1: 75733035-75733035 on Build GRCh38
rs764637486 Chr.1: 75732781-75732781 on Build GRCh38
rs765175601 Chr.1: 75732911-75732911 on Build GRCh38
rs772298613 Chr.1: 75733052-75733052 on Build GRCh38
rs766173642 Chr.1: 75732898-75732899 on Build GRCh38
rs751312329 Chr.1: 75733098-75733098 on Build GRCh38
rs190995228 Chr.1: 75733163-75733163 on Build GRCh38
rs780351735 Chr.1: 75733176-75733176 on Build GRCh38
rs753412421 Chr.1: 75733137-75733137 on Build GRCh38
rs398123075 Chr.1: 75732889-75732889 on Build GRCh38
rs752451629 Chr.1: 75732792-75732792 on Build GRCh38
rs571581685 Chr.1: 75732773-75732773 on Build GRCh38
rs756915078 Chr.1: 75732810-75732810 on Build GRCh38
rs1004892543 Chr.1: 75732957-75732957 on Build GRCh38
rs371194356 Chr.1: 75732901-75732901 on Build GRCh38
rs1057516428 Chr.1: 75732906-75732907 on Build GRCh38
rs766418868 Chr.1: 75733067-75733067 on Build GRCh38
rs764360637 Chr.1: 75732760-75732760 on Build GRCh38
rs760188948 Chr.1: 75733190-75733190 on Build GRCh38
rs751365301 Chr.1: 75732938-75732938 on Build GRCh38
rs777784664 Chr.1: 75732978-75732978 on Build GRCh38
rs368385016 Chr.1: 75732920-75732920 on Build GRCh38
rs535952185 Chr.1: 75733146-75733146 on Build GRCh38
rs758376450 Chr.1: 75732925-75732925 on Build GRCh38
rs776162089 Chr.1: 75732863-75732863 on Build GRCh38
rs767211797 Chr.1: 75733173-75733173 on Build GRCh38
rs778055470 Chr.1: 75732788-75732788 on Build GRCh38
rs201221339 Chr.1: 75732890-75732890 on Build GRCh38
rs536299764 Chr.1: 75733080-75733080 on Build GRCh38
rs761009839 Chr.1: 75733060-75733060 on Build GRCh38
rs771150762 Chr.1: 75733158-75733158 on Build GRCh38
rs762114560 Chr.1: 75732886-75732886 on Build GRCh38
rs41291506 Chr.1: 75732787-75732787 on Build GRCh38
rs367640956 Chr.1: 75733099-75733099 on Build GRCh38

View Full Product Details