Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75931749-75932246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB17
Entrez Gene ID
127247
Gene Name
ankyrin repeat and SOCS box containing 17
Gene Aliases
Asb-17
UniGene
Hs.125423
Gene Chromosome Location
Chr.1: 75918873-75937625 on Build GRCh38
Transcripts
NM_080868.2 NR_026546.1 XM_006710345.3 XM_006710346.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6127608 c.365T>C p.V122A Chr.1: 75931927-75931927 on Build GRCh38
2242106 c.204A>T p.T68T Chr.1: 75932088-75932088 on Build GRCh38
4522382 c.115G>A p.G39R Chr.1: 75932177-75932177 on Build GRCh38
2242102 c.353C>A p.T118K Chr.1: 75931939-75931939 on Build GRCh38
6127606 c.111G>A p.Q37Q Chr.1: 75932181-75932181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766357033 Chr.1: 75932200-75932200 on Build GRCh38
rs763022858 Chr.1: 75932097-75932097 on Build GRCh38
rs770024127 Chr.1: 75932087-75932087 on Build GRCh38
rs140553025 Chr.1: 75931852-75931852 on Build GRCh38
rs745671695 Chr.1: 75931897-75931897 on Build GRCh38
rs762649708 Chr.1: 75931841-75931841 on Build GRCh38
rs369867386 Chr.1: 75931958-75931958 on Build GRCh38
rs553833321 Chr.1: 75932048-75932048 on Build GRCh38
rs973280593 Chr.1: 75931771-75931771 on Build GRCh38
rs867745250 Chr.1: 75931984-75931984 on Build GRCh38
rs567753897 Chr.1: 75931890-75931890 on Build GRCh38
rs998788362 Chr.1: 75931895-75931895 on Build GRCh38
rs773571008 Chr.1: 75931956-75931956 on Build GRCh38
rs750164857 Chr.1: 75931998-75931998 on Build GRCh38
rs375915435 Chr.1: 75932170-75932170 on Build GRCh38
rs767340808 Chr.1: 75932099-75932099 on Build GRCh38
rs766620765 Chr.1: 75931969-75931969 on Build GRCh38
rs1796814 Chr.1: 75931990-75931990 on Build GRCh38
rs192225086 Chr.1: 75931865-75931865 on Build GRCh38
rs776881601 Chr.1: 75932065-75932065 on Build GRCh38
rs199681630 Chr.1: 75932163-75932163 on Build GRCh38
rs778322729 Chr.1: 75932158-75932158 on Build GRCh38
rs533619388 Chr.1: 75931914-75931914 on Build GRCh38
rs201319157 Chr.1: 75932165-75932165 on Build GRCh38
rs759590791 Chr.1: 75931972-75931972 on Build GRCh38
rs200235376 Chr.1: 75932117-75932117 on Build GRCh38
rs142741766 Chr.1: 75931896-75931896 on Build GRCh38
rs188825833 Chr.1: 75932035-75932035 on Build GRCh38
rs760485730 Chr.1: 75932198-75932198 on Build GRCh38
rs577407367 Chr.1: 75931809-75931809 on Build GRCh38
rs768665485 Chr.1: 75932182-75932182 on Build GRCh38
rs367592336 Chr.1: 75931857-75931857 on Build GRCh38
rs139449858 Chr.1: 75931920-75931920 on Build GRCh38
rs568808058 Chr.1: 75931970-75931970 on Build GRCh38
rs752253768 Chr.1: 75932142-75932142 on Build GRCh38
rs562878541 Chr.1: 75931816-75931816 on Build GRCh38
rs763072264 Chr.1: 75932177-75932177 on Build GRCh38
rs779576040 Chr.1: 75931911-75931911 on Build GRCh38
rs544412374 Chr.1: 75931813-75931813 on Build GRCh38
rs201311331 Chr.1: 75931976-75931976 on Build GRCh38
rs772732746 Chr.1: 75931945-75931945 on Build GRCh38
rs764007757 Chr.1: 75931877-75931877 on Build GRCh38
rs753891186 Chr.1: 75931871-75931871 on Build GRCh38
rs767367355 Chr.1: 75931994-75931994 on Build GRCh38
rs746440539 Chr.1: 75932095-75932095 on Build GRCh38
rs755926016 Chr.1: 75932001-75932001 on Build GRCh38
rs754504070 Chr.1: 75932033-75932033 on Build GRCh38
rs755732849 Chr.1: 75931908-75931908 on Build GRCh38
rs757905959 Chr.1: 75932153-75932153 on Build GRCh38
rs559179957 Chr.1: 75932107-75932107 on Build GRCh38
rs778717349 Chr.1: 75932132-75932132 on Build GRCh38
rs888895669 Chr.1: 75931840-75931840 on Build GRCh38
rs754622969 Chr.1: 75932121-75932121 on Build GRCh38
rs771251338 Chr.1: 75932054-75932054 on Build GRCh38
rs753624936 Chr.1: 75932220-75932220 on Build GRCh38
rs534539433 Chr.1: 75932100-75932100 on Build GRCh38
rs757015802 Chr.1: 75931892-75931892 on Build GRCh38
rs779137911 Chr.1: 75932073-75932073 on Build GRCh38
rs372726081 Chr.1: 75932167-75932167 on Build GRCh38
rs17097710 Chr.1: 75931832-75931832 on Build GRCh38
rs187995565 Chr.1: 75931842-75931842 on Build GRCh38
rs146811000 Chr.1: 75931830-75931830 on Build GRCh38
rs755119576 Chr.1: 75931872-75931872 on Build GRCh38
rs182689417 Chr.1: 75932034-75932034 on Build GRCh38
rs898825862 Chr.1: 75931829-75931829 on Build GRCh38
rs574736873 Chr.1: 75932169-75932169 on Build GRCh38
rs759715484 Chr.1: 75932082-75932082 on Build GRCh38
rs775490158 Chr.1: 75932090-75932090 on Build GRCh38
rs773165747 Chr.1: 75932192-75932192 on Build GRCh38
rs202233278 Chr.1: 75931928-75931928 on Build GRCh38
rs772432111 Chr.1: 75932102-75932103 on Build GRCh38
rs753713759 Chr.1: 75932120-75932120 on Build GRCh38
rs569365958 Chr.1: 75932175-75932175 on Build GRCh38
rs148576874 Chr.1: 75932053-75932053 on Build GRCh38
rs11811988 Chr.1: 75932031-75932031 on Build GRCh38
rs752805168 Chr.1: 75931983-75931983 on Build GRCh38
rs1050062567 Chr.1: 75932204-75932204 on Build GRCh38
rs761821157 Chr.1: 75932046-75932046 on Build GRCh38
rs779741486 Chr.1: 75932012-75932012 on Build GRCh38
rs754012654 Chr.1: 75932203-75932203 on Build GRCh38
rs772565854 Chr.1: 75932045-75932045 on Build GRCh38
rs149865221 Chr.1: 75931991-75931991 on Build GRCh38

View Full Product Details