Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3672043-3672537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
UniGene
Hs.173716
Gene Chromosome Location
Chr.20: 3667973-3682131 on Build GRCh38
Transcripts
NM_001282447.2 NM_025220.4 NM_153202.3 XM_005260843.1 XM_006723639.1 XM_006723640.1 XM_006723644.2 XM_011529366.1 XM_011529367.1 XM_011529368.1 XM_011529370.1 XM_011529371.1 XM_011529373.1 XM_017028080.1 XM_017028081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4423929 c.1552G>A p.A518T Chr.20: 3672179-3672179 on Build GRCh38
4987835 c.1543T>C p.W515R Chr.20: 3672188-3672188 on Build GRCh38
5857813 c.1490delC p.P497fs*49 Chr.20: 3672241-3672241 on Build GRCh38
6093198 c.1541G>C p.C514S Chr.20: 3672190-3672190 on Build GRCh38
2760773 c.1459T>C p.F487L Chr.20: 3672272-3672272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055064428 Chr.20: 3672403-3672403 on Build GRCh38
rs902593198 Chr.20: 3672472-3672472 on Build GRCh38
rs534375620 Chr.20: 3672480-3672480 on Build GRCh38
rs558390737 Chr.20: 3672106-3672106 on Build GRCh38
rs745403787 Chr.20: 3672222-3672222 on Build GRCh38
rs906447939 Chr.20: 3672158-3672158 on Build GRCh38
rs769288537 Chr.20: 3672226-3672226 on Build GRCh38
rs756572107 Chr.20: 3672304-3672304 on Build GRCh38
rs759541350 Chr.20: 3672258-3672258 on Build GRCh38
rs534541783 Chr.20: 3672173-3672173 on Build GRCh38
rs762901156 Chr.20: 3672358-3672358 on Build GRCh38
rs927792492 Chr.20: 3672392-3672392 on Build GRCh38
rs763286689 Chr.20: 3672274-3672274 on Build GRCh38
rs894054181 Chr.20: 3672122-3672122 on Build GRCh38
rs201325299 Chr.20: 3672349-3672349 on Build GRCh38
rs56355127 Chr.20: 3672333-3672337 on Build GRCh38
rs186219489 Chr.20: 3672334-3672334 on Build GRCh38
rs762168627 Chr.20: 3672372-3672372 on Build GRCh38
rs55880335 Chr.20: 3672500-3672500 on Build GRCh38
rs778170022 Chr.20: 3672207-3672207 on Build GRCh38
rs758546289 Chr.20: 3672203-3672203 on Build GRCh38
rs539488105 Chr.20: 3672097-3672097 on Build GRCh38
rs6037648 Chr.20: 3672342-3672342 on Build GRCh38
rs376654723 Chr.20: 3672350-3672350 on Build GRCh38
rs578045242 Chr.20: 3672246-3672246 on Build GRCh38
rs373056338 Chr.20: 3672237-3672237 on Build GRCh38
rs3918395 Chr.20: 3672502-3672502 on Build GRCh38
rs539161162 Chr.20: 3672382-3672382 on Build GRCh38
rs751952346 Chr.20: 3672282-3672282 on Build GRCh38
rs781634258 Chr.20: 3672221-3672221 on Build GRCh38
rs750879408 Chr.20: 3672393-3672393 on Build GRCh38
rs767934313 Chr.20: 3672374-3672374 on Build GRCh38
rs765426516 Chr.20: 3672260-3672260 on Build GRCh38
rs758111889 Chr.20: 3672509-3672509 on Build GRCh38
rs1010267423 Chr.20: 3672147-3672147 on Build GRCh38
rs766629277 Chr.20: 3672108-3672108 on Build GRCh38
rs868398896 Chr.20: 3672327-3672327 on Build GRCh38
rs939213717 Chr.20: 3672398-3672398 on Build GRCh38
rs556123031 Chr.20: 3672486-3672486 on Build GRCh38
rs748487562 Chr.20: 3672121-3672121 on Build GRCh38
rs545054862 Chr.20: 3672301-3672301 on Build GRCh38
rs762134030 Chr.20: 3672243-3672243 on Build GRCh38
rs781672715 Chr.20: 3672289-3672289 on Build GRCh38
rs773229803 Chr.20: 3672253-3672253 on Build GRCh38
rs41525645 Chr.20: 3672234-3672234 on Build GRCh38
rs764270138 Chr.20: 3672279-3672279 on Build GRCh38
rs615436 Chr.20: 3672188-3672188 on Build GRCh38
rs1009781531 Chr.20: 3672405-3672405 on Build GRCh38
rs929143262 Chr.20: 3672109-3672109 on Build GRCh38
rs949937807 Chr.20: 3672345-3672345 on Build GRCh38
rs767119399 Chr.20: 3672358-3672359 on Build GRCh38
rs556083898 Chr.20: 3672174-3672174 on Build GRCh38
rs779669835 Chr.20: 3672305-3672305 on Build GRCh38
rs973044929 Chr.20: 3672223-3672223 on Build GRCh38
rs546525095 Chr.20: 3672319-3672319 on Build GRCh38
rs755199691 Chr.20: 3672114-3672114 on Build GRCh38
rs73076686 Chr.20: 3672103-3672103 on Build GRCh38
rs563988014 Chr.20: 3672312-3672312 on Build GRCh38
rs752708082 Chr.20: 3672265-3672265 on Build GRCh38
rs568797571 Chr.20: 3672363-3672363 on Build GRCh38
rs77629002 Chr.20: 3672089-3672089 on Build GRCh38
rs776168462 Chr.20: 3672148-3672148 on Build GRCh38
rs983277774 Chr.20: 3672391-3672391 on Build GRCh38
rs757800807 Chr.20: 3672215-3672215 on Build GRCh38
rs374693928 Chr.20: 3672336-3672336 on Build GRCh38
rs778148486 Chr.20: 3672141-3672141 on Build GRCh38
rs41531745 Chr.20: 3672416-3672416 on Build GRCh38
rs1044137569 Chr.20: 3672457-3672458 on Build GRCh38
rs756654266 Chr.20: 3672399-3672399 on Build GRCh38
rs774332374 Chr.20: 3672239-3672239 on Build GRCh38
rs994599931 Chr.20: 3672516-3672516 on Build GRCh38
rs761895855 Chr.20: 3672179-3672179 on Build GRCh38
rs866094556 Chr.20: 3672388-3672388 on Build GRCh38
rs751721432 Chr.20: 3672371-3672371 on Build GRCh38
rs748720838 Chr.20: 3672308-3672308 on Build GRCh38
rs551495617 Chr.20: 3672474-3672474 on Build GRCh38
rs757567846 Chr.20: 3672283-3672283 on Build GRCh38
rs755039455 Chr.20: 3672185-3672185 on Build GRCh38
rs41395251 Chr.20: 3672477-3672477 on Build GRCh38
rs1017377022 Chr.20: 3672202-3672202 on Build GRCh38
rs1011381990 Chr.20: 3672213-3672213 on Build GRCh38
rs774280799 Chr.20: 3672178-3672178 on Build GRCh38
rs766761218 Chr.20: 3672491-3672491 on Build GRCh38
rs147260297 Chr.20: 3672321-3672321 on Build GRCh38
rs772161942 Chr.20: 3672249-3672249 on Build GRCh38
rs200809635 Chr.20: 3672127-3672127 on Build GRCh38
rs368007049 Chr.20: 3672116-3672116 on Build GRCh38
rs763879659 Chr.20: 3672177-3672177 on Build GRCh38
rs769362525 Chr.20: 3672169-3672169 on Build GRCh38
rs751899277 Chr.20: 3672214-3672214 on Build GRCh38
rs571197585 Chr.20: 3672361-3672361 on Build GRCh38
rs143884879 Chr.20: 3672332-3672336 on Build GRCh38
rs747583666 Chr.20: 3672146-3672146 on Build GRCh38
rs773138145 Chr.20: 3672332-3672332 on Build GRCh38
rs528261077 Chr.20: 3672316-3672316 on Build GRCh38
rs779159532 Chr.20: 3672120-3672120 on Build GRCh38
rs370574211 Chr.20: 3672515-3672515 on Build GRCh38
rs926692856 Chr.20: 3672355-3672355 on Build GRCh38
rs377599074 Chr.20: 3672506-3672506 on Build GRCh38
rs745350807 Chr.20: 3672159-3672159 on Build GRCh38

View Full Product Details