Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3758303-3758839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf27
Entrez Gene ID
54976
Gene Name
chromosome 20 open reading frame 27
Gene Aliases
-
UniGene
Hs.274422
Gene Chromosome Location
Chr.20: 3753499-3768388 on Build GRCh38
Transcripts
NM_001039140.2 NM_001258429.1 NM_001258430.1 NR_047675.1 XM_011529266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1026706 c.122G>A p.R41H Chr.20: 3758651-3758651 on Build GRCh38
4098300 c.192G>A p.S64S Chr.20: 3758581-3758581 on Build GRCh38
5494052 c.164T>C p.V55A Chr.20: 3758609-3758609 on Build GRCh38
1026705 c.219C>T p.D73D Chr.20: 3758554-3758554 on Build GRCh38
6615591 c.121C>T p.R41C Chr.20: 3758652-3758652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113588223 Chr.20: 3758629-3758629 on Build GRCh38
rs577872015 Chr.20: 3758582-3758582 on Build GRCh38
rs368072108 Chr.20: 3758325-3758325 on Build GRCh38
rs1034782463 Chr.20: 3758379-3758379 on Build GRCh38
rs201043568 Chr.20: 3758497-3758497 on Build GRCh38
rs41279396 Chr.20: 3758645-3758645 on Build GRCh38
rs538755689 Chr.20: 3758450-3758450 on Build GRCh38
rs891107536 Chr.20: 3758439-3758439 on Build GRCh38
rs946151602 Chr.20: 3758374-3758374 on Build GRCh38
rs747234420 Chr.20: 3758657-3758657 on Build GRCh38
rs748692383 Chr.20: 3758572-3758572 on Build GRCh38
rs553966252 Chr.20: 3758705-3758705 on Build GRCh38
rs986833612 Chr.20: 3758784-3758784 on Build GRCh38
rs1052736417 Chr.20: 3758336-3758336 on Build GRCh38
rs200033063 Chr.20: 3758526-3758526 on Build GRCh38
rs540176224 Chr.20: 3758346-3758346 on Build GRCh38
rs1005073395 Chr.20: 3758373-3758373 on Build GRCh38
rs191092611 Chr.20: 3758510-3758510 on Build GRCh38
rs766255021 Chr.20: 3758680-3758681 on Build GRCh38
rs903766956 Chr.20: 3758706-3758706 on Build GRCh38
rs1006900205 Chr.20: 3758442-3758442 on Build GRCh38
rs753933743 Chr.20: 3758552-3758552 on Build GRCh38
rs757603758 Chr.20: 3758660-3758660 on Build GRCh38
rs775127054 Chr.20: 3758631-3758631 on Build GRCh38
rs757221491 Chr.20: 3758530-3758530 on Build GRCh38
rs572167160 Chr.20: 3758748-3758748 on Build GRCh38
rs752946290 Chr.20: 3758335-3758335 on Build GRCh38
rs374067825 Chr.20: 3758688-3758688 on Build GRCh38
rs987977929 Chr.20: 3758610-3758610 on Build GRCh38
rs767315991 Chr.20: 3758548-3758548 on Build GRCh38
rs746348018 Chr.20: 3758618-3758618 on Build GRCh38
rs148614096 Chr.20: 3758635-3758635 on Build GRCh38
rs747298394 Chr.20: 3758585-3758585 on Build GRCh38
rs749304056 Chr.20: 3758377-3758377 on Build GRCh38
rs867843460 Chr.20: 3758580-3758580 on Build GRCh38
rs748704717 Chr.20: 3758702-3758702 on Build GRCh38
rs142709018 Chr.20: 3758596-3758596 on Build GRCh38
rs760383629 Chr.20: 3758679-3758680 on Build GRCh38
rs956898568 Chr.20: 3758770-3758770 on Build GRCh38
rs891860065 Chr.20: 3758366-3758366 on Build GRCh38
rs768948450 Chr.20: 3758677-3758678 on Build GRCh38
rs781687163 Chr.20: 3758661-3758661 on Build GRCh38
rs566900619 Chr.20: 3758449-3758449 on Build GRCh38
rs746435942 Chr.20: 3758670-3758670 on Build GRCh38
rs562308692 Chr.20: 3758493-3758493 on Build GRCh38
rs755096843 Chr.20: 3758557-3758557 on Build GRCh38
rs377153188 Chr.20: 3758707-3758707 on Build GRCh38
rs777812096 Chr.20: 3758656-3758656 on Build GRCh38
rs142042927 Chr.20: 3758674-3758674 on Build GRCh38
rs550570969 Chr.20: 3758492-3758492 on Build GRCh38
rs762679386 Chr.20: 3758634-3758634 on Build GRCh38
rs571531973 Chr.20: 3758652-3758652 on Build GRCh38
rs762508671 Chr.20: 3758521-3758521 on Build GRCh38
rs549556115 Chr.20: 3758583-3758583 on Build GRCh38
rs755997482 Chr.20: 3758462-3758462 on Build GRCh38
rs751253961 Chr.20: 3758529-3758529 on Build GRCh38
rs116204784 Chr.20: 3758454-3758454 on Build GRCh38
rs960782656 Chr.20: 3758399-3758399 on Build GRCh38
rs529106362 Chr.20: 3758463-3758463 on Build GRCh38
rs140834500 Chr.20: 3758494-3758494 on Build GRCh38
rs771493186 Chr.20: 3758606-3758606 on Build GRCh38
rs540043004 Chr.20: 3758755-3758755 on Build GRCh38
rs756037326 Chr.20: 3758644-3758644 on Build GRCh38
rs146253705 Chr.20: 3758573-3758573 on Build GRCh38
rs776410476 Chr.20: 3758682-3758682 on Build GRCh38
rs1056591139 Chr.20: 3758666-3758666 on Build GRCh38
rs1047325462 Chr.20: 3758807-3758807 on Build GRCh38
rs1012103053 Chr.20: 3758422-3758422 on Build GRCh38
rs774720717 Chr.20: 3758678-3758679 on Build GRCh38
rs1020105272 Chr.20: 3758813-3758813 on Build GRCh38
rs567961140 Chr.20: 3758611-3758611 on Build GRCh38
rs752711158 Chr.20: 3758651-3758651 on Build GRCh38
rs1051200501 Chr.20: 3758389-3758389 on Build GRCh38
rs980436473 Chr.20: 3758562-3758562 on Build GRCh38
rs773437215 Chr.20: 3758778-3758778 on Build GRCh38
rs138016250 Chr.20: 3758581-3758581 on Build GRCh38
rs11539185 Chr.20: 3758638-3758638 on Build GRCh38
rs920160078 Chr.20: 3758627-3758627 on Build GRCh38
rs1000752640 Chr.20: 3758747-3758747 on Build GRCh38
rs1039766848 Chr.20: 3758696-3758696 on Build GRCh38
rs758656944 Chr.20: 3758348-3758348 on Build GRCh38
rs887253379 Chr.20: 3758816-3758816 on Build GRCh38

View Full Product Details