Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3800370-3800874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25B
Entrez Gene ID
994
Gene Name
cell division cycle 25B
Gene Aliases
-
UniGene
Hs.153752
Gene Chromosome Location
Chr.20: 3786772-3806121 on Build GRCh38
Transcripts
NM_001287516.1 NM_001287517.1 NM_001287518.1 NM_001287519.1 NM_001287520.1 NM_001287522.1 NM_001287524.1 NM_004358.4 NM_021872.3 NM_021873.3 NR_136335.1 NR_136336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6624181 c.332G>A p.R111Q Chr.20: 3800807-3800807 on Build GRCh38
6858085 c.348G>A p.A116A Chr.20: 3800823-3800823 on Build GRCh38
6624179 c.524G>A p.R175Q Chr.20: 3800807-3800807 on Build GRCh38
5857855 c.508C>T p.Q170* Chr.20: 3800791-3800791 on Build GRCh38
6300448 c.484G>C p.V162L Chr.20: 3800767-3800767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777091404 Chr.20: 3800706-3800707 on Build GRCh38
rs111654983 Chr.20: 3800473-3800473 on Build GRCh38
rs763680599 Chr.20: 3800453-3800453 on Build GRCh38
rs924314061 Chr.20: 3800549-3800549 on Build GRCh38
rs750060566 Chr.20: 3800471-3800471 on Build GRCh38
rs550851634 Chr.20: 3800511-3800511 on Build GRCh38
rs897796646 Chr.20: 3800630-3800630 on Build GRCh38
rs775666911 Chr.20: 3800497-3800497 on Build GRCh38
rs894619445 Chr.20: 3800407-3800407 on Build GRCh38
rs746114677 Chr.20: 3800699-3800699 on Build GRCh38
rs751526183 Chr.20: 3800840-3800840 on Build GRCh38
rs932976178 Chr.20: 3800609-3800609 on Build GRCh38
rs562467419 Chr.20: 3800527-3800527 on Build GRCh38
rs1021777274 Chr.20: 3800447-3800447 on Build GRCh38
rs549552103 Chr.20: 3800723-3800723 on Build GRCh38
rs1036451818 Chr.20: 3800604-3800604 on Build GRCh38
rs769049251 Chr.20: 3800729-3800729 on Build GRCh38
rs759162155 Chr.20: 3800825-3800825 on Build GRCh38
rs904266009 Chr.20: 3800683-3800683 on Build GRCh38
rs371350767 Chr.20: 3800773-3800773 on Build GRCh38
rs780196030 Chr.20: 3800800-3800800 on Build GRCh38
rs766222083 Chr.20: 3800514-3800514 on Build GRCh38
rs778784069 Chr.20: 3800537-3800537 on Build GRCh38
rs373068861 Chr.20: 3800795-3800795 on Build GRCh38
rs11570001 Chr.20: 3800540-3800540 on Build GRCh38
rs754772727 Chr.20: 3800531-3800531 on Build GRCh38
rs920383476 Chr.20: 3800822-3800822 on Build GRCh38
rs773393529 Chr.20: 3800821-3800821 on Build GRCh38
rs11552106 Chr.20: 3800796-3800796 on Build GRCh38
rs147172963 Chr.20: 3800481-3800481 on Build GRCh38
rs910307144 Chr.20: 3800637-3800637 on Build GRCh38
rs894752608 Chr.20: 3800670-3800670 on Build GRCh38
rs776161634 Chr.20: 3800823-3800823 on Build GRCh38
rs371528141 Chr.20: 3800482-3800482 on Build GRCh38
rs868511318 Chr.20: 3800739-3800739 on Build GRCh38
rs754035549 Chr.20: 3800851-3800851 on Build GRCh38
rs749465321 Chr.20: 3800802-3800802 on Build GRCh38
rs758361574 Chr.20: 3800789-3800789 on Build GRCh38
rs199988612 Chr.20: 3800812-3800812 on Build GRCh38
rs1057375613 Chr.20: 3800663-3800663 on Build GRCh38
rs773681532 Chr.20: 3800809-3800809 on Build GRCh38
rs762236479 Chr.20: 3800748-3800748 on Build GRCh38
rs765024374 Chr.20: 3800776-3800776 on Build GRCh38
rs938678351 Chr.20: 3800633-3800633 on Build GRCh38
rs201911534 Chr.20: 3800770-3800770 on Build GRCh38
rs766950960 Chr.20: 3800505-3800505 on Build GRCh38
rs755912697 Chr.20: 3800477-3800477 on Build GRCh38
rs771606389 Chr.20: 3800765-3800765 on Build GRCh38
rs532926530 Chr.20: 3800681-3800681 on Build GRCh38
rs536795359 Chr.20: 3800842-3800842 on Build GRCh38
rs776068972 Chr.20: 3800417-3800417 on Build GRCh38
rs148574309 Chr.20: 3800715-3800726 on Build GRCh38
rs1035953532 Chr.20: 3800402-3800402 on Build GRCh38
rs749680272 Chr.20: 3800724-3800724 on Build GRCh38
rs577638878 Chr.20: 3800831-3800831 on Build GRCh38
rs376072567 Chr.20: 3800817-3800817 on Build GRCh38
rs538158719 Chr.20: 3800774-3800774 on Build GRCh38
rs538614130 Chr.20: 3800839-3800839 on Build GRCh38
rs770312291 Chr.20: 3800496-3800496 on Build GRCh38
rs571020625 Chr.20: 3800781-3800781 on Build GRCh38
rs375465916 Chr.20: 3800767-3800767 on Build GRCh38
rs772664176 Chr.20: 3800766-3800766 on Build GRCh38
rs968527639 Chr.20: 3800472-3800472 on Build GRCh38
rs1011064003 Chr.20: 3800751-3800751 on Build GRCh38
rs111526979 Chr.20: 3800686-3800686 on Build GRCh38
rs984352715 Chr.20: 3800737-3800737 on Build GRCh38
rs770189227 Chr.20: 3800702-3800702 on Build GRCh38
rs1052778969 Chr.20: 3800836-3800836 on Build GRCh38
rs373954063 Chr.20: 3800740-3800740 on Build GRCh38
rs144359318 Chr.20: 3800807-3800807 on Build GRCh38
rs140433155 Chr.20: 3800493-3800493 on Build GRCh38
rs774913855 Chr.20: 3800499-3800499 on Build GRCh38
rs756726870 Chr.20: 3800462-3800462 on Build GRCh38
rs765684296 Chr.20: 3800412-3800412 on Build GRCh38
rs201436986 Chr.20: 3800498-3800498 on Build GRCh38
rs372135675 Chr.20: 3800585-3800585 on Build GRCh38
rs61755298 Chr.20: 3800489-3800489 on Build GRCh38
rs368675680 Chr.20: 3800492-3800492 on Build GRCh38
rs779300536 Chr.20: 3800806-3800806 on Build GRCh38
rs11570002 Chr.20: 3800687-3800687 on Build GRCh38
rs756159379 Chr.20: 3800841-3800841 on Build GRCh38
rs146714806 Chr.20: 3800409-3800409 on Build GRCh38
rs770463028 Chr.20: 3800818-3800818 on Build GRCh38
rs1013491251 Chr.20: 3800677-3800677 on Build GRCh38
rs757426655 Chr.20: 3800546-3800546 on Build GRCh38
rs367573455 Chr.20: 3800803-3800803 on Build GRCh38
rs752588927 Chr.20: 3800835-3800835 on Build GRCh38
rs1037966855 Chr.20: 3800673-3800673 on Build GRCh38
rs1003088366 Chr.20: 3800401-3800401 on Build GRCh38
rs534156738 Chr.20: 3800692-3800692 on Build GRCh38
rs868003275 Chr.20: 3800445-3800445 on Build GRCh38
rs763329080 Chr.20: 3800448-3800448 on Build GRCh38
rs780110600 Chr.20: 3800846-3800846 on Build GRCh38
rs41279400 Chr.20: 3800691-3800691 on Build GRCh38
rs11570003 Chr.20: 3800722-3800733 on Build GRCh38
rs752711886 Chr.20: 3800786-3800786 on Build GRCh38
rs767220701 Chr.20: 3800463-3800463 on Build GRCh38
rs761367060 Chr.20: 3800501-3800501 on Build GRCh38
rs751078014 Chr.20: 3800459-3800459 on Build GRCh38
rs762808228 Chr.20: 3800838-3800838 on Build GRCh38
rs771131604 Chr.20: 3800491-3800491 on Build GRCh38
rs766161444 Chr.20: 3800519-3800520 on Build GRCh38

View Full Product Details