Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6075117-6075651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

SNP Detail

SNP ID Chromosome Location
rs1019189976 Chr.20: 6075510-6075510 on Build GRCh38
rs929102911 Chr.20: 6075609-6075609 on Build GRCh38
rs6117066 Chr.20: 6075157-6075157 on Build GRCh38
rs555733355 Chr.20: 6075251-6075251 on Build GRCh38
rs956236689 Chr.20: 6075403-6075403 on Build GRCh38
rs914633281 Chr.20: 6075482-6075482 on Build GRCh38
rs553342112 Chr.20: 6075460-6075460 on Build GRCh38
rs932738873 Chr.20: 6075486-6075486 on Build GRCh38
rs1040321890 Chr.20: 6075546-6075546 on Build GRCh38
rs1027378742 Chr.20: 6075197-6075197 on Build GRCh38
rs984867286 Chr.20: 6075562-6075562 on Build GRCh38
rs1056269122 Chr.20: 6075606-6075606 on Build GRCh38
rs887546808 Chr.20: 6075238-6075238 on Build GRCh38
rs930774750 Chr.20: 6075272-6075272 on Build GRCh38
rs1056352549 Chr.20: 6075304-6075304 on Build GRCh38
rs993236100 Chr.20: 6075178-6075178 on Build GRCh38
rs182415956 Chr.20: 6075247-6075247 on Build GRCh38
rs397756084 Chr.20: 6075396-6075397 on Build GRCh38
rs869285484 Chr.20: 6075397-6075398 on Build GRCh38
rs542885232 Chr.20: 6075532-6075532 on Build GRCh38
rs896433786 Chr.20: 6075320-6075320 on Build GRCh38
rs1049626231 Chr.20: 6075274-6075274 on Build GRCh38
rs759236996 Chr.20: 6075478-6075478 on Build GRCh38
rs757326093 Chr.20: 6075357-6075369 on Build GRCh38
rs560958476 Chr.20: 6075613-6075613 on Build GRCh38
rs6139908 Chr.20: 6075489-6075489 on Build GRCh38
rs940520040 Chr.20: 6075594-6075594 on Build GRCh38
rs1011868040 Chr.20: 6075330-6075330 on Build GRCh38
rs146142837 Chr.20: 6075288-6075288 on Build GRCh38
rs1000582625 Chr.20: 6075446-6075446 on Build GRCh38
rs577291407 Chr.20: 6075399-6075399 on Build GRCh38
rs952142365 Chr.20: 6075539-6075539 on Build GRCh38
rs1031188046 Chr.20: 6075454-6075454 on Build GRCh38
rs572055568 Chr.20: 6075519-6075519 on Build GRCh38
rs555458571 Chr.20: 6075499-6075499 on Build GRCh38
rs907659820 Chr.20: 6075287-6075287 on Build GRCh38
rs139481302 Chr.20: 6075332-6075332 on Build GRCh38
rs576200282 Chr.20: 6075617-6075617 on Build GRCh38
rs944681018 Chr.20: 6075240-6075240 on Build GRCh38
rs544608353 Chr.20: 6075400-6075400 on Build GRCh38
rs567183901 Chr.20: 6075183-6075183 on Build GRCh38
rs537579176 Chr.20: 6075289-6075289 on Build GRCh38
rs897760931 Chr.20: 6075581-6075581 on Build GRCh38
rs912596820 Chr.20: 6075520-6075520 on Build GRCh38
rs956420976 Chr.20: 6075148-6075148 on Build GRCh38

View Full Product Details