Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6094547-6095053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684342 c.1132T>G p.L378V Chr.20: 6094946-6094946 on Build GRCh38
1028830 c.1122T>C p.D374D Chr.20: 6094956-6094956 on Build GRCh38
5423639 c.1127T>G p.L376R Chr.20: 6094951-6094951 on Build GRCh38
1028831 c.1122T>C p.D374D Chr.20: 6094956-6094956 on Build GRCh38
4684343 c.1132T>G p.L378V Chr.20: 6094946-6094946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535496713 Chr.20: 6094752-6094752 on Build GRCh38
rs998180716 Chr.20: 6094601-6094601 on Build GRCh38
rs868254492 Chr.20: 6094958-6094958 on Build GRCh38
rs913387320 Chr.20: 6094802-6094802 on Build GRCh38
rs565288503 Chr.20: 6095001-6095004 on Build GRCh38
rs549926185 Chr.20: 6094641-6094641 on Build GRCh38
rs998581193 Chr.20: 6094875-6094875 on Build GRCh38
rs143153145 Chr.20: 6094926-6094926 on Build GRCh38
rs953591503 Chr.20: 6094713-6094713 on Build GRCh38
rs769449192 Chr.20: 6095004-6095004 on Build GRCh38
rs769704237 Chr.20: 6094782-6094782 on Build GRCh38
rs750445504 Chr.20: 6094923-6094923 on Build GRCh38
rs112981254 Chr.20: 6094678-6094678 on Build GRCh38
rs904917774 Chr.20: 6094841-6094841 on Build GRCh38
rs759352186 Chr.20: 6094896-6094896 on Build GRCh38
rs796855469 Chr.20: 6094806-6094806 on Build GRCh38
rs749810209 Chr.20: 6094977-6094977 on Build GRCh38
rs139486209 Chr.20: 6094743-6094743 on Build GRCh38
rs2144938 Chr.20: 6094863-6094863 on Build GRCh38
rs780491245 Chr.20: 6094943-6094943 on Build GRCh38
rs533333661 Chr.20: 6094673-6094673 on Build GRCh38
rs1014738337 Chr.20: 6094693-6094693 on Build GRCh38
rs202227887 Chr.20: 6095029-6095029 on Build GRCh38
rs755272944 Chr.20: 6094982-6094982 on Build GRCh38
rs887127415 Chr.20: 6094985-6094985 on Build GRCh38
rs936206744 Chr.20: 6094823-6094823 on Build GRCh38
rs1030645655 Chr.20: 6094615-6094615 on Build GRCh38
rs538690401 Chr.20: 6094820-6094820 on Build GRCh38
rs909060944 Chr.20: 6094744-6094744 on Build GRCh38
rs765133175 Chr.20: 6094900-6094900 on Build GRCh38
rs779144445 Chr.20: 6095019-6095019 on Build GRCh38
rs751736473 Chr.20: 6094911-6094911 on Build GRCh38
rs904664284 Chr.20: 6094821-6094821 on Build GRCh38
rs777442299 Chr.20: 6094659-6094659 on Build GRCh38
rs992532382 Chr.20: 6094789-6094789 on Build GRCh38
rs753855187 Chr.20: 6094892-6094892 on Build GRCh38
rs566063621 Chr.20: 6094920-6094920 on Build GRCh38
rs781532839 Chr.20: 6094922-6094922 on Build GRCh38
rs770558247 Chr.20: 6094790-6094790 on Build GRCh38
rs370788314 Chr.20: 6095020-6095020 on Build GRCh38
rs899950974 Chr.20: 6094874-6094874 on Build GRCh38
rs766385147 Chr.20: 6094891-6094891 on Build GRCh38
rs869312727 Chr.20: 6094937-6094937 on Build GRCh38
rs548118494 Chr.20: 6094602-6094602 on Build GRCh38
rs550252614 Chr.20: 6094788-6094788 on Build GRCh38
rs963212431 Chr.20: 6094718-6094718 on Build GRCh38
rs2232067 Chr.20: 6094810-6094810 on Build GRCh38
rs186734728 Chr.20: 6095024-6095024 on Build GRCh38

View Full Product Details