Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6768009-6768501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2
Entrez Gene ID
650
Gene Name
bone morphogenetic protein 2
Gene Aliases
BDA2, BMP2A
UniGene
Hs.73853
Gene Chromosome Location
Chr.20: 6767700-6780280 on Build GRCh38
Transcripts
NM_001200.3 XM_011529323.2

SNP Detail

SNP ID Chromosome Location
rs550120877 Chr.20: 6768372-6768372 on Build GRCh38
rs546620110 Chr.20: 6768192-6768192 on Build GRCh38
rs951349513 Chr.20: 6768218-6768218 on Build GRCh38
rs535209432 Chr.20: 6768426-6768426 on Build GRCh38
rs553625436 Chr.20: 6768281-6768282 on Build GRCh38
rs1046219512 Chr.20: 6768427-6768427 on Build GRCh38
rs1057437274 Chr.20: 6768366-6768366 on Build GRCh38
rs909671165 Chr.20: 6768265-6768265 on Build GRCh38
rs550317794 Chr.20: 6768083-6768083 on Build GRCh38
rs1039931150 Chr.20: 6768390-6768390 on Build GRCh38
rs937340084 Chr.20: 6768050-6768050 on Build GRCh38
rs568626550 Chr.20: 6768380-6768380 on Build GRCh38
rs906397234 Chr.20: 6768221-6768221 on Build GRCh38
rs983091938 Chr.20: 6768143-6768143 on Build GRCh38
rs1057093580 Chr.20: 6768076-6768076 on Build GRCh38
rs1023929551 Chr.20: 6768150-6768150 on Build GRCh38
rs982730414 Chr.20: 6768259-6768259 on Build GRCh38
rs980611565 Chr.20: 6768312-6768312 on Build GRCh38
rs1037824978 Chr.20: 6768161-6768161 on Build GRCh38
rs976907321 Chr.20: 6768084-6768084 on Build GRCh38
rs1030940741 Chr.20: 6768249-6768249 on Build GRCh38
rs894913430 Chr.20: 6768178-6768178 on Build GRCh38
rs567557768 Chr.20: 6768370-6768370 on Build GRCh38
rs141364472 Chr.20: 6768210-6768210 on Build GRCh38
rs951084983 Chr.20: 6768416-6768416 on Build GRCh38
rs1042255722 Chr.20: 6768105-6768105 on Build GRCh38
rs964100362 Chr.20: 6768321-6768321 on Build GRCh38
rs958103709 Chr.20: 6768028-6768028 on Build GRCh38
rs2746245 Chr.20: 6768395-6768395 on Build GRCh38
rs927371742 Chr.20: 6768316-6768316 on Build GRCh38
rs1016849542 Chr.20: 6768441-6768441 on Build GRCh38
rs941732341 Chr.20: 6768153-6768153 on Build GRCh38
rs904703752 Chr.20: 6768124-6768124 on Build GRCh38
rs997951816 Chr.20: 6768398-6768398 on Build GRCh38
rs964152994 Chr.20: 6768064-6768064 on Build GRCh38
rs999913483 Chr.20: 6768142-6768142 on Build GRCh38
rs899572269 Chr.20: 6768396-6768396 on Build GRCh38
rs1004496234 Chr.20: 6768423-6768423 on Build GRCh38
rs1010920475 Chr.20: 6768300-6768300 on Build GRCh38
rs6054510 Chr.20: 6768269-6768269 on Build GRCh38
rs999846085 Chr.20: 6768223-6768223 on Build GRCh38
rs962520466 Chr.20: 6768289-6768289 on Build GRCh38
rs1018389484 Chr.20: 6768056-6768056 on Build GRCh38
rs904654197 Chr.20: 6768376-6768376 on Build GRCh38
rs553746369 Chr.20: 6768473-6768473 on Build GRCh38
rs963829188 Chr.20: 6768187-6768187 on Build GRCh38
rs535084968 Chr.20: 6768371-6768371 on Build GRCh38
rs906346736 Chr.20: 6768436-6768436 on Build GRCh38
rs577398067 Chr.20: 6768410-6768410 on Build GRCh38
rs531688342 Chr.20: 6768149-6768149 on Build GRCh38
rs927257212 Chr.20: 6768029-6768029 on Build GRCh38
rs539132309 Chr.20: 6768385-6768385 on Build GRCh38
rs937454625 Chr.20: 6768359-6768359 on Build GRCh38
rs763738426 Chr.20: 6768402-6768402 on Build GRCh38
rs983424099 Chr.20: 6768375-6768375 on Build GRCh38
rs997845497 Chr.20: 6768199-6768199 on Build GRCh38
rs115405923 Chr.20: 6768093-6768093 on Build GRCh38
rs557811126 Chr.20: 6768409-6768409 on Build GRCh38
rs989953179 Chr.20: 6768051-6768051 on Build GRCh38
rs1010512879 Chr.20: 6768099-6768099 on Build GRCh38
rs916298436 Chr.20: 6768418-6768418 on Build GRCh38

View Full Product Details