Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6768789-6769294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2
Entrez Gene ID
650
Gene Name
bone morphogenetic protein 2
Gene Aliases
BDA2, BMP2A
UniGene
Hs.73853
Gene Chromosome Location
Chr.20: 6767700-6780280 on Build GRCh38
Transcripts
NM_001200.3 XM_011529323.2

SNP Detail

SNP ID Chromosome Location
rs896434684 Chr.20: 6769026-6769026 on Build GRCh38
rs981791909 Chr.20: 6769085-6769085 on Build GRCh38
rs946648804 Chr.20: 6768880-6768880 on Build GRCh38
rs546343265 Chr.20: 6769140-6769140 on Build GRCh38
rs529118444 Chr.20: 6769251-6769251 on Build GRCh38
rs939789149 Chr.20: 6768922-6768922 on Build GRCh38
rs1023250928 Chr.20: 6769078-6769078 on Build GRCh38
rs1002042069 Chr.20: 6768957-6768957 on Build GRCh38
rs975668124 Chr.20: 6768867-6768867 on Build GRCh38
rs921196551 Chr.20: 6769175-6769175 on Build GRCh38
rs1024392861 Chr.20: 6768830-6768830 on Build GRCh38
rs185340495 Chr.20: 6768884-6768884 on Build GRCh38
rs370216838 Chr.20: 6769217-6769217 on Build GRCh38
rs886848470 Chr.20: 6768919-6768919 on Build GRCh38
rs1032975882 Chr.20: 6769158-6769158 on Build GRCh38
rs1021237688 Chr.20: 6769030-6769030 on Build GRCh38
rs887200085 Chr.20: 6768981-6768981 on Build GRCh38
rs1052278931 Chr.20: 6768845-6768845 on Build GRCh38
rs546017272 Chr.20: 6768844-6768844 on Build GRCh38
rs1051102606 Chr.20: 6768958-6768958 on Build GRCh38
rs1013413847 Chr.20: 6769029-6769029 on Build GRCh38
rs201411076 Chr.20: 6769141-6769144 on Build GRCh38
rs967901952 Chr.20: 6768841-6768841 on Build GRCh38
rs933951603 Chr.20: 6769230-6769230 on Build GRCh38
rs927750087 Chr.20: 6768817-6768817 on Build GRCh38
rs1037976312 Chr.20: 6768930-6768930 on Build GRCh38
rs1013155806 Chr.20: 6769061-6769061 on Build GRCh38
rs235702 Chr.20: 6769024-6769024 on Build GRCh38
rs1051052765 Chr.20: 6769260-6769260 on Build GRCh38
rs925968814 Chr.20: 6768860-6768860 on Build GRCh38
rs988332838 Chr.20: 6769140-6769141 on Build GRCh38
rs765665193 Chr.20: 6769013-6769013 on Build GRCh38
rs957358799 Chr.20: 6769167-6769167 on Build GRCh38
rs956491782 Chr.20: 6769108-6769108 on Build GRCh38
rs946578333 Chr.20: 6769164-6769164 on Build GRCh38
rs189454574 Chr.20: 6768893-6768893 on Build GRCh38
rs867037244 Chr.20: 6769082-6769082 on Build GRCh38
rs934171168 Chr.20: 6768895-6768895 on Build GRCh38
rs967472754 Chr.20: 6769046-6769046 on Build GRCh38
rs936207402 Chr.20: 6768864-6768864 on Build GRCh38
rs1039637853 Chr.20: 6768899-6768899 on Build GRCh38
rs999365138 Chr.20: 6769063-6769063 on Build GRCh38
rs561657894 Chr.20: 6769174-6769174 on Build GRCh38
rs1051734275 Chr.20: 6768908-6768908 on Build GRCh38
rs963242831 Chr.20: 6769048-6769048 on Build GRCh38
rs998151596 Chr.20: 6768913-6768913 on Build GRCh38
rs927665937 Chr.20: 6769099-6769099 on Build GRCh38
rs921507337 Chr.20: 6769222-6769222 on Build GRCh38
rs898180356 Chr.20: 6768942-6768942 on Build GRCh38

View Full Product Details