Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10560509-10561003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4IP
Entrez Gene ID
128710
Gene Name
SLX4 interacting protein
Gene Aliases
C20orf94, bA204H22.1, bA254M13.1, dJ1099D15.3
UniGene
Hs.668782
Gene Chromosome Location
Chr.20: 10434453-10628138 on Build GRCh38
Transcripts
NM_001009608.2 XM_017027647.1 XM_017027648.1 XM_017027649.1 XM_017027650.1 XM_017027651.1 XM_017027652.1 XM_017027653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4953238 c.238G>C p.G80R Chr.20: 10560820-10560820 on Build GRCh38
2702620 c.185A>G p.Q62R Chr.20: 10560767-10560767 on Build GRCh38
4096612 c.188A>G p.H63R Chr.20: 10560770-10560770 on Build GRCh38
1024395 c.224C>T p.P75L Chr.20: 10560806-10560806 on Build GRCh38
1024394 c.186G>A p.Q62Q Chr.20: 10560768-10560768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012853691 Chr.20: 10560586-10560586 on Build GRCh38
rs1027300406 Chr.20: 10560642-10560642 on Build GRCh38
rs759393316 Chr.20: 10560675-10560675 on Build GRCh38
rs779288593 Chr.20: 10560762-10560762 on Build GRCh38
rs1046030156 Chr.20: 10560611-10560611 on Build GRCh38
rs192540899 Chr.20: 10560882-10560882 on Build GRCh38
rs761610386 Chr.20: 10560661-10560661 on Build GRCh38
rs373469943 Chr.20: 10560843-10560843 on Build GRCh38
rs576133690 Chr.20: 10560772-10560772 on Build GRCh38
rs200510882 Chr.20: 10560691-10560691 on Build GRCh38
rs764366699 Chr.20: 10560684-10560684 on Build GRCh38
rs757811811 Chr.20: 10560581-10560581 on Build GRCh38
rs57274459 Chr.20: 10560583-10560583 on Build GRCh38
rs766919112 Chr.20: 10560751-10560751 on Build GRCh38
rs998498331 Chr.20: 10560613-10560613 on Build GRCh38
rs370756387 Chr.20: 10560677-10560677 on Build GRCh38
rs57951964 Chr.20: 10560544-10560544 on Build GRCh38
rs745614443 Chr.20: 10560794-10560794 on Build GRCh38
rs760827148 Chr.20: 10560861-10560861 on Build GRCh38
rs144091902 Chr.20: 10560765-10560765 on Build GRCh38
rs756506975 Chr.20: 10560728-10560728 on Build GRCh38
rs534129651 Chr.20: 10560622-10560622 on Build GRCh38
rs957154290 Chr.20: 10560738-10560738 on Build GRCh38
rs187348905 Chr.20: 10560601-10560601 on Build GRCh38
rs530074798 Chr.20: 10560543-10560543 on Build GRCh38
rs768363561 Chr.20: 10560717-10560721 on Build GRCh38
rs146530916 Chr.20: 10560768-10560768 on Build GRCh38
rs762118860 Chr.20: 10560823-10560823 on Build GRCh38
rs139067448 Chr.20: 10560894-10560894 on Build GRCh38
rs748871512 Chr.20: 10560650-10560650 on Build GRCh38
rs1016137923 Chr.20: 10560568-10560568 on Build GRCh38
rs758105551 Chr.20: 10560766-10560766 on Build GRCh38
rs946815292 Chr.20: 10560916-10560916 on Build GRCh38
rs986412493 Chr.20: 10560883-10560883 on Build GRCh38
rs183264601 Chr.20: 10560891-10560891 on Build GRCh38
rs767689042 Chr.20: 10560719-10560719 on Build GRCh38
rs907103903 Chr.20: 10560612-10560612 on Build GRCh38
rs780563280 Chr.20: 10560760-10560760 on Build GRCh38
rs755366780 Chr.20: 10560761-10560761 on Build GRCh38
rs766445670 Chr.20: 10560749-10560749 on Build GRCh38
rs751117152 Chr.20: 10560587-10560587 on Build GRCh38
rs774684260 Chr.20: 10560864-10560864 on Build GRCh38
rs200047011 Chr.20: 10560726-10560726 on Build GRCh38
rs770717675 Chr.20: 10560773-10560773 on Build GRCh38
rs891342995 Chr.20: 10560561-10560561 on Build GRCh38
rs776223316 Chr.20: 10560649-10560649 on Build GRCh38
rs370701195 Chr.20: 10560841-10560841 on Build GRCh38
rs899897195 Chr.20: 10560921-10560921 on Build GRCh38
rs769684364 Chr.20: 10560795-10560795 on Build GRCh38
rs772424810 Chr.20: 10560836-10560836 on Build GRCh38
rs766755120 Chr.20: 10560870-10560870 on Build GRCh38
rs762169764 Chr.20: 10560713-10560713 on Build GRCh38
rs953649559 Chr.20: 10560833-10560833 on Build GRCh38
rs1004391567 Chr.20: 10560564-10560564 on Build GRCh38
rs775473989 Chr.20: 10560774-10560774 on Build GRCh38
rs1043817419 Chr.20: 10560920-10560920 on Build GRCh38
rs41280284 Chr.20: 10560814-10560814 on Build GRCh38
rs756561451 Chr.20: 10560607-10560607 on Build GRCh38
rs542424354 Chr.20: 10560733-10560733 on Build GRCh38

View Full Product Details