Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 11924061-11924559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
UniGene
Hs.709366
Gene Chromosome Location
Chr.20: 11890723-11926595 on Build GRCh38
Transcripts
NM_001282550.1 NM_001282551.1 NM_001282552.2 NM_001282554.1 NM_014962.3 NM_181443.3 XM_017027728.1 XM_017027729.1 XM_017027730.1

SNP Detail

SNP ID Chromosome Location
rs946182549 Chr.20: 11924175-11924175 on Build GRCh38
rs931639097 Chr.20: 11924084-11924084 on Build GRCh38
rs368826666 Chr.20: 11924114-11924114 on Build GRCh38
rs879129276 Chr.20: 11924494-11924494 on Build GRCh38
rs951724209 Chr.20: 11924132-11924132 on Build GRCh38
rs756642148 Chr.20: 11924227-11924227 on Build GRCh38
rs887534274 Chr.20: 11924425-11924425 on Build GRCh38
rs966311582 Chr.20: 11924346-11924346 on Build GRCh38
rs975957997 Chr.20: 11924356-11924356 on Build GRCh38
rs1054173293 Chr.20: 11924099-11924099 on Build GRCh38
rs541378456 Chr.20: 11924412-11924412 on Build GRCh38
rs917777095 Chr.20: 11924222-11924222 on Build GRCh38
rs1034600474 Chr.20: 11924236-11924236 on Build GRCh38
rs959319379 Chr.20: 11924201-11924201 on Build GRCh38
rs530791651 Chr.20: 11924507-11924507 on Build GRCh38
rs377445708 Chr.20: 11924489-11924489 on Build GRCh38
rs1004735686 Chr.20: 11924322-11924322 on Build GRCh38
rs376037202 Chr.20: 11924264-11924264 on Build GRCh38
rs755300828 Chr.20: 11924501-11924501 on Build GRCh38
rs993666494 Chr.20: 11924411-11924411 on Build GRCh38
rs549992658 Chr.20: 11924516-11924516 on Build GRCh38
rs41280294 Chr.20: 11924285-11924285 on Build GRCh38
rs1041871065 Chr.20: 11924220-11924220 on Build GRCh38
rs1033590641 Chr.20: 11924173-11924173 on Build GRCh38
rs754164483 Chr.20: 11924317-11924317 on Build GRCh38
rs991933345 Chr.20: 11924217-11924217 on Build GRCh38
rs758231143 Chr.20: 11924326-11924330 on Build GRCh38
rs537258594 Chr.20: 11924180-11924180 on Build GRCh38
rs1054893228 Chr.20: 11924302-11924302 on Build GRCh38
rs910788341 Chr.20: 11924320-11924320 on Build GRCh38
rs186536576 Chr.20: 11924474-11924474 on Build GRCh38
rs182539488 Chr.20: 11924394-11924394 on Build GRCh38
rs557163357 Chr.20: 11924233-11924233 on Build GRCh38
rs1003071092 Chr.20: 11924228-11924228 on Build GRCh38
rs921854662 Chr.20: 11924390-11924390 on Build GRCh38
rs147299646 Chr.20: 11924240-11924240 on Build GRCh38
rs901976370 Chr.20: 11924408-11924408 on Build GRCh38
rs951480402 Chr.20: 11924292-11924292 on Build GRCh38
rs546102868 Chr.20: 11924278-11924278 on Build GRCh38
rs577845668 Chr.20: 11924234-11924237 on Build GRCh38
rs373983851 Chr.20: 11924485-11924485 on Build GRCh38
rs892900644 Chr.20: 11924138-11924138 on Build GRCh38
rs984868440 Chr.20: 11924151-11924151 on Build GRCh38
rs1047447409 Chr.20: 11924423-11924423 on Build GRCh38
rs1014754822 Chr.20: 11924323-11924323 on Build GRCh38

View Full Product Details