Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17531039-17531558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP1
Entrez Gene ID
631
Gene Name
beaded filament structural protein 1
Gene Aliases
CP115, CP94, CTRCT33, LIFL-H
UniGene
Hs.129702
Gene Chromosome Location
Chr.20: 17493905-17569220 on Build GRCh38
Transcripts
NM_001161705.1 NM_001195.4 NM_001278606.1 NM_001278607.1 NM_001278608.1 XM_017028005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5641141 c.261C>A p.A87A Chr.20: 17531069-17531069 on Build GRCh38
3389606 c.68G>T p.R23L Chr.20: 17531262-17531262 on Build GRCh38
4802690 c.46G>C p.E16Q Chr.20: 17531284-17531284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917119794 Chr.20: 17531203-17531203 on Build GRCh38
rs966180206 Chr.20: 17531118-17531118 on Build GRCh38
rs772525555 Chr.20: 17531200-17531200 on Build GRCh38
rs930145055 Chr.20: 17531485-17531485 on Build GRCh38
rs900178236 Chr.20: 17531366-17531366 on Build GRCh38
rs992902242 Chr.20: 17531473-17531473 on Build GRCh38
rs773615057 Chr.20: 17531272-17531272 on Build GRCh38
rs767310600 Chr.20: 17531381-17531381 on Build GRCh38
rs1047291246 Chr.20: 17531337-17531337 on Build GRCh38
rs958707239 Chr.20: 17531453-17531453 on Build GRCh38
rs1039980114 Chr.20: 17531435-17531435 on Build GRCh38
rs923937893 Chr.20: 17531192-17531192 on Build GRCh38
rs551353395 Chr.20: 17531491-17531491 on Build GRCh38
rs941574204 Chr.20: 17531521-17531521 on Build GRCh38
rs757920706 Chr.20: 17531237-17531237 on Build GRCh38
rs188623262 Chr.20: 17531432-17531432 on Build GRCh38
rs200899362 Chr.20: 17531284-17531284 on Build GRCh38
rs940369345 Chr.20: 17531426-17531426 on Build GRCh38
rs772173689 Chr.20: 17531264-17531264 on Build GRCh38
rs764863815 Chr.20: 17531309-17531309 on Build GRCh38
rs954546532 Chr.20: 17531388-17531388 on Build GRCh38
rs1049231623 Chr.20: 17531231-17531231 on Build GRCh38
rs949869524 Chr.20: 17531210-17531210 on Build GRCh38
rs757062789 Chr.20: 17531239-17531239 on Build GRCh38
rs907567384 Chr.20: 17531424-17531424 on Build GRCh38
rs761174568 Chr.20: 17531214-17531214 on Build GRCh38
rs753577775 Chr.20: 17531218-17531218 on Build GRCh38
rs749513898 Chr.20: 17531244-17531244 on Build GRCh38
rs777079491 Chr.20: 17531288-17531288 on Build GRCh38
rs773742320 Chr.20: 17531213-17531213 on Build GRCh38
rs971280187 Chr.20: 17531387-17531388 on Build GRCh38
rs867175014 Chr.20: 17531299-17531299 on Build GRCh38
rs549462369 Chr.20: 17531223-17531223 on Build GRCh38
rs762571379 Chr.20: 17531255-17531255 on Build GRCh38
rs1005833334 Chr.20: 17531359-17531359 on Build GRCh38
rs868488668 Chr.20: 17531294-17531294 on Build GRCh38
rs1057355369 Chr.20: 17531119-17531119 on Build GRCh38
rs1030127643 Chr.20: 17531205-17531205 on Build GRCh38
rs569842630 Chr.20: 17531512-17531512 on Build GRCh38
rs1040596803 Chr.20: 17531246-17531246 on Build GRCh38
rs762418779 Chr.20: 17531311-17531311 on Build GRCh38
rs992122571 Chr.20: 17531367-17531367 on Build GRCh38
rs367562045 Chr.20: 17531377-17531377 on Build GRCh38
rs904016466 Chr.20: 17531509-17531509 on Build GRCh38
rs561046384 Chr.20: 17531224-17531224 on Build GRCh38
rs996305718 Chr.20: 17531269-17531269 on Build GRCh38
rs376904264 Chr.20: 17531124-17531124 on Build GRCh38
rs895956223 Chr.20: 17531318-17531318 on Build GRCh38
rs888725539 Chr.20: 17531349-17531349 on Build GRCh38
rs143850362 Chr.20: 17531193-17531193 on Build GRCh38
rs978756517 Chr.20: 17531404-17531404 on Build GRCh38
rs974832705 Chr.20: 17531120-17531120 on Build GRCh38
rs1048803062 Chr.20: 17531413-17531413 on Build GRCh38
rs927245000 Chr.20: 17531447-17531447 on Build GRCh38
rs956682295 Chr.20: 17531343-17531343 on Build GRCh38
rs752363092 Chr.20: 17531310-17531310 on Build GRCh38
rs546576207 Chr.20: 17531339-17531339 on Build GRCh38
rs749458907 Chr.20: 17531108-17531108 on Build GRCh38
rs761225289 Chr.20: 17531276-17531276 on Build GRCh38
rs1005080255 Chr.20: 17531062-17531062 on Build GRCh38
rs1015919822 Chr.20: 17531365-17531365 on Build GRCh38
rs780922338 Chr.20: 17531243-17531243 on Build GRCh38
rs1010510957 Chr.20: 17531391-17531391 on Build GRCh38
rs536850732 Chr.20: 17531526-17531526 on Build GRCh38
rs779194352 Chr.20: 17531258-17531258 on Build GRCh38
rs964462179 Chr.20: 17531194-17531194 on Build GRCh38
rs1020189418 Chr.20: 17531392-17531392 on Build GRCh38
rs895975320 Chr.20: 17531128-17531128 on Build GRCh38
rs1019095682 Chr.20: 17531077-17531077 on Build GRCh38
rs565665888 Chr.20: 17531416-17531416 on Build GRCh38
rs1051062244 Chr.20: 17531508-17531508 on Build GRCh38
rs968657523 Chr.20: 17531402-17531402 on Build GRCh38
rs994396405 Chr.20: 17531146-17531146 on Build GRCh38
rs111913127 Chr.20: 17531368-17531368 on Build GRCh38
rs1039942305 Chr.20: 17531527-17531527 on Build GRCh38
rs1043139559 Chr.20: 17531068-17531068 on Build GRCh38
rs937479628 Chr.20: 17531096-17531096 on Build GRCh38
rs759963940 Chr.20: 17531292-17531292 on Build GRCh38
rs886081089 Chr.20: 17531175-17531175 on Build GRCh38
rs930152908 Chr.20: 17531322-17531322 on Build GRCh38
rs1005864305 Chr.20: 17531177-17531177 on Build GRCh38
rs757116164 Chr.20: 17531328-17531328 on Build GRCh38
rs759199022 Chr.20: 17531219-17531219 on Build GRCh38
rs930412983 Chr.20: 17531405-17531408 on Build GRCh38
rs751524897 Chr.20: 17531324-17531324 on Build GRCh38
rs905972474 Chr.20: 17531069-17531069 on Build GRCh38
rs768926369 Chr.20: 17531255-17531255 on Build GRCh38
rs910130799 Chr.20: 17531520-17531520 on Build GRCh38
rs886038419 Chr.20: 17531223-17531224 on Build GRCh38
rs763606769 Chr.20: 17531323-17531323 on Build GRCh38
rs761942068 Chr.20: 17531197-17531197 on Build GRCh38
rs917344149 Chr.20: 17531482-17531482 on Build GRCh38
rs545346553 Chr.20: 17531147-17531147 on Build GRCh38
rs748220083 Chr.20: 17531260-17531260 on Build GRCh38
rs947352219 Chr.20: 17531227-17531227 on Build GRCh38
rs868032714 Chr.20: 17531257-17531257 on Build GRCh38
rs563918837 Chr.20: 17531153-17531153 on Build GRCh38
rs1030510090 Chr.20: 17531384-17531384 on Build GRCh38
rs953795792 Chr.20: 17531361-17531361 on Build GRCh38

View Full Product Details