Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17591543-17592085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTN
Entrez Gene ID
11034
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
ACTDP, ADF, HEL32, bA462D18.2
UniGene
Hs.304192
Gene Chromosome Location
Chr.20: 17569173-17608242 on Build GRCh38
Transcripts
NM_001011546.1 NM_006870.3 XM_011529142.1 XM_011529143.1 XM_011529144.1

SNP Detail

SNP ID Chromosome Location
rs115150380 Chr.20: 17592040-17592040 on Build GRCh38
rs73258680 Chr.20: 17591779-17591779 on Build GRCh38
rs1009853667 Chr.20: 17591597-17591597 on Build GRCh38
rs762517074 Chr.20: 17591656-17591656 on Build GRCh38
rs541890802 Chr.20: 17591908-17591908 on Build GRCh38
rs901298828 Chr.20: 17591630-17591630 on Build GRCh38
rs535465630 Chr.20: 17591650-17591650 on Build GRCh38
rs6080746 Chr.20: 17592021-17592021 on Build GRCh38
rs974949758 Chr.20: 17591755-17591755 on Build GRCh38
rs889952037 Chr.20: 17592023-17592023 on Build GRCh38
rs964974670 Chr.20: 17591754-17591754 on Build GRCh38
rs759984435 Chr.20: 17591962-17591962 on Build GRCh38
rs1054934375 Chr.20: 17591929-17591929 on Build GRCh38
rs913664595 Chr.20: 17591875-17591875 on Build GRCh38
rs903809096 Chr.20: 17591601-17591601 on Build GRCh38
rs954964666 Chr.20: 17592061-17592061 on Build GRCh38
rs78446128 Chr.20: 17592028-17592028 on Build GRCh38
rs756408424 Chr.20: 17592019-17592019 on Build GRCh38
rs574546458 Chr.20: 17591886-17591886 on Build GRCh38
rs979934033 Chr.20: 17591953-17591953 on Build GRCh38
rs564591924 Chr.20: 17591957-17591957 on Build GRCh38
rs576981317 Chr.20: 17591798-17591798 on Build GRCh38
rs566047826 Chr.20: 17592063-17592063 on Build GRCh38
rs544394180 Chr.20: 17591847-17591847 on Build GRCh38
rs956327596 Chr.20: 17592052-17592052 on Build GRCh38
rs572677892 Chr.20: 17591921-17591921 on Build GRCh38
rs1002048333 Chr.20: 17591613-17591613 on Build GRCh38
rs909369804 Chr.20: 17591698-17591698 on Build GRCh38
rs960952224 Chr.20: 17591626-17591626 on Build GRCh38
rs188312624 Chr.20: 17591667-17591667 on Build GRCh38
rs945114520 Chr.20: 17591896-17591896 on Build GRCh38
rs1026851616 Chr.20: 17591653-17591653 on Build GRCh38
rs6136142 Chr.20: 17591979-17591979 on Build GRCh38
rs77955557 Chr.20: 17591685-17591685 on Build GRCh38
rs940001588 Chr.20: 17591978-17591978 on Build GRCh38
rs998335382 Chr.20: 17591633-17591633 on Build GRCh38
rs556298266 Chr.20: 17591850-17591850 on Build GRCh38
rs949833177 Chr.20: 17591873-17591873 on Build GRCh38
rs955712046 Chr.20: 17591767-17591767 on Build GRCh38
rs1016441281 Chr.20: 17592000-17592000 on Build GRCh38
rs561887320 Chr.20: 17592002-17592002 on Build GRCh38
rs1050780588 Chr.20: 17591797-17591797 on Build GRCh38
rs916993118 Chr.20: 17591781-17591781 on Build GRCh38
rs1043553748 Chr.20: 17591911-17591911 on Build GRCh38
rs546461489 Chr.20: 17591926-17591926 on Build GRCh38
rs930537807 Chr.20: 17591960-17591961 on Build GRCh38
rs1043522524 Chr.20: 17591599-17591599 on Build GRCh38
rs998283150 Chr.20: 17592006-17592006 on Build GRCh38
rs147180239 Chr.20: 17591961-17591961 on Build GRCh38
rs534069232 Chr.20: 17591778-17591778 on Build GRCh38
rs181216651 Chr.20: 17591917-17591917 on Build GRCh38
rs1033660022 Chr.20: 17591617-17591617 on Build GRCh38
rs752990358 Chr.20: 17591995-17591995 on Build GRCh38
rs774062291 Chr.20: 17591754-17591755 on Build GRCh38

View Full Product Details