Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17600921-17601419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTN
Entrez Gene ID
11034
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
ACTDP, ADF, HEL32, bA462D18.2
UniGene
Hs.304192
Gene Chromosome Location
Chr.20: 17569173-17608242 on Build GRCh38
Transcripts
NM_001011546.1 NM_006870.3 XM_011529142.1 XM_011529143.1 XM_011529144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4141345 c.270A>C p.E90D Chr.20: 17601004-17601004 on Build GRCh38
6588340 c.291A>G p.E97E Chr.20: 17601025-17601025 on Build GRCh38
3939267 c.204C>G p.F68L Chr.20: 17600938-17600938 on Build GRCh38
1024796 c.264C>A p.S88R Chr.20: 17600998-17600998 on Build GRCh38
1024795 c.241C>T p.R81C Chr.20: 17600975-17600975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139152501 Chr.20: 17601324-17601324 on Build GRCh38
rs558036055 Chr.20: 17601275-17601275 on Build GRCh38
rs963371803 Chr.20: 17601255-17601255 on Build GRCh38
rs755368695 Chr.20: 17601047-17601047 on Build GRCh38
rs971784898 Chr.20: 17601259-17601259 on Build GRCh38
rs755218253 Chr.20: 17601048-17601048 on Build GRCh38
rs954503609 Chr.20: 17601273-17601273 on Build GRCh38
rs557055543 Chr.20: 17601365-17601365 on Build GRCh38
rs750962255 Chr.20: 17601035-17601035 on Build GRCh38
rs754252238 Chr.20: 17601037-17601037 on Build GRCh38
rs759421994 Chr.20: 17600957-17600957 on Build GRCh38
rs114415913 Chr.20: 17601064-17601064 on Build GRCh38
rs376065996 Chr.20: 17601017-17601017 on Build GRCh38
rs149130905 Chr.20: 17600968-17600968 on Build GRCh38
rs1046206379 Chr.20: 17601098-17601098 on Build GRCh38
rs776649627 Chr.20: 17601097-17601097 on Build GRCh38
rs958905983 Chr.20: 17601214-17601214 on Build GRCh38
rs538998994 Chr.20: 17601266-17601266 on Build GRCh38
rs762059569 Chr.20: 17600969-17600969 on Build GRCh38
rs1008834559 Chr.20: 17601354-17601354 on Build GRCh38
rs528101010 Chr.20: 17600994-17600994 on Build GRCh38
rs79882063 Chr.20: 17601384-17601384 on Build GRCh38
rs906281818 Chr.20: 17601115-17601115 on Build GRCh38
rs779182585 Chr.20: 17601048-17601048 on Build GRCh38
rs915183349 Chr.20: 17601375-17601375 on Build GRCh38
rs777589904 Chr.20: 17601051-17601052 on Build GRCh38
rs760684521 Chr.20: 17601045-17601049 on Build GRCh38
rs746203319 Chr.20: 17600975-17600975 on Build GRCh38
rs73260507 Chr.20: 17601155-17601155 on Build GRCh38
rs374553589 Chr.20: 17601008-17601008 on Build GRCh38
rs1016550399 Chr.20: 17601253-17601253 on Build GRCh38
rs767965259 Chr.20: 17601034-17601035 on Build GRCh38
rs6044900 Chr.20: 17601265-17601265 on Build GRCh38
rs1053625850 Chr.20: 17601374-17601374 on Build GRCh38
rs11907911 Chr.20: 17601104-17601104 on Build GRCh38
rs940555371 Chr.20: 17601120-17601120 on Build GRCh38
rs889143654 Chr.20: 17601351-17601351 on Build GRCh38
rs201824558 Chr.20: 17601276-17601276 on Build GRCh38
rs747816395 Chr.20: 17601055-17601055 on Build GRCh38
rs762287475 Chr.20: 17601034-17601034 on Build GRCh38
rs746571714 Chr.20: 17601086-17601086 on Build GRCh38
rs757138839 Chr.20: 17601058-17601059 on Build GRCh38
rs766386717 Chr.20: 17601046-17601047 on Build GRCh38
rs150970081 Chr.20: 17601259-17601260 on Build GRCh38
rs770754811 Chr.20: 17601378-17601378 on Build GRCh38
rs776571513 Chr.20: 17601335-17601335 on Build GRCh38
rs553766225 Chr.20: 17601250-17601250 on Build GRCh38
rs751357930 Chr.20: 17601056-17601057 on Build GRCh38
rs765071845 Chr.20: 17600959-17600959 on Build GRCh38
rs770323267 Chr.20: 17601090-17601090 on Build GRCh38
rs549702820 Chr.20: 17601065-17601065 on Build GRCh38
rs535202042 Chr.20: 17601229-17601229 on Build GRCh38
rs73260509 Chr.20: 17601225-17601225 on Build GRCh38
rs750572697 Chr.20: 17600985-17600985 on Build GRCh38
rs754073931 Chr.20: 17601047-17601048 on Build GRCh38
rs201891224 Chr.20: 17601265-17601265 on Build GRCh38

View Full Product Details