Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17990263-17990761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGME1
Entrez Gene ID
92667
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, DDK1, MTDPS11, bA504H3.4
UniGene
Hs.320823
Gene Chromosome Location
Chr.20: 17968594-17991122 on Build GRCh38
Transcripts
NM_001310338.1 NM_001310339.1 NM_052865.3 XM_005260867.2 XM_005260870.3 XM_006723663.3 XM_011529394.1 XM_011529395.1 XM_017028127.1 XM_017028128.1

SNP Detail

SNP ID Chromosome Location
rs530099517 Chr.20: 17990566-17990566 on Build GRCh38
rs3748496 Chr.20: 17990413-17990413 on Build GRCh38
rs908835747 Chr.20: 17990644-17990644 on Build GRCh38
rs1004453588 Chr.20: 17990391-17990391 on Build GRCh38
rs552946250 Chr.20: 17990726-17990726 on Build GRCh38
rs887676300 Chr.20: 17990315-17990315 on Build GRCh38
rs986178212 Chr.20: 17990416-17990417 on Build GRCh38
rs1037243061 Chr.20: 17990498-17990498 on Build GRCh38
rs1027306585 Chr.20: 17990382-17990382 on Build GRCh38
rs887739232 Chr.20: 17990425-17990425 on Build GRCh38
rs896765809 Chr.20: 17990422-17990422 on Build GRCh38
rs1031690809 Chr.20: 17990380-17990380 on Build GRCh38
rs202125794 Chr.20: 17990411-17990412 on Build GRCh38
rs545584314 Chr.20: 17990418-17990418 on Build GRCh38
rs956043547 Chr.20: 17990409-17990409 on Build GRCh38
rs1015430261 Chr.20: 17990399-17990399 on Build GRCh38
rs1007433526 Chr.20: 17990316-17990316 on Build GRCh38
rs976130049 Chr.20: 17990361-17990361 on Build GRCh38
rs1036932750 Chr.20: 17990737-17990737 on Build GRCh38
rs763357643 Chr.20: 17990582-17990582 on Build GRCh38
rs560563158 Chr.20: 17990611-17990611 on Build GRCh38
rs3748495 Chr.20: 17990423-17990423 on Build GRCh38
rs1017445775 Chr.20: 17990325-17990325 on Build GRCh38
rs953475078 Chr.20: 17990581-17990581 on Build GRCh38
rs1001850627 Chr.20: 17990365-17990365 on Build GRCh38
rs553436022 Chr.20: 17990313-17990313 on Build GRCh38
rs115922799 Chr.20: 17990620-17990620 on Build GRCh38
rs144655387 Chr.20: 17990547-17990547 on Build GRCh38
rs928824046 Chr.20: 17990606-17990606 on Build GRCh38
rs970340056 Chr.20: 17990576-17990576 on Build GRCh38
rs980279929 Chr.20: 17990414-17990414 on Build GRCh38
rs997379785 Chr.20: 17990670-17990670 on Build GRCh38
rs184976420 Chr.20: 17990629-17990629 on Build GRCh38
rs909564140 Chr.20: 17990684-17990684 on Build GRCh38
rs570912934 Chr.20: 17990667-17990667 on Build GRCh38
rs891920766 Chr.20: 17990725-17990725 on Build GRCh38
rs888406068 Chr.20: 17990387-17990387 on Build GRCh38
rs939908805 Chr.20: 17990711-17990711 on Build GRCh38
rs1007443422 Chr.20: 17990454-17990454 on Build GRCh38
rs547383695 Chr.20: 17990419-17990420 on Build GRCh38
rs192430919 Chr.20: 17990571-17990571 on Build GRCh38
rs901743952 Chr.20: 17990663-17990663 on Build GRCh38
rs571838355 Chr.20: 17990360-17990360 on Build GRCh38
rs773589568 Chr.20: 17990552-17990552 on Build GRCh38
rs1038946862 Chr.20: 17990651-17990651 on Build GRCh38
rs71194213 Chr.20: 17990412-17990413 on Build GRCh38
rs1052651005 Chr.20: 17990308-17990308 on Build GRCh38
rs937668651 Chr.20: 17990422-17990423 on Build GRCh38
rs866147005 Chr.20: 17990424-17990424 on Build GRCh38
rs537608341 Chr.20: 17990410-17990411 on Build GRCh38
rs1048724190 Chr.20: 17990643-17990643 on Build GRCh38
rs369475314 Chr.20: 17990409-17990410 on Build GRCh38
rs543323042 Chr.20: 17990528-17990528 on Build GRCh38
rs6075273 Chr.20: 17990388-17990388 on Build GRCh38
rs576188119 Chr.20: 17990504-17990504 on Build GRCh38
rs939114166 Chr.20: 17990417-17990417 on Build GRCh38
rs566327913 Chr.20: 17990570-17990570 on Build GRCh38
rs1045183711 Chr.20: 17990426-17990426 on Build GRCh38
rs764081989 Chr.20: 17990575-17990575 on Build GRCh38
rs185446685 Chr.20: 17990299-17990299 on Build GRCh38
rs1032345053 Chr.20: 17990585-17990585 on Build GRCh38
rs538456209 Chr.20: 17990691-17990691 on Build GRCh38
rs1021637223 Chr.20: 17990739-17990739 on Build GRCh38
rs774423656 Chr.20: 17990561-17990561 on Build GRCh38
rs1007969501 Chr.20: 17990584-17990584 on Build GRCh38
rs1017455622 Chr.20: 17990513-17990513 on Build GRCh38
rs373212996 Chr.20: 17990351-17990351 on Build GRCh38
rs3748494 Chr.20: 17990630-17990630 on Build GRCh38
rs865789703 Chr.20: 17990412-17990412 on Build GRCh38
rs943278856 Chr.20: 17990645-17990645 on Build GRCh38
rs1046148682 Chr.20: 17990421-17990421 on Build GRCh38
rs928769523 Chr.20: 17990416-17990416 on Build GRCh38
rs990538409 Chr.20: 17990411-17990411 on Build GRCh38
rs897550345 Chr.20: 17990419-17990419 on Build GRCh38
rs958354036 Chr.20: 17990417-17990418 on Build GRCh38

View Full Product Details