Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18383286-18383800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZANK1
Entrez Gene ID
55184
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
UniGene
Hs.472225
Gene Chromosome Location
Chr.20: 18383367-18467185 on Build GRCh38
Transcripts
NM_001099407.1 XM_005260742.3 XM_005260743.3 XM_006723584.2 XM_011529268.2 XM_011529271.2 XM_011529272.2 XM_011529275.2 XM_011529277.2 XM_017027909.1 XM_017027910.1 XM_017027911.1 XM_017027912.1 XM_017027913.1 XM_017027914.1 XM_017027915.1 XM_017027916.1 XM_017027917.1 XM_017027918.1 XM_017027919.1 XM_017027920.1 XM_017027921.1

SNP Detail

SNP ID Chromosome Location
rs79456487 Chr.20: 18383379-18383379 on Build GRCh38
rs978272226 Chr.20: 18383632-18383632 on Build GRCh38
rs963630811 Chr.20: 18383323-18383323 on Build GRCh38
rs958097921 Chr.20: 18383760-18383760 on Build GRCh38
rs923856844 Chr.20: 18383471-18383471 on Build GRCh38
rs564451559 Chr.20: 18383478-18383478 on Build GRCh38
rs878973459 Chr.20: 18383376-18383376 on Build GRCh38
rs939307107 Chr.20: 18383402-18383402 on Build GRCh38
rs776901748 Chr.20: 18383563-18383563 on Build GRCh38
rs1008892654 Chr.20: 18383700-18383700 on Build GRCh38
rs759250425 Chr.20: 18383524-18383525 on Build GRCh38
rs941453738 Chr.20: 18383439-18383439 on Build GRCh38
rs995073306 Chr.20: 18383650-18383650 on Build GRCh38
rs990343513 Chr.20: 18383368-18383368 on Build GRCh38
rs1038430997 Chr.20: 18383764-18383764 on Build GRCh38
rs987574761 Chr.20: 18383403-18383403 on Build GRCh38
rs374873788 Chr.20: 18383639-18383639 on Build GRCh38
rs1005471040 Chr.20: 18383600-18383600 on Build GRCh38
rs967448210 Chr.20: 18383715-18383715 on Build GRCh38
rs762009790 Chr.20: 18383612-18383612 on Build GRCh38
rs765035216 Chr.20: 18383725-18383725 on Build GRCh38
rs889157650 Chr.20: 18383698-18383698 on Build GRCh38
rs531831154 Chr.20: 18383495-18383495 on Build GRCh38
rs776275357 Chr.20: 18383487-18383487 on Build GRCh38
rs928148014 Chr.20: 18383702-18383702 on Build GRCh38
rs957296082 Chr.20: 18383564-18383565 on Build GRCh38
rs966882064 Chr.20: 18383616-18383616 on Build GRCh38
rs541314291 Chr.20: 18383525-18383525 on Build GRCh38
rs1029229164 Chr.20: 18383660-18383660 on Build GRCh38
rs568679187 Chr.20: 18383532-18383532 on Build GRCh38
rs775661383 Chr.20: 18383417-18383417 on Build GRCh38
rs4814740 Chr.20: 18383585-18383585 on Build GRCh38
rs1017890341 Chr.20: 18383550-18383550 on Build GRCh38
rs750839178 Chr.20: 18383726-18383726 on Build GRCh38
rs1030804739 Chr.20: 18383759-18383759 on Build GRCh38
rs965620574 Chr.20: 18383424-18383424 on Build GRCh38
rs547971242 Chr.20: 18383407-18383407 on Build GRCh38
rs896947497 Chr.20: 18383452-18383452 on Build GRCh38
rs187235404 Chr.20: 18383757-18383757 on Build GRCh38
rs995250821 Chr.20: 18383344-18383344 on Build GRCh38
rs182533298 Chr.20: 18383470-18383470 on Build GRCh38
rs574557363 Chr.20: 18383540-18383540 on Build GRCh38
rs138503094 Chr.20: 18383598-18383598 on Build GRCh38
rs527337019 Chr.20: 18383736-18383736 on Build GRCh38
rs1028976929 Chr.20: 18383358-18383358 on Build GRCh38
rs991108992 Chr.20: 18383739-18383739 on Build GRCh38
rs977916336 Chr.20: 18383434-18383434 on Build GRCh38
rs928221481 Chr.20: 18383380-18383380 on Build GRCh38
rs1038895362 Chr.20: 18383450-18383450 on Build GRCh38
rs947705874 Chr.20: 18383533-18383533 on Build GRCh38
rs765607782 Chr.20: 18383637-18383637 on Build GRCh38
rs760842746 Chr.20: 18383474-18383474 on Build GRCh38
rs1017924472 Chr.20: 18383318-18383318 on Build GRCh38
rs1019045425 Chr.20: 18383701-18383701 on Build GRCh38
rs35733501 Chr.20: 18383686-18383687 on Build GRCh38
rs915402768 Chr.20: 18383372-18383372 on Build GRCh38

View Full Product Details