Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20052200-20052701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
UniGene
Hs.729168
Gene Chromosome Location
Chr.20: 20052514-20360714 on Build GRCh38
Transcripts
NM_015585.3 XM_005260688.3 XM_005260689.2 XM_005260690.3 XM_005260691.3 XM_005260692.3 XM_005260693.3 XM_005260697.1 XM_011529211.1 XM_011529212.2 XM_011529213.1 XM_017027786.1 XM_017027787.1 XM_017027788.1 XM_017027789.1 XM_017027790.1 XM_017027795.1 NM_001278625.1 NM_001278626.1 NM_001278627.1 NM_001278628.1 NM_016652.5

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
UniGene
Hs.171342
Gene Chromosome Location
Chr.20: 20034361-20056046 on Build GRCh38
Transcripts
NM_015585.3 XM_005260688.3 XM_005260689.2 XM_005260690.3 XM_005260691.3 XM_005260692.3 XM_005260693.3 XM_005260697.1 XM_011529211.1 XM_011529212.2 XM_011529213.1 XM_017027786.1 XM_017027787.1 XM_017027788.1 XM_017027789.1 XM_017027790.1 XM_017027795.1 NM_001278625.1 NM_001278626.1 NM_001278627.1 NM_001278628.1 NM_016652.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232100 c.305G>A p.G102E Chr.20: 20052521-20052521 on Build GRCh38
5596536 c.310G>A p.G104S Chr.20: 20052516-20052516 on Build GRCh38
3758475 c.228T>A p.V76V Chr.20: 20052598-20052598 on Build GRCh38
6840009 c.502G>A p.A168T Chr.20: 20052324-20052324 on Build GRCh38
5723093 c.320C>T p.S107F Chr.20: 20052506-20052506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751937518 Chr.20: 20052478-20052478 on Build GRCh38
rs750771442 Chr.20: 20052533-20052533 on Build GRCh38
rs774259133 Chr.20: 20052563-20052563 on Build GRCh38
rs755270384 Chr.20: 20052677-20052677 on Build GRCh38
rs550890942 Chr.20: 20052505-20052505 on Build GRCh38
rs781171564 Chr.20: 20052335-20052337 on Build GRCh38
rs1036189250 Chr.20: 20052356-20052356 on Build GRCh38
rs201188951 Chr.20: 20052410-20052410 on Build GRCh38
rs752316306 Chr.20: 20052637-20052637 on Build GRCh38
rs779297090 Chr.20: 20052328-20052328 on Build GRCh38
rs746986628 Chr.20: 20052565-20052566 on Build GRCh38
rs777680922 Chr.20: 20052586-20052586 on Build GRCh38
rs149465617 Chr.20: 20052374-20052374 on Build GRCh38
rs759687119 Chr.20: 20052462-20052462 on Build GRCh38
rs1050359212 Chr.20: 20052560-20052560 on Build GRCh38
rs748479342 Chr.20: 20052331-20052331 on Build GRCh38
rs766788917 Chr.20: 20052574-20052574 on Build GRCh38
rs569431052 Chr.20: 20052520-20052520 on Build GRCh38
rs77988877 Chr.20: 20052468-20052468 on Build GRCh38
rs770071178 Chr.20: 20052559-20052559 on Build GRCh38
rs780325218 Chr.20: 20052465-20052465 on Build GRCh38
rs147964176 Chr.20: 20052496-20052496 on Build GRCh38
rs766122480 Chr.20: 20052546-20052546 on Build GRCh38
rs141785689 Chr.20: 20052596-20052596 on Build GRCh38
rs766402866 Chr.20: 20052390-20052390 on Build GRCh38
rs150473222 Chr.20: 20052528-20052528 on Build GRCh38
rs749271930 Chr.20: 20052448-20052448 on Build GRCh38
rs762438664 Chr.20: 20052661-20052661 on Build GRCh38
rs778892952 Chr.20: 20052500-20052500 on Build GRCh38
rs201618370 Chr.20: 20052396-20052396 on Build GRCh38
rs754934321 Chr.20: 20052498-20052498 on Build GRCh38
rs188266043 Chr.20: 20052401-20052401 on Build GRCh38
rs191096342 Chr.20: 20052531-20052531 on Build GRCh38
rs1053925392 Chr.20: 20052233-20052233 on Build GRCh38
rs748147610 Chr.20: 20052552-20052552 on Build GRCh38
rs776812300 Chr.20: 20052625-20052625 on Build GRCh38
rs777682651 Chr.20: 20052555-20052555 on Build GRCh38
rs755440432 Chr.20: 20052322-20052322 on Build GRCh38
rs765238125 Chr.20: 20052429-20052429 on Build GRCh38
rs755883285 Chr.20: 20052535-20052535 on Build GRCh38
rs867115962 Chr.20: 20052490-20052490 on Build GRCh38
rs2255255 Chr.20: 20052354-20052354 on Build GRCh38
rs771112536 Chr.20: 20052461-20052461 on Build GRCh38
rs200178595 Chr.20: 20052668-20052676 on Build GRCh38
rs746772587 Chr.20: 20052557-20052557 on Build GRCh38
rs373695701 Chr.20: 20052382-20052382 on Build GRCh38
rs761193049 Chr.20: 20052622-20052622 on Build GRCh38
rs989589288 Chr.20: 20052665-20052665 on Build GRCh38
rs757605082 Chr.20: 20052286-20052286 on Build GRCh38
rs372828683 Chr.20: 20052471-20052471 on Build GRCh38
rs1044246126 Chr.20: 20052419-20052419 on Build GRCh38
rs201634961 Chr.20: 20052469-20052469 on Build GRCh38
rs762160996 Chr.20: 20052529-20052529 on Build GRCh38
rs148603082 Chr.20: 20052415-20052415 on Build GRCh38
rs978279355 Chr.20: 20052360-20052360 on Build GRCh38
rs372474436 Chr.20: 20052501-20052501 on Build GRCh38
rs982161407 Chr.20: 20052571-20052571 on Build GRCh38
rs753117976 Chr.20: 20052399-20052399 on Build GRCh38
rs760886113 Chr.20: 20052386-20052386 on Build GRCh38
rs369544240 Chr.20: 20052370-20052370 on Build GRCh38
rs780124915 Chr.20: 20052301-20052301 on Build GRCh38
rs766589968 Chr.20: 20052424-20052424 on Build GRCh38
rs745429919 Chr.20: 20052438-20052438 on Build GRCh38
rs756134981 Chr.20: 20052296-20052296 on Build GRCh38
rs774158149 Chr.20: 20052421-20052421 on Build GRCh38
rs2273057 Chr.20: 20052598-20052598 on Build GRCh38
rs756247332 Chr.20: 20052443-20052444 on Build GRCh38
rs140622884 Chr.20: 20052508-20052508 on Build GRCh38
rs768591710 Chr.20: 20052449-20052449 on Build GRCh38
rs757058902 Chr.20: 20052588-20052588 on Build GRCh38
rs2255258 Chr.20: 20052493-20052493 on Build GRCh38
rs867110263 Chr.20: 20052338-20052338 on Build GRCh38
rs771688473 Chr.20: 20052243-20052243 on Build GRCh38
rs540598140 Chr.20: 20052274-20052274 on Build GRCh38
rs775559150 Chr.20: 20052411-20052411 on Build GRCh38
rs1002828352 Chr.20: 20052236-20052236 on Build GRCh38
rs771908967 Chr.20: 20052515-20052515 on Build GRCh38
rs780618922 Chr.20: 20052404-20052404 on Build GRCh38
rs749766098 Chr.20: 20052319-20052319 on Build GRCh38
rs759448633 Chr.20: 20052269-20052269 on Build GRCh38
rs761661820 Chr.20: 20052459-20052459 on Build GRCh38
rs751416259 Chr.20: 20052587-20052587 on Build GRCh38
rs767820162 Chr.20: 20052288-20052288 on Build GRCh38
rs773909827 Chr.20: 20052330-20052330 on Build GRCh38
rs142988900 Chr.20: 20052672-20052672 on Build GRCh38
rs141746930 Chr.20: 20052290-20052290 on Build GRCh38
rs763086392 Chr.20: 20052522-20052522 on Build GRCh38
rs780899904 Chr.20: 20052593-20052593 on Build GRCh38
rs760099136 Chr.20: 20052251-20052251 on Build GRCh38
rs771448769 Chr.20: 20052623-20052623 on Build GRCh38
rs772726100 Chr.20: 20052349-20052349 on Build GRCh38
rs779736792 Chr.20: 20052494-20052494 on Build GRCh38
rs144315980 Chr.20: 20052431-20052431 on Build GRCh38
rs145079188 Chr.20: 20052554-20052554 on Build GRCh38
rs148307781 Chr.20: 20052551-20052551 on Build GRCh38
rs770494524 Chr.20: 20052240-20052240 on Build GRCh38
rs758771266 Chr.20: 20052400-20052400 on Build GRCh38
rs753929976 Chr.20: 20052394-20052394 on Build GRCh38
rs377334990 Chr.20: 20052261-20052261 on Build GRCh38
rs770565261 Chr.20: 20052403-20052403 on Build GRCh38
rs774052514 Chr.20: 20052454-20052454 on Build GRCh38
rs748148401 Chr.20: 20052606-20052606 on Build GRCh38
rs143884554 Chr.20: 20052407-20052407 on Build GRCh38
rs958218587 Chr.20: 20052647-20052647 on Build GRCh38
rs938506015 Chr.20: 20052422-20052422 on Build GRCh38
rs557483220 Chr.20: 20052671-20052671 on Build GRCh38
rs764568104 Chr.20: 20052525-20052525 on Build GRCh38
rs530981737 Chr.20: 20052398-20052398 on Build GRCh38
rs771778103 Chr.20: 20052347-20052347 on Build GRCh38
rs757412467 Chr.20: 20052259-20052259 on Build GRCh38
rs74404969 Chr.20: 20052678-20052678 on Build GRCh38
rs766393616 Chr.20: 20052253-20052253 on Build GRCh38
rs139192640 Chr.20: 20052371-20052371 on Build GRCh38
rs762409299 Chr.20: 20052564-20052564 on Build GRCh38
rs936922928 Chr.20: 20052227-20052227 on Build GRCh38
rs147785600 Chr.20: 20052436-20052436 on Build GRCh38
rs752125657 Chr.20: 20052585-20052585 on Build GRCh38
rs145198573 Chr.20: 20052550-20052550 on Build GRCh38
rs765151718 Chr.20: 20052277-20052277 on Build GRCh38
rs529488737 Chr.20: 20052666-20052666 on Build GRCh38
rs747875764 Chr.20: 20052490-20052499 on Build GRCh38
rs745343930 Chr.20: 20052406-20052406 on Build GRCh38
rs759595000 Chr.20: 20052425-20052425 on Build GRCh38
rs773755524 Chr.20: 20052569-20052569 on Build GRCh38
rs1023869790 Chr.20: 20052669-20052669 on Build GRCh38
rs916589352 Chr.20: 20052262-20052262 on Build GRCh38
rs751504038 Chr.20: 20052668-20052668 on Build GRCh38
rs545269988 Chr.20: 20052619-20052619 on Build GRCh38
rs764825513 Chr.20: 20052581-20052581 on Build GRCh38
rs916636693 Chr.20: 20052381-20052381 on Build GRCh38
rs2273056 Chr.20: 20052579-20052579 on Build GRCh38
rs768785835 Chr.20: 20052562-20052562 on Build GRCh38
rs775584333 Chr.20: 20052516-20052516 on Build GRCh38
rs772951381 Chr.20: 20052504-20052504 on Build GRCh38
rs764763818 Chr.20: 20052633-20052633 on Build GRCh38
rs779648844 Chr.20: 20052442-20052442 on Build GRCh38
rs889025020 Chr.20: 20052576-20052576 on Build GRCh38
rs750639927 Chr.20: 20052442-20052443 on Build GRCh38
rs759281207 Chr.20: 20052627-20052627 on Build GRCh38
rs2273058 Chr.20: 20052675-20052675 on Build GRCh38
rs1033146736 Chr.20: 20052645-20052645 on Build GRCh38
rs969604281 Chr.20: 20052670-20052670 on Build GRCh38
rs762926211 Chr.20: 20052417-20052417 on Build GRCh38
rs778019592 Chr.20: 20052433-20052433 on Build GRCh38
rs377204100 Chr.20: 20052575-20052575 on Build GRCh38
rs749414547 Chr.20: 20052561-20052561 on Build GRCh38
rs777592703 Chr.20: 20052556-20052556 on Build GRCh38
rs761105056 Chr.20: 20052572-20052572 on Build GRCh38
rs772748103 Chr.20: 20052246-20052246 on Build GRCh38
rs772687775 Chr.20: 20052565-20052565 on Build GRCh38
rs755517643 Chr.20: 20052395-20052395 on Build GRCh38
rs769446966 Chr.20: 20052440-20052440 on Build GRCh38
rs867662867 Chr.20: 20052450-20052450 on Build GRCh38
rs767166892 Chr.20: 20052491-20052491 on Build GRCh38
rs779101364 Chr.20: 20052599-20052599 on Build GRCh38
rs372675083 Chr.20: 20052377-20052377 on Build GRCh38
rs370706858 Chr.20: 20052607-20052607 on Build GRCh38
rs370311079 Chr.20: 20052279-20052279 on Build GRCh38

View Full Product Details